Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 101391367-101391870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPL36A
Entrez Gene ID
6173
Gene Name
ribosomal protein L36a
Gene Aliases
L36A, L44L, MIG6, RPL44
UniGene
Hs.432485
Gene Chromosome Location
Chr.X: 101390890-101396154 on Build GRCh38
Transcripts
NM_001199972.1 NM_021029.5 NM_001199973.1 NM_001199974.1

Gene Symbol
RPL36A-HNRNPH2
Entrez Gene ID
100529097
Gene Name
RPL36A-HNRNPH2 readthrough
Gene Aliases
-
UniGene
Hs.432485
Gene Chromosome Location
Chr.X: 101390890-101414140 on Build GRCh38
Transcripts
NM_001199972.1 NM_021029.5 NM_001199973.1 NM_001199974.1

SNP Detail

SNP ID Chromosome Location
rs1046512992 Chr.X: 101391630-101391630 on Build GRCh38
rs376149766 Chr.X: 101391712-101391712 on Build GRCh38
rs369758384 Chr.X: 101391426-101391426 on Build GRCh38
rs931236792 Chr.X: 101391581-101391581 on Build GRCh38
rs375882151 Chr.X: 101391846-101391846 on Build GRCh38
rs781895008 Chr.X: 101391768-101391768 on Build GRCh38
rs781956801 Chr.X: 101391835-101391836 on Build GRCh38
rs782666451 Chr.X: 101391568-101391568 on Build GRCh38
rs370531812 Chr.X: 101391764-101391764 on Build GRCh38
rs781844897 Chr.X: 101391443-101391443 on Build GRCh38
rs999684471 Chr.X: 101391661-101391661 on Build GRCh38
rs139329988 Chr.X: 101391625-101391625 on Build GRCh38
rs782761522 Chr.X: 101391792-101391792 on Build GRCh38
rs782061794 Chr.X: 101391412-101391412 on Build GRCh38
rs782273995 Chr.X: 101391593-101391593 on Build GRCh38
rs373953678 Chr.X: 101391836-101391836 on Build GRCh38
rs986518374 Chr.X: 101391390-101391390 on Build GRCh38
rs3027596 Chr.X: 101391602-101391602 on Build GRCh38
rs782356566 Chr.X: 101391817-101391817 on Build GRCh38
rs1054385580 Chr.X: 101391705-101391705 on Build GRCh38
rs142755460 Chr.X: 101391405-101391405 on Build GRCh38
rs782102007 Chr.X: 101391713-101391713 on Build GRCh38
rs782260191 Chr.X: 101391583-101391584 on Build GRCh38
rs973007509 Chr.X: 101391456-101391456 on Build GRCh38
rs374093946 Chr.X: 101391584-101391584 on Build GRCh38
rs34249618 Chr.X: 101391749-101391750 on Build GRCh38
rs782323425 Chr.X: 101391577-101391577 on Build GRCh38
rs781946083 Chr.X: 101391738-101391738 on Build GRCh38
rs146187652 Chr.X: 101391539-101391539 on Build GRCh38
rs782492796 Chr.X: 101391553-101391553 on Build GRCh38
rs782133871 Chr.X: 101391440-101391440 on Build GRCh38
rs782048174 Chr.X: 101391758-101391758 on Build GRCh38
rs782205827 Chr.X: 101391576-101391576 on Build GRCh38
rs782549416 Chr.X: 101391844-101391844 on Build GRCh38
rs3027595 Chr.X: 101391786-101391786 on Build GRCh38
rs782309344 Chr.X: 101391422-101391422 on Build GRCh38
rs781783917 Chr.X: 101391837-101391837 on Build GRCh38
rs782432620 Chr.X: 101391441-101391441 on Build GRCh38
rs902630239 Chr.X: 101391641-101391641 on Build GRCh38
rs782331196 Chr.X: 101391795-101391795 on Build GRCh38
rs782569704 Chr.X: 101391653-101391653 on Build GRCh38
rs782088988 Chr.X: 101391449-101391449 on Build GRCh38
rs949410016 Chr.X: 101391629-101391629 on Build GRCh38
rs782345921 Chr.X: 101391719-101391719 on Build GRCh38
rs782671537 Chr.X: 101391515-101391515 on Build GRCh38
rs782556598 Chr.X: 101391505-101391505 on Build GRCh38
rs782479617 Chr.X: 101391814-101391814 on Build GRCh38
rs782717175 Chr.X: 101391464-101391464 on Build GRCh38
rs781792177 Chr.X: 101391474-101391474 on Build GRCh38

View Full Product Details