Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 9567225-9567794 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC25A33
Entrez Gene ID
84275
Gene Name
solute carrier family 25 member 33
Gene Aliases
BMSC-MCP, PNC1
UniGene
Hs.568613
Gene Chromosome Location
Chr.1: 9539469-9582773 on Build GRCh38
Transcripts
NM_032315.2 XM_005263503.3 XM_011542296.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6780703 c.255G>T p.E85D Chr.1: 9567302-9567302 on Build GRCh38
5577053 c.286C>T p.P96S Chr.1: 9567333-9567333 on Build GRCh38
2260994 c.283G>A p.G95S Chr.1: 9567330-9567330 on Build GRCh38
6780704 c.263A>G p.K88R Chr.1: 9567310-9567310 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1019029121 Chr.1: 9567755-9567755 on Build GRCh38
rs776826348 Chr.1: 9567312-9567312 on Build GRCh38
rs369567909 Chr.1: 9567389-9567389 on Build GRCh38
rs779264868 Chr.1: 9567386-9567424 on Build GRCh38
rs762265928 Chr.1: 9567356-9567356 on Build GRCh38
rs776307626 Chr.1: 9567530-9567530 on Build GRCh38
rs370922985 Chr.1: 9567470-9567470 on Build GRCh38
rs564942915 Chr.1: 9567756-9567756 on Build GRCh38
rs542193831 Chr.1: 9567658-9567658 on Build GRCh38
rs554008278 Chr.1: 9567522-9567522 on Build GRCh38
rs778522338 Chr.1: 9567299-9567299 on Build GRCh38
rs764804627 Chr.1: 9567555-9567555 on Build GRCh38
rs149976338 Chr.1: 9567355-9567355 on Build GRCh38
rs528186150 Chr.1: 9567401-9567401 on Build GRCh38
rs769700549 Chr.1: 9567329-9567329 on Build GRCh38
rs984560425 Chr.1: 9567603-9567604 on Build GRCh38
rs555928367 Chr.1: 9567452-9567452 on Build GRCh38
rs1022944050 Chr.1: 9567595-9567595 on Build GRCh38
rs755690700 Chr.1: 9567385-9567385 on Build GRCh38
rs759886615 Chr.1: 9567441-9567441 on Build GRCh38
rs532299548 Chr.1: 9567764-9567764 on Build GRCh38
rs959645915 Chr.1: 9567260-9567260 on Build GRCh38
rs6698575 Chr.1: 9567463-9567463 on Build GRCh38
rs181146587 Chr.1: 9567368-9567368 on Build GRCh38
rs756327632 Chr.1: 9567417-9567417 on Build GRCh38
rs151258346 Chr.1: 9567343-9567343 on Build GRCh38
rs376335225 Chr.1: 9567377-9567377 on Build GRCh38
rs34733498 Chr.1: 9567340-9567341 on Build GRCh38
rs760233421 Chr.1: 9567514-9567514 on Build GRCh38
rs780986133 Chr.1: 9567261-9567261 on Build GRCh38
rs114935495 Chr.1: 9567726-9567726 on Build GRCh38
rs1011137736 Chr.1: 9567568-9567568 on Build GRCh38
rs185403207 Chr.1: 9567519-9567519 on Build GRCh38
rs780054403 Chr.1: 9567276-9567276 on Build GRCh38
rs1034348491 Chr.1: 9567251-9567251 on Build GRCh38
rs755549236 Chr.1: 9567301-9567301 on Build GRCh38
rs534156437 Chr.1: 9567433-9567433 on Build GRCh38
rs200821207 Chr.1: 9567305-9567305 on Build GRCh38
rs948571426 Chr.1: 9567381-9567381 on Build GRCh38
rs764643143 Chr.1: 9567352-9567352 on Build GRCh38
rs776099808 Chr.1: 9567330-9567330 on Build GRCh38
rs768526283 Chr.1: 9567287-9567287 on Build GRCh38
rs571729047 Chr.1: 9567605-9567605 on Build GRCh38
rs771181070 Chr.1: 9567311-9567311 on Build GRCh38
rs767337678 Chr.1: 9567364-9567364 on Build GRCh38
rs560683312 Chr.1: 9567663-9567663 on Build GRCh38
rs373847848 Chr.1: 9567611-9567611 on Build GRCh38
rs201608198 Chr.1: 9567274-9567274 on Build GRCh38
rs576047092 Chr.1: 9567696-9567696 on Build GRCh38
rs111904501 Chr.1: 9567590-9567590 on Build GRCh38
rs1012202934 Chr.1: 9567451-9567451 on Build GRCh38
rs965432215 Chr.1: 9567527-9567527 on Build GRCh38
rs761473866 Chr.1: 9567531-9567531 on Build GRCh38
rs146265200 Chr.1: 9567505-9567505 on Build GRCh38
rs1049392208 Chr.1: 9567449-9567449 on Build GRCh38
rs754918928 Chr.1: 9567403-9567403 on Build GRCh38
rs560832261 Chr.1: 9567487-9567487 on Build GRCh38
rs918615836 Chr.1: 9567326-9567326 on Build GRCh38
rs531002667 Chr.1: 9567744-9567744 on Build GRCh38
rs750225976 Chr.1: 9567714-9567714 on Build GRCh38
rs373779347 Chr.1: 9567286-9567286 on Build GRCh38
rs921879687 Chr.1: 9567431-9567431 on Build GRCh38
rs750021556 Chr.1: 9567265-9567265 on Build GRCh38
rs747914396 Chr.1: 9567309-9567309 on Build GRCh38
rs532308777 Chr.1: 9567341-9567341 on Build GRCh38
rs906912760 Chr.1: 9567517-9567517 on Build GRCh38
rs1002594695 Chr.1: 9567422-9567422 on Build GRCh38

View Full Product Details