Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32040175-32040723 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP21A2
Entrez Gene ID
1589
Gene Name
cytochrome P450 family 21 subfamily A member 2
Gene Aliases
CA21H, CAH1, CPS1, CYP21, CYP21B, P450c21B
UniGene
Hs.654479
Gene Chromosome Location
Chr.6: 32038316-32041670 on Build GRCh38
Transcripts
NM_000500.7 NM_001128590.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3624999 c.1144G>A p.G382S Chr.6: 32040693-32040693 on Build GRCh38
3624998 c.1007C>T p.P336L Chr.6: 32040473-32040473 on Build GRCh38
1131914 c.1024C>T p.R342W Chr.6: 32040490-32040490 on Build GRCh38
1077771 c.1132G>A p.D378N Chr.6: 32040681-32040681 on Build GRCh38
3624997 c.978G>A p.L326L Chr.6: 32040444-32040444 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs72552753 Chr.6: 32040207-32040207 on Build GRCh38
rs773218988 Chr.6: 32040438-32040438 on Build GRCh38
rs754209599 Chr.6: 32040475-32040475 on Build GRCh38
rs747079101 Chr.6: 32040491-32040491 on Build GRCh38
rs373579128 Chr.6: 32040541-32040541 on Build GRCh38
rs750984938 Chr.6: 32040257-32040257 on Build GRCh38
rs374872656 Chr.6: 32040217-32040217 on Build GRCh38
rs528524868 Chr.6: 32040681-32040681 on Build GRCh38
rs921238920 Chr.6: 32040320-32040320 on Build GRCh38
rs758658540 Chr.6: 32040565-32040565 on Build GRCh38
rs778895502 Chr.6: 32040585-32040585 on Build GRCh38
rs771912317 Chr.6: 32040588-32040588 on Build GRCh38
rs775304709 Chr.6: 32040231-32040231 on Build GRCh38
rs780316540 Chr.6: 32040380-32040380 on Build GRCh38
rs370908729 Chr.6: 32040512-32040512 on Build GRCh38
rs139392370 Chr.6: 32040477-32040477 on Build GRCh38
rs763782360 Chr.6: 32040239-32040239 on Build GRCh38
rs753107694 Chr.6: 32040621-32040621 on Build GRCh38
rs780412947 Chr.6: 32040209-32040209 on Build GRCh38
rs764955614 Chr.6: 32040224-32040224 on Build GRCh38
rs563109642 Chr.6: 32040591-32040591 on Build GRCh38
rs369087577 Chr.6: 32040211-32040211 on Build GRCh38
rs376421504 Chr.6: 32040575-32040575 on Build GRCh38
rs1027588145 Chr.6: 32040582-32040582 on Build GRCh38
rs775951156 Chr.6: 32040594-32040594 on Build GRCh38
rs766779361 Chr.6: 32040237-32040237 on Build GRCh38
rs759899014 Chr.6: 32040540-32040540 on Build GRCh38
rs548446169 Chr.6: 32040484-32040484 on Build GRCh38
rs6443 Chr.6: 32040291-32040291 on Build GRCh38
rs756621561 Chr.6: 32040467-32040467 on Build GRCh38
rs770483467 Chr.6: 32040593-32040593 on Build GRCh38
rs759398534 Chr.6: 32040223-32040223 on Build GRCh38
rs6461 Chr.6: 32040634-32040634 on Build GRCh38
rs774151904 Chr.6: 32040455-32040455 on Build GRCh38
rs772910112 Chr.6: 32040589-32040589 on Build GRCh38
rs770059546 Chr.6: 32040197-32040197 on Build GRCh38
rs537111179 Chr.6: 32040233-32040233 on Build GRCh38
rs557564791 Chr.6: 32040210-32040210 on Build GRCh38
rs574370139 Chr.6: 32040536-32040536 on Build GRCh38
rs774242376 Chr.6: 32040518-32040518 on Build GRCh38
rs151344506 Chr.6: 32040675-32040675 on Build GRCh38
rs146473100 Chr.6: 32040519-32040519 on Build GRCh38
rs760216630 Chr.6: 32040530-32040530 on Build GRCh38
rs7769409 Chr.6: 32040535-32040535 on Build GRCh38
rs991620424 Chr.6: 32040205-32040205 on Build GRCh38
rs6469 Chr.6: 32040674-32040674 on Build GRCh38
rs7755898 Chr.6: 32040421-32040421 on Build GRCh38
rs564419538 Chr.6: 32040640-32040640 on Build GRCh38
rs142058202 Chr.6: 32040433-32040433 on Build GRCh38
rs755201790 Chr.6: 32040388-32040388 on Build GRCh38
rs756824528 Chr.6: 32040255-32040255 on Build GRCh38
rs6442 Chr.6: 32040216-32040216 on Build GRCh38
rs144104274 Chr.6: 32040516-32040516 on Build GRCh38
rs891233264 Chr.6: 32040448-32040448 on Build GRCh38
rs756598451 Chr.6: 32040357-32040357 on Build GRCh38
rs769730855 Chr.6: 32040445-32040445 on Build GRCh38
rs368463249 Chr.6: 32040459-32040459 on Build GRCh38
rs766860260 Chr.6: 32040531-32040531 on Build GRCh38
rs747936478 Chr.6: 32040587-32040587 on Build GRCh38
rs781074931 Chr.6: 32040574-32040574 on Build GRCh38
rs765702263 Chr.6: 32040598-32040598 on Build GRCh38
rs767161968 Chr.6: 32040460-32040460 on Build GRCh38
rs771822460 Chr.6: 32040520-32040520 on Build GRCh38
rs778983393 Chr.6: 32040683-32040683 on Build GRCh38
rs749635628 Chr.6: 32040382-32040382 on Build GRCh38
rs6444 Chr.6: 32040296-32040296 on Build GRCh38
rs745826667 Chr.6: 32040440-32040440 on Build GRCh38
rs72552754 Chr.6: 32040485-32040485 on Build GRCh38
rs536088585 Chr.6: 32040200-32040200 on Build GRCh38
rs72552755 Chr.6: 32040490-32040490 on Build GRCh38
rs768586475 Chr.6: 32040517-32040517 on Build GRCh38
rs1027284604 Chr.6: 32040196-32040196 on Build GRCh38
rs767069613 Chr.6: 32040256-32040256 on Build GRCh38
rs754380599 Chr.6: 32040373-32040373 on Build GRCh38
rs755228276 Chr.6: 32040657-32040658 on Build GRCh38
rs376415981 Chr.6: 32040406-32040406 on Build GRCh38
rs72552756 Chr.6: 32040692-32040692 on Build GRCh38
rs748290896 Chr.6: 32040415-32040415 on Build GRCh38
rs763722637 Chr.6: 32040453-32040453 on Build GRCh38
rs376035565 Chr.6: 32040566-32040566 on Build GRCh38
rs774336886 Chr.6: 32040215-32040215 on Build GRCh38
rs540662963 Chr.6: 32040493-32040493 on Build GRCh38
rs932682037 Chr.6: 32040374-32040374 on Build GRCh38
rs530916937 Chr.6: 32040597-32040597 on Build GRCh38
rs150804717 Chr.6: 32040439-32040439 on Build GRCh38
rs772900496 Chr.6: 32040529-32040529 on Build GRCh38
rs749917058 Chr.6: 32040466-32040466 on Build GRCh38
rs751174502 Chr.6: 32040249-32040249 on Build GRCh38
rs776542176 Chr.6: 32040220-32040220 on Build GRCh38
rs377484718 Chr.6: 32040447-32040447 on Build GRCh38
rs764378215 Chr.6: 32040629-32040629 on Build GRCh38
rs750443304 Chr.6: 32040680-32040680 on Build GRCh38
rs766890396 Chr.6: 32040471-32040471 on Build GRCh38
rs779090292 Chr.6: 32040393-32040393 on Build GRCh38

View Full Product Details