Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 28021913-28022481 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUGGC
Entrez Gene ID
389643
Gene Name
nuclear GTPase, germinal center associated
Gene Aliases
C8orf80, HMFN0672, SLIP-GC
UniGene
Hs.370129
Gene Chromosome Location
Chr.8: 28021964-28084068 on Build GRCh38
Transcripts
NM_001010906.1 XM_011544523.2 XM_011544524.2 XM_011544525.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3785944 c.2406C>T p.D802D Chr.8: 28022195-28022195 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs28729520 Chr.8: 28022159-28022159 on Build GRCh38
rs866156560 Chr.8: 28022262-28022262 on Build GRCh38
rs772866878 Chr.8: 28022062-28022062 on Build GRCh38
rs959340333 Chr.8: 28022073-28022073 on Build GRCh38
rs7459596 Chr.8: 28022020-28022020 on Build GRCh38
rs538292425 Chr.8: 28022121-28022121 on Build GRCh38
rs867668428 Chr.8: 28022133-28022133 on Build GRCh38
rs535311563 Chr.8: 28021975-28021975 on Build GRCh38
rs1049580757 Chr.8: 28021978-28021978 on Build GRCh38
rs145983334 Chr.8: 28022404-28022404 on Build GRCh38
rs113432190 Chr.8: 28022143-28022143 on Build GRCh38
rs143257376 Chr.8: 28022209-28022218 on Build GRCh38
rs890916858 Chr.8: 28022005-28022005 on Build GRCh38
rs754454687 Chr.8: 28022205-28022206 on Build GRCh38
rs919711170 Chr.8: 28022349-28022349 on Build GRCh38
rs533915470 Chr.8: 28022374-28022374 on Build GRCh38
rs892485503 Chr.8: 28022021-28022021 on Build GRCh38
rs1053387354 Chr.8: 28022144-28022144 on Build GRCh38
rs971310904 Chr.8: 28022151-28022151 on Build GRCh38
rs35977212 Chr.8: 28022176-28022176 on Build GRCh38
rs552358277 Chr.8: 28022120-28022120 on Build GRCh38
rs556309709 Chr.8: 28022421-28022421 on Build GRCh38
rs1026620661 Chr.8: 28022035-28022035 on Build GRCh38
rs984620706 Chr.8: 28022433-28022433 on Build GRCh38
rs113806414 Chr.8: 28022206-28022215 on Build GRCh38
rs199620008 Chr.8: 28022131-28022132 on Build GRCh38
rs919020194 Chr.8: 28022389-28022389 on Build GRCh38
rs1022073527 Chr.8: 28021952-28021952 on Build GRCh38
rs531480818 Chr.8: 28022163-28022163 on Build GRCh38
rs1032370879 Chr.8: 28022068-28022068 on Build GRCh38
rs192016370 Chr.8: 28022400-28022400 on Build GRCh38
rs961062791 Chr.8: 28022224-28022224 on Build GRCh38
rs932423436 Chr.8: 28022366-28022366 on Build GRCh38
rs903928144 Chr.8: 28022098-28022098 on Build GRCh38
rs567951241 Chr.8: 28022316-28022316 on Build GRCh38
rs182751863 Chr.8: 28022405-28022405 on Build GRCh38
rs923277388 Chr.8: 28022140-28022140 on Build GRCh38
rs929043355 Chr.8: 28022398-28022398 on Build GRCh38
rs1033344938 Chr.8: 28022019-28022019 on Build GRCh38
rs750630306 Chr.8: 28022208-28022217 on Build GRCh38
rs541794052 Chr.8: 28022145-28022145 on Build GRCh38
rs1015828447 Chr.8: 28022161-28022161 on Build GRCh38
rs370322563 Chr.8: 28022457-28022457 on Build GRCh38
rs975185394 Chr.8: 28022122-28022122 on Build GRCh38
rs11136025 Chr.8: 28021953-28021953 on Build GRCh38
rs187545263 Chr.8: 28022009-28022009 on Build GRCh38
rs368831624 Chr.8: 28022283-28022283 on Build GRCh38
rs984550104 Chr.8: 28022065-28022065 on Build GRCh38
rs1033788678 Chr.8: 28022385-28022385 on Build GRCh38
rs972886692 Chr.8: 28022157-28022157 on Build GRCh38
rs1013645275 Chr.8: 28022401-28022401 on Build GRCh38
rs187889500 Chr.8: 28022434-28022434 on Build GRCh38
rs532333868 Chr.8: 28022107-28022107 on Build GRCh38
rs548186074 Chr.8: 28022261-28022261 on Build GRCh38
rs919675691 Chr.8: 28021964-28021964 on Build GRCh38
rs765593676 Chr.8: 28022207-28022216 on Build GRCh38
rs564046944 Chr.8: 28022158-28022158 on Build GRCh38
rs528772563 Chr.8: 28021977-28021977 on Build GRCh38
rs903639383 Chr.8: 28022315-28022315 on Build GRCh38
rs541516224 Chr.8: 28022030-28022030 on Build GRCh38
rs1033853772 Chr.8: 28022105-28022106 on Build GRCh38
rs781163654 Chr.8: 28022197-28022197 on Build GRCh38
rs1043215693 Chr.8: 28022287-28022287 on Build GRCh38
rs933594486 Chr.8: 28022142-28022142 on Build GRCh38
rs997549 Chr.8: 28022078-28022078 on Build GRCh38

View Full Product Details