Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 27977948-27978509 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DCAF8L1
Entrez Gene ID
139425
Gene Name
DDB1 and CUL4 associated factor 8 like 1
Gene Aliases
WDR42B
UniGene
Hs.447365
Gene Chromosome Location
Chr.X: 27977993-27981449 on Build GRCh38
Transcripts
NM_001017930.1

SNP Detail

SNP ID Chromosome Location
rs1004451559 Chr.X: 27978266-27978266 on Build GRCh38
rs906792647 Chr.X: 27978422-27978422 on Build GRCh38
rs779503300 Chr.X: 27978094-27978094 on Build GRCh38
rs776567997 Chr.X: 27978469-27978469 on Build GRCh38
rs755292381 Chr.X: 27978276-27978276 on Build GRCh38
rs62590483 Chr.X: 27978407-27978407 on Build GRCh38
rs562583726 Chr.X: 27978079-27978079 on Build GRCh38
rs887303391 Chr.X: 27978246-27978246 on Build GRCh38
rs1002178761 Chr.X: 27978464-27978464 on Build GRCh38
rs745483488 Chr.X: 27978302-27978302 on Build GRCh38
rs73628870 Chr.X: 27978184-27978184 on Build GRCh38
rs371729408 Chr.X: 27978072-27978072 on Build GRCh38
rs769294755 Chr.X: 27978485-27978485 on Build GRCh38
rs1046660383 Chr.X: 27978294-27978295 on Build GRCh38
rs745802702 Chr.X: 27978482-27978482 on Build GRCh38
rs770992554 Chr.X: 27978426-27978426 on Build GRCh38
rs769384179 Chr.X: 27978322-27978322 on Build GRCh38
rs145254507 Chr.X: 27978310-27978310 on Build GRCh38
rs182641406 Chr.X: 27978077-27978077 on Build GRCh38
rs751958744 Chr.X: 27977985-27977985 on Build GRCh38
rs190937563 Chr.X: 27978031-27978031 on Build GRCh38
rs780020112 Chr.X: 27978163-27978163 on Build GRCh38
rs1034101494 Chr.X: 27978481-27978481 on Build GRCh38

View Full Product Details