Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 48801-49366 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TUBB8
Entrez Gene ID
347688
Gene Name
tubulin beta 8 class VIII
Gene Aliases
OOMD, OOMD2, bA631M21.2
UniGene
Hs.532659
Gene Chromosome Location
Chr.10: 46437-49504 on Build GRCh38
Transcripts
NM_177987.2 XM_011519459.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3978916 c.57+4G>T p.? Chr.10: 49178-49178 on Build GRCh38
6130840 c.97G>T p.A33S Chr.10: 48873-48873 on Build GRCh38
3935212 c.165C>T p.S55S Chr.10: 48805-48805 on Build GRCh38
3868061 c.113G>A p.G38E Chr.10: 48857-48857 on Build GRCh38
6433925 c.103A>G p.T35A Chr.10: 48867-48867 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782556398 Chr.10: 48949-48949 on Build GRCh38
rs781926447 Chr.10: 48924-48924 on Build GRCh38
rs558699368 Chr.10: 49315-49315 on Build GRCh38
rs782044041 Chr.10: 48838-48838 on Build GRCh38
rs536294088 Chr.10: 49287-49287 on Build GRCh38
rs200438923 Chr.10: 49256-49256 on Build GRCh38
rs374587676 Chr.10: 49276-49276 on Build GRCh38
rs371572684 Chr.10: 48908-48908 on Build GRCh38
rs532653317 Chr.10: 49266-49266 on Build GRCh38
rs56184415 Chr.10: 49308-49308 on Build GRCh38
rs565148518 Chr.10: 49157-49157 on Build GRCh38
rs6560828 Chr.10: 49134-49134 on Build GRCh38
rs782245099 Chr.10: 49290-49290 on Build GRCh38
rs782151041 Chr.10: 48923-48923 on Build GRCh38
rs782189229 Chr.10: 49132-49132 on Build GRCh38
rs782037271 Chr.10: 48918-48918 on Build GRCh38
rs565866290 Chr.10: 49063-49063 on Build GRCh38
rs782028974 Chr.10: 49139-49139 on Build GRCh38
rs189206149 Chr.10: 48883-48883 on Build GRCh38
rs782066604 Chr.10: 49211-49211 on Build GRCh38
rs782226878 Chr.10: 48957-48957 on Build GRCh38
rs782643929 Chr.10: 48962-48962 on Build GRCh38
rs571518881 Chr.10: 49112-49112 on Build GRCh38
rs782646758 Chr.10: 48877-48877 on Build GRCh38
rs545289800 Chr.10: 48959-48959 on Build GRCh38
rs113214939 Chr.10: 49321-49321 on Build GRCh38
rs201477872 Chr.10: 49203-49203 on Build GRCh38
rs34828119 Chr.10: 48879-48879 on Build GRCh38
rs782135088 Chr.10: 49135-49135 on Build GRCh38
rs199981930 Chr.10: 49213-49213 on Build GRCh38
rs6560829 Chr.10: 49286-49286 on Build GRCh38
rs782471513 Chr.10: 48872-48872 on Build GRCh38
rs368410155 Chr.10: 48878-48878 on Build GRCh38
rs782657768 Chr.10: 49228-49228 on Build GRCh38
rs530111185 Chr.10: 49047-49047 on Build GRCh38
rs572670473 Chr.10: 49150-49150 on Build GRCh38
rs574738305 Chr.10: 49327-49327 on Build GRCh38
rs367863774 Chr.10: 48856-48856 on Build GRCh38
rs781930898 Chr.10: 49163-49163 on Build GRCh38
rs575824723 Chr.10: 49316-49316 on Build GRCh38
rs781886505 Chr.10: 48940-48940 on Build GRCh38
rs202105357 Chr.10: 49255-49255 on Build GRCh38
rs536693166 Chr.10: 48846-48846 on Build GRCh38
rs375765076 Chr.10: 49142-49142 on Build GRCh38
rs375330271 Chr.10: 49037-49037 on Build GRCh38
rs202227666 Chr.10: 49227-49227 on Build GRCh38
rs782400971 Chr.10: 48831-48831 on Build GRCh38
rs201207808 Chr.10: 49248-49248 on Build GRCh38
rs370158636 Chr.10: 49151-49151 on Build GRCh38
rs782410536 Chr.10: 48963-48963 on Build GRCh38
rs368985079 Chr.10: 48943-48943 on Build GRCh38
rs182056908 Chr.10: 49280-49280 on Build GRCh38
rs561104222 Chr.10: 49230-49230 on Build GRCh38
rs772690436 Chr.10: 48859-48859 on Build GRCh38
rs377636729 Chr.10: 49278-49278 on Build GRCh38
rs776449844 Chr.10: 49269-49269 on Build GRCh38
rs371961820 Chr.10: 49032-49032 on Build GRCh38
rs566191947 Chr.10: 49306-49306 on Build GRCh38
rs782355581 Chr.10: 49178-49178 on Build GRCh38
rs547105830 Chr.10: 49259-49259 on Build GRCh38
rs782429159 Chr.10: 49250-49250 on Build GRCh38
rs781964423 Chr.10: 48896-48896 on Build GRCh38
rs782378907 Chr.10: 48909-48909 on Build GRCh38
rs782303131 Chr.10: 48888-48888 on Build GRCh38
rs68128259 Chr.10: 48932-48932 on Build GRCh38
rs552693939 Chr.10: 49147-49147 on Build GRCh38
rs74113794 Chr.10: 49349-49349 on Build GRCh38
rs782587024 Chr.10: 49131-49131 on Build GRCh38
rs560614954 Chr.10: 49264-49264 on Build GRCh38
rs781940421 Chr.10: 48832-48832 on Build GRCh38
rs530142525 Chr.10: 49249-49249 on Build GRCh38
rs565504140 Chr.10: 48968-48969 on Build GRCh38
rs782362715 Chr.10: 49133-49133 on Build GRCh38
rs11251920 Chr.10: 48932-48932 on Build GRCh38
rs199892705 Chr.10: 49197-49197 on Build GRCh38
rs782281949 Chr.10: 49158-49158 on Build GRCh38
rs781842009 Chr.10: 48917-48917 on Build GRCh38
rs781949445 Chr.10: 49188-49188 on Build GRCh38
rs540586607 Chr.10: 49346-49346 on Build GRCh38
rs782698324 Chr.10: 48873-48873 on Build GRCh38
rs534855745 Chr.10: 49090-49090 on Build GRCh38
rs200919246 Chr.10: 49214-49214 on Build GRCh38
rs782661117 Chr.10: 49148-49148 on Build GRCh38
rs782672679 Chr.10: 48884-48884 on Build GRCh38
rs149334029 Chr.10: 48874-48874 on Build GRCh38
rs869025273 Chr.10: 49234-49234 on Build GRCh38
rs199817418 Chr.10: 49229-49229 on Build GRCh38
rs112134897 Chr.10: 48928-48928 on Build GRCh38
rs782437559 Chr.10: 48939-48940 on Build GRCh38
rs782145810 Chr.10: 49140-49140 on Build GRCh38
rs575816007 Chr.10: 49160-49160 on Build GRCh38
rs558290342 Chr.10: 49272-49272 on Build GRCh38
rs782742393 Chr.10: 48841-48841 on Build GRCh38
rs369136403 Chr.10: 49146-49146 on Build GRCh38
rs116058315 Chr.10: 49257-49257 on Build GRCh38
rs782100591 Chr.10: 49144-49144 on Build GRCh38
rs782708970 Chr.10: 49212-49212 on Build GRCh38
rs181491095 Chr.10: 49092-49092 on Build GRCh38
rs550661825 Chr.10: 48986-48986 on Build GRCh38
rs544563719 Chr.10: 49320-49320 on Build GRCh38
rs11251919 Chr.10: 48930-48930 on Build GRCh38
rs547100867 Chr.10: 49057-49057 on Build GRCh38
rs565286582 Chr.10: 48971-48971 on Build GRCh38
rs782444001 Chr.10: 49281-49281 on Build GRCh38
rs201277162 Chr.10: 49258-49258 on Build GRCh38
rs782048968 Chr.10: 49171-49171 on Build GRCh38
rs556647159 Chr.10: 49127-49127 on Build GRCh38
rs372323345 Chr.10: 49126-49126 on Build GRCh38
rs11251929 Chr.10: 49156-49156 on Build GRCh38
rs186208482 Chr.10: 49297-49297 on Build GRCh38
rs781887693 Chr.10: 48861-48861 on Build GRCh38
rs782652485 Chr.10: 49239-49239 on Build GRCh38
rs544912034 Chr.10: 49221-49221 on Build GRCh38
rs781975937 Chr.10: 48823-48823 on Build GRCh38
rs577701532 Chr.10: 48827-48827 on Build GRCh38
rs782190891 Chr.10: 48886-48886 on Build GRCh38
rs782223716 Chr.10: 49153-49153 on Build GRCh38
rs782362140 Chr.10: 49137-49137 on Build GRCh38
rs369719593 Chr.10: 49282-49282 on Build GRCh38
rs6560830 Chr.10: 49289-49289 on Build GRCh38
rs782122091 Chr.10: 48854-48854 on Build GRCh38
rs782499869 Chr.10: 48933-48933 on Build GRCh38
rs782714133 Chr.10: 49141-49141 on Build GRCh38

View Full Product Details