Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 32972994-32973486 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BRD2
Entrez Gene ID
6046
Gene Name
bromodomain containing 2
Gene Aliases
D6S113E, FSH, FSRG1, NAT, O27.1.1, RING3, RNF3
UniGene
Hs.75243
Gene Chromosome Location
Chr.6: 32968660-32981505 on Build GRCh38
Transcripts
NM_001113182.2 NM_001199455.1 NM_001199456.1 NM_001291986.1 NM_005104.3

SNP Detail

SNP ID Chromosome Location
rs888797797 Chr.6: 32973018-32973033 on Build GRCh38
rs181999924 Chr.6: 32973428-32973428 on Build GRCh38
rs17220479 Chr.6: 32973038-32973038 on Build GRCh38
rs756134836 Chr.6: 32973259-32973260 on Build GRCh38
rs931597580 Chr.6: 32973261-32973261 on Build GRCh38
rs912596483 Chr.6: 32973347-32973347 on Build GRCh38
rs12196597 Chr.6: 32973437-32973437 on Build GRCh38
rs531095072 Chr.6: 32973102-32973102 on Build GRCh38
rs781636153 Chr.6: 32973036-32973036 on Build GRCh38
rs568412719 Chr.6: 32973143-32973143 on Build GRCh38
rs768013841 Chr.6: 32973047-32973047 on Build GRCh38
rs975602905 Chr.6: 32973157-32973157 on Build GRCh38
rs116719162 Chr.6: 32973027-32973027 on Build GRCh38
rs564695425 Chr.6: 32973129-32973129 on Build GRCh38
rs563834112 Chr.6: 32973066-32973066 on Build GRCh38
rs1032191138 Chr.6: 32973298-32973298 on Build GRCh38
rs995093214 Chr.6: 32973094-32973094 on Build GRCh38
rs112099614 Chr.6: 32973265-32973266 on Build GRCh38
rs1025111713 Chr.6: 32973293-32973293 on Build GRCh38
rs781234144 Chr.6: 32973231-32973231 on Build GRCh38
rs756942725 Chr.6: 32973166-32973166 on Build GRCh38
rs894845009 Chr.6: 32973281-32973281 on Build GRCh38
rs760201056 Chr.6: 32973016-32973016 on Build GRCh38
rs999305116 Chr.6: 32973461-32973461 on Build GRCh38
rs1017262569 Chr.6: 32973325-32973325 on Build GRCh38
rs546413107 Chr.6: 32973118-32973118 on Build GRCh38
rs757656991 Chr.6: 32973035-32973035 on Build GRCh38
rs1053529443 Chr.6: 32973042-32973042 on Build GRCh38
rs558369363 Chr.6: 32973026-32973026 on Build GRCh38
rs771746626 Chr.6: 32973052-32973052 on Build GRCh38
rs529274703 Chr.6: 32973145-32973145 on Build GRCh38
rs1037162298 Chr.6: 32973257-32973257 on Build GRCh38
rs772102542 Chr.6: 32973183-32973183 on Build GRCh38
rs756520301 Chr.6: 32973107-32973107 on Build GRCh38
rs28361067 Chr.6: 32973425-32973425 on Build GRCh38
rs540756581 Chr.6: 32973029-32973029 on Build GRCh38
rs772484415 Chr.6: 32973356-32973356 on Build GRCh38
rs149792692 Chr.6: 32973302-32973302 on Build GRCh38
rs757260877 Chr.6: 32973262-32973262 on Build GRCh38
rs72865879 Chr.6: 32973031-32973031 on Build GRCh38
rs12196639 Chr.6: 32973350-32973350 on Build GRCh38
rs745727025 Chr.6: 32973151-32973151 on Build GRCh38
rs977928766 Chr.6: 32973142-32973142 on Build GRCh38
rs145779446 Chr.6: 32973326-32973326 on Build GRCh38
rs552815158 Chr.6: 32973296-32973296 on Build GRCh38
rs754252649 Chr.6: 32973113-32973113 on Build GRCh38
rs997094605 Chr.6: 32973338-32973338 on Build GRCh38
rs1016121811 Chr.6: 32973388-32973388 on Build GRCh38
rs1009010165 Chr.6: 32973316-32973316 on Build GRCh38
rs1019852143 Chr.6: 32973138-32973138 on Build GRCh38
rs983239630 Chr.6: 32973173-32973173 on Build GRCh38
rs140063261 Chr.6: 32973159-32973159 on Build GRCh38
rs573496552 Chr.6: 32973401-32973401 on Build GRCh38
rs762007996 Chr.6: 32973088-32973088 on Build GRCh38
rs747686119 Chr.6: 32973050-32973050 on Build GRCh38
rs569840028 Chr.6: 32973156-32973156 on Build GRCh38
rs761933919 Chr.6: 32973175-32973175 on Build GRCh38
rs548997601 Chr.6: 32973387-32973387 on Build GRCh38
rs371150722 Chr.6: 32973140-32973140 on Build GRCh38
rs937925805 Chr.6: 32973283-32973283 on Build GRCh38
rs890144214 Chr.6: 32973310-32973310 on Build GRCh38
rs767840665 Chr.6: 32973092-32973092 on Build GRCh38
rs972843929 Chr.6: 32973375-32973375 on Build GRCh38
rs748850063 Chr.6: 32973043-32973043 on Build GRCh38
rs902232399 Chr.6: 32973445-32973445 on Build GRCh38
rs982115236 Chr.6: 32973407-32973407 on Build GRCh38
rs3130597 Chr.6: 32973414-32973414 on Build GRCh38
rs749406326 Chr.6: 32973234-32973234 on Build GRCh38
rs776509186 Chr.6: 32973060-32973060 on Build GRCh38
rs751685448 Chr.6: 32973034-32973034 on Build GRCh38
rs542484085 Chr.6: 32973062-32973062 on Build GRCh38
rs754517349 Chr.6: 32973128-32973128 on Build GRCh38
rs986825450 Chr.6: 32973346-32973346 on Build GRCh38
rs1053599509 Chr.6: 32973464-32973464 on Build GRCh38
rs780688150 Chr.6: 32973040-32973040 on Build GRCh38
rs556958927 Chr.6: 32973303-32973303 on Build GRCh38
rs760440868 Chr.6: 32973058-32973058 on Build GRCh38
rs893606012 Chr.6: 32973297-32973297 on Build GRCh38
rs952967088 Chr.6: 32973368-32973368 on Build GRCh38
rs926565359 Chr.6: 32973266-32973266 on Build GRCh38
rs559757518 Chr.6: 32973420-32973420 on Build GRCh38
rs949384573 Chr.6: 32973222-32973222 on Build GRCh38
rs747309549 Chr.6: 32973389-32973389 on Build GRCh38
rs17214071 Chr.6: 32973178-32973178 on Build GRCh38
rs548424479 Chr.6: 32973147-32973147 on Build GRCh38
rs540681808 Chr.6: 32973411-32973411 on Build GRCh38
rs1013330677 Chr.6: 32973289-32973289 on Build GRCh38
rs190465055 Chr.6: 32973190-32973190 on Build GRCh38
rs780740941 Chr.6: 32973111-32973111 on Build GRCh38
rs878972647 Chr.6: 32973185-32973185 on Build GRCh38
rs114215603 Chr.6: 32973136-32973136 on Build GRCh38
rs764023804 Chr.6: 32973065-32973065 on Build GRCh38
rs751813812 Chr.6: 32973077-32973077 on Build GRCh38
rs912721271 Chr.6: 32973180-32973180 on Build GRCh38
rs558309791 Chr.6: 32973165-32973165 on Build GRCh38
rs747677219 Chr.6: 32973132-32973132 on Build GRCh38
rs902263244 Chr.6: 32973295-32973295 on Build GRCh38
rs1020624767 Chr.6: 32973071-32973071 on Build GRCh38
rs750869793 Chr.6: 32973103-32973103 on Build GRCh38

View Full Product Details