Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 170544332-170544878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GORAB
Entrez Gene ID
92344
Gene Name
golgin, RAB6 interacting
Gene Aliases
GO, NTKLBP1, SCYL1BP1
UniGene
Hs.183702
Gene Chromosome Location
Chr.1: 170532122-170553834 on Build GRCh38
Transcripts
NM_001146039.1 NM_001320252.1 NM_152281.2 NR_027397.1 XM_006711628.3 XM_011510149.2 XM_011510150.2 XM_011510151.1 XM_017002807.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5485502 c.635G>A p.R212Q Chr.1: 170544743-170544743 on Build GRCh38
6672827 c.681C>T p.D227D Chr.1: 170544789-170544789 on Build GRCh38
6672824 c.634C>T p.R212W Chr.1: 170544742-170544742 on Build GRCh38
3478163 c.679G>A p.D227N Chr.1: 170544787-170544787 on Build GRCh38
7126316 c.706G>A p.D236N Chr.1: 170544814-170544814 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985697164 Chr.1: 170544532-170544532 on Build GRCh38
rs138045019 Chr.1: 170544576-170544577 on Build GRCh38
rs528084129 Chr.1: 170544403-170544403 on Build GRCh38
rs764978536 Chr.1: 170544427-170544427 on Build GRCh38
rs999521107 Chr.1: 170544817-170544817 on Build GRCh38
rs138502097 Chr.1: 170544808-170544808 on Build GRCh38
rs375910593 Chr.1: 170544682-170544682 on Build GRCh38
rs958439493 Chr.1: 170544396-170544396 on Build GRCh38
rs774619703 Chr.1: 170544747-170544747 on Build GRCh38
rs978103881 Chr.1: 170544486-170544486 on Build GRCh38
rs745963369 Chr.1: 170544712-170544712 on Build GRCh38
rs993265536 Chr.1: 170544637-170544637 on Build GRCh38
rs539291301 Chr.1: 170544640-170544640 on Build GRCh38
rs747896164 Chr.1: 170544683-170544683 on Build GRCh38
rs750477207 Chr.1: 170544658-170544658 on Build GRCh38
rs185701616 Chr.1: 170544696-170544696 on Build GRCh38
rs894433598 Chr.1: 170544576-170544576 on Build GRCh38
rs775889980 Chr.1: 170544732-170544732 on Build GRCh38
rs35872826 Chr.1: 170544801-170544801 on Build GRCh38
rs569049244 Chr.1: 170544635-170544635 on Build GRCh38
rs750368506 Chr.1: 170544669-170544669 on Build GRCh38
rs34521824 Chr.1: 170544679-170544680 on Build GRCh38
rs766291965 Chr.1: 170544828-170544828 on Build GRCh38
rs183596463 Chr.1: 170544841-170544841 on Build GRCh38
rs568072369 Chr.1: 170544521-170544521 on Build GRCh38
rs764819857 Chr.1: 170544779-170544779 on Build GRCh38
rs762148026 Chr.1: 170544657-170544659 on Build GRCh38
rs906818390 Chr.1: 170544362-170544362 on Build GRCh38
rs142444073 Chr.1: 170544775-170544775 on Build GRCh38
rs989703175 Chr.1: 170544405-170544405 on Build GRCh38
rs557062602 Chr.1: 170544854-170544854 on Build GRCh38
rs758084831 Chr.1: 170544806-170544806 on Build GRCh38
rs375601907 Chr.1: 170544392-170544392 on Build GRCh38
rs753322057 Chr.1: 170544770-170544770 on Build GRCh38
rs749490786 Chr.1: 170544832-170544832 on Build GRCh38
rs922501984 Chr.1: 170544510-170544510 on Build GRCh38
rs765367090 Chr.1: 170544814-170544814 on Build GRCh38
rs17855279 Chr.1: 170544708-170544708 on Build GRCh38
rs752132099 Chr.1: 170544655-170544655 on Build GRCh38
rs1055787626 Chr.1: 170544778-170544778 on Build GRCh38
rs76365864 Chr.1: 170544840-170544840 on Build GRCh38
rs187430236 Chr.1: 170544523-170544523 on Build GRCh38
rs959718777 Chr.1: 170544417-170544417 on Build GRCh38
rs765883131 Chr.1: 170544672-170544672 on Build GRCh38
rs754566739 Chr.1: 170544681-170544681 on Build GRCh38
rs763362522 Chr.1: 170544742-170544742 on Build GRCh38
rs546229609 Chr.1: 170544520-170544520 on Build GRCh38
rs768828994 Chr.1: 170544833-170544833 on Build GRCh38
rs773728833 Chr.1: 170544845-170544845 on Build GRCh38
rs770115158 Chr.1: 170544745-170544745 on Build GRCh38
rs200802067 Chr.1: 170544809-170544809 on Build GRCh38
rs1022806933 Chr.1: 170544450-170544450 on Build GRCh38
rs772589935 Chr.1: 170544844-170544844 on Build GRCh38
rs966671066 Chr.1: 170544481-170544481 on Build GRCh38
rs890134115 Chr.1: 170544649-170544649 on Build GRCh38
rs190557815 Chr.1: 170544743-170544743 on Build GRCh38
rs762378708 Chr.1: 170544759-170544759 on Build GRCh38
rs770077182 Chr.1: 170544722-170544722 on Build GRCh38
rs780332229 Chr.1: 170544831-170544831 on Build GRCh38
rs746929438 Chr.1: 170544689-170544689 on Build GRCh38
rs753345878 Chr.1: 170544680-170544680 on Build GRCh38
rs1033885057 Chr.1: 170544416-170544416 on Build GRCh38
rs941199170 Chr.1: 170544600-170544600 on Build GRCh38
rs767921831 Chr.1: 170544763-170544763 on Build GRCh38
rs1037570542 Chr.1: 170544586-170544586 on Build GRCh38
rs780999041 Chr.1: 170544821-170544821 on Build GRCh38
rs562039849 Chr.1: 170544480-170544480 on Build GRCh38
rs550715545 Chr.1: 170544572-170544572 on Build GRCh38
rs750572178 Chr.1: 170544838-170544839 on Build GRCh38
rs745905750 Chr.1: 170544825-170544825 on Build GRCh38
rs193265861 Chr.1: 170544641-170544641 on Build GRCh38
rs115208114 Chr.1: 170544365-170544365 on Build GRCh38
rs929627053 Chr.1: 170544675-170544675 on Build GRCh38
rs368316601 Chr.1: 170544820-170544820 on Build GRCh38
rs777474951 Chr.1: 170544686-170544686 on Build GRCh38
rs941841650 Chr.1: 170544585-170544585 on Build GRCh38

View Full Product Details