Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 100274735-100275313 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BLOC1S2
Entrez Gene ID
282991
Gene Name
biogenesis of lysosomal organelles complex 1 subunit 2
Gene Aliases
BLOS2, BORCS2, CEAP, CEAP11
UniGene
Hs.34906
Gene Chromosome Location
Chr.10: 100273278-100286712 on Build GRCh38
Transcripts
NM_001001342.2 NM_001282436.1 NM_001282437.1 NM_001282438.1 NM_001282439.1 NM_173809.4

SNP Detail

SNP ID Chromosome Location
rs1014472638 Chr.10: 100274862-100274862 on Build GRCh38
rs979680361 Chr.10: 100275029-100275029 on Build GRCh38
rs968692461 Chr.10: 100275025-100275025 on Build GRCh38
rs905224831 Chr.10: 100274810-100274810 on Build GRCh38
rs535904434 Chr.10: 100275229-100275229 on Build GRCh38
rs914098620 Chr.10: 100274971-100274971 on Build GRCh38
rs889034358 Chr.10: 100275245-100275245 on Build GRCh38
rs988329447 Chr.10: 100274761-100274761 on Build GRCh38
rs565835348 Chr.10: 100274883-100274883 on Build GRCh38
rs940157323 Chr.10: 100275149-100275149 on Build GRCh38
rs770078029 Chr.10: 100275266-100275266 on Build GRCh38
rs570589752 Chr.10: 100274938-100274938 on Build GRCh38
rs112642355 Chr.10: 100274900-100274900 on Build GRCh38
rs368154127 Chr.10: 100275154-100275154 on Build GRCh38
rs986953267 Chr.10: 100275036-100275036 on Build GRCh38
rs78751060 Chr.10: 100275146-100275146 on Build GRCh38
rs965775175 Chr.10: 100274954-100274954 on Build GRCh38
rs917996349 Chr.10: 100275147-100275147 on Build GRCh38
rs1015974132 Chr.10: 100274917-100274917 on Build GRCh38
rs926883042 Chr.10: 100275039-100275039 on Build GRCh38
rs148896290 Chr.10: 100274830-100274830 on Build GRCh38
rs1056489851 Chr.10: 100275131-100275131 on Build GRCh38
rs189813357 Chr.10: 100275206-100275206 on Build GRCh38
rs572571279 Chr.10: 100275251-100275251 on Build GRCh38
rs754904948 Chr.10: 100274858-100274858 on Build GRCh38
rs774650178 Chr.10: 100275204-100275205 on Build GRCh38
rs1043735731 Chr.10: 100274792-100274792 on Build GRCh38
rs777332391 Chr.10: 100274928-100274928 on Build GRCh38
rs143606335 Chr.10: 100274868-100274868 on Build GRCh38
rs771921392 Chr.10: 100275240-100275240 on Build GRCh38
rs778568428 Chr.10: 100274860-100274860 on Build GRCh38
rs12267390 Chr.10: 100274982-100274982 on Build GRCh38
rs914132510 Chr.10: 100274769-100274769 on Build GRCh38
rs182147651 Chr.10: 100275236-100275236 on Build GRCh38
rs1044100080 Chr.10: 100274809-100274809 on Build GRCh38
rs185447113 Chr.10: 100275122-100275122 on Build GRCh38
rs911376755 Chr.10: 100275103-100275103 on Build GRCh38
rs749420231 Chr.10: 100275049-100275049 on Build GRCh38
rs551242337 Chr.10: 100274970-100274970 on Build GRCh38
rs1013660268 Chr.10: 100275241-100275241 on Build GRCh38
rs890012643 Chr.10: 100274784-100274784 on Build GRCh38
rs115662071 Chr.10: 100274929-100274929 on Build GRCh38
rs897732106 Chr.10: 100274875-100274875 on Build GRCh38
rs561616509 Chr.10: 100274786-100274786 on Build GRCh38
rs1027719930 Chr.10: 100274925-100274925 on Build GRCh38
rs768573547 Chr.10: 100275138-100275138 on Build GRCh38
rs1052962766 Chr.10: 100275209-100275209 on Build GRCh38
rs879783481 Chr.10: 100274841-100274841 on Build GRCh38
rs752321204 Chr.10: 100274861-100274861 on Build GRCh38
rs903846142 Chr.10: 100274794-100274794 on Build GRCh38
rs556185335 Chr.10: 100275157-100275157 on Build GRCh38
rs905021686 Chr.10: 100275271-100275271 on Build GRCh38

View Full Product Details