Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 44523133-44523555 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NELL2
Entrez Gene ID
4753
Gene Name
neural EGFL like 2
Gene Aliases
NRP2
UniGene
Hs.505326
Gene Chromosome Location
Chr.12: 44508275-44921848 on Build GRCh38
Transcripts
NM_001145107.1 NM_001145108.1 NM_001145109.1 NM_001145110.1 NM_006159.2 XM_005268905.3 XM_011538396.1 XM_017019341.1 XM_017019342.1 XM_017019343.1 XM_017019344.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3731625 c.1935A>C p.K645N Chr.12: 44523354-44523354 on Build GRCh38
3460907 c.2024G>A p.G675E Chr.12: 44523415-44523415 on Build GRCh38
693951 c.1826G>T p.G609V Chr.12: 44523463-44523463 on Build GRCh38
6136539 c.1914C>A p.D638E Chr.12: 44523372-44523372 on Build GRCh38
468367 c.1821G>A p.G607G Chr.12: 44523468-44523468 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771202210 Chr.12: 44523479-44523479 on Build GRCh38
rs745705477 Chr.12: 44523415-44523415 on Build GRCh38
rs762324368 Chr.12: 44523345-44523345 on Build GRCh38
rs572731322 Chr.12: 44523489-44523489 on Build GRCh38
rs369136591 Chr.12: 44523487-44523487 on Build GRCh38
rs746502275 Chr.12: 44523336-44523336 on Build GRCh38
rs761759696 Chr.12: 44523526-44523526 on Build GRCh38
rs955204718 Chr.12: 44523464-44523464 on Build GRCh38
rs199859063 Chr.12: 44523442-44523442 on Build GRCh38
rs767875389 Chr.12: 44523241-44523241 on Build GRCh38
rs368334753 Chr.12: 44523298-44523298 on Build GRCh38
rs748885133 Chr.12: 44523340-44523340 on Build GRCh38
rs761818482 Chr.12: 44523501-44523501 on Build GRCh38
rs767825403 Chr.12: 44523360-44523360 on Build GRCh38
rs765364103 Chr.12: 44523491-44523491 on Build GRCh38
rs34214960 Chr.12: 44523387-44523388 on Build GRCh38
rs905354195 Chr.12: 44523220-44523220 on Build GRCh38
rs182914291 Chr.12: 44523307-44523307 on Build GRCh38
rs750179536 Chr.12: 44523511-44523511 on Build GRCh38
rs750590443 Chr.12: 44523244-44523244 on Build GRCh38
rs758247591 Chr.12: 44523277-44523277 on Build GRCh38
rs192963979 Chr.12: 44523230-44523230 on Build GRCh38
rs554020057 Chr.12: 44523444-44523444 on Build GRCh38
rs772611169 Chr.12: 44523466-44523466 on Build GRCh38
rs757077926 Chr.12: 44523178-44523178 on Build GRCh38
rs1050710 Chr.12: 44523397-44523397 on Build GRCh38
rs757059383 Chr.12: 44523297-44523297 on Build GRCh38
rs773873793 Chr.12: 44523460-44523460 on Build GRCh38
rs753992072 Chr.12: 44523266-44523266 on Build GRCh38
rs779821233 Chr.12: 44523437-44523437 on Build GRCh38
rs201592050 Chr.12: 44523417-44523417 on Build GRCh38
rs760778711 Chr.12: 44523366-44523366 on Build GRCh38
rs372252823 Chr.12: 44523278-44523278 on Build GRCh38
rs757288037 Chr.12: 44523403-44523403 on Build GRCh38
rs909937854 Chr.12: 44523174-44523174 on Build GRCh38
rs372448410 Chr.12: 44523250-44523250 on Build GRCh38
rs376601496 Chr.12: 44523287-44523287 on Build GRCh38
rs778717407 Chr.12: 44523272-44523272 on Build GRCh38
rs773598023 Chr.12: 44523364-44523364 on Build GRCh38
rs144132963 Chr.12: 44523358-44523358 on Build GRCh38
rs748614100 Chr.12: 44523461-44523461 on Build GRCh38
rs753751564 Chr.12: 44523261-44523264 on Build GRCh38
rs200328352 Chr.12: 44523492-44523492 on Build GRCh38
rs773546810 Chr.12: 44523468-44523468 on Build GRCh38
rs779727944 Chr.12: 44523519-44523519 on Build GRCh38
rs146099301 Chr.12: 44523326-44523326 on Build GRCh38
rs768156247 Chr.12: 44523445-44523445 on Build GRCh38
rs188113747 Chr.12: 44523512-44523512 on Build GRCh38
rs1036994131 Chr.12: 44523193-44523193 on Build GRCh38
rs766329552 Chr.12: 44523259-44523259 on Build GRCh38
rs1028891695 Chr.12: 44523427-44523427 on Build GRCh38
rs751489476 Chr.12: 44523400-44523400 on Build GRCh38
rs755873024 Chr.12: 44523517-44523517 on Build GRCh38
rs372305255 Chr.12: 44523343-44523343 on Build GRCh38
rs953203186 Chr.12: 44523436-44523436 on Build GRCh38
rs766673976 Chr.12: 44523379-44523379 on Build GRCh38
rs374154916 Chr.12: 44523416-44523416 on Build GRCh38
rs1012652680 Chr.12: 44523189-44523189 on Build GRCh38
rs138882159 Chr.12: 44523408-44523408 on Build GRCh38
rs188361127 Chr.12: 44523203-44523203 on Build GRCh38
rs973175341 Chr.12: 44523480-44523480 on Build GRCh38
rs753483104 Chr.12: 44523533-44523533 on Build GRCh38
rs767619586 Chr.12: 44523504-44523504 on Build GRCh38
rs374706671 Chr.12: 44523524-44523524 on Build GRCh38
rs199920966 Chr.12: 44523395-44523395 on Build GRCh38
rs1003838231 Chr.12: 44523227-44523227 on Build GRCh38
rs142328202 Chr.12: 44523525-44523525 on Build GRCh38
rs375353348 Chr.12: 44523469-44523469 on Build GRCh38

View Full Product Details