Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 119042171-119042543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
WARS2
Entrez Gene ID
10352
Gene Name
tryptophanyl tRNA synthetase 2, mitochondrial
Gene Aliases
TrpRS
UniGene
Hs.523506
Gene Chromosome Location
Chr.1: 119031216-119140701 on Build GRCh38
Transcripts
NM_015836.3 NM_201263.2 XM_005270350.3 XM_011540494.2 XM_011540495.2 XM_017000038.1 XM_017000039.1 XM_017000040.1 XM_017000041.1 XM_017000042.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4004735 c.475A>G p.T159A Chr.1: 119042304-119042304 on Build GRCh38
7301627 c.498C>T p.A166A Chr.1: 119042281-119042281 on Build GRCh38
7397559 c.458G>T p.G153V Chr.1: 119042321-119042321 on Build GRCh38
2179520 c.450G>T p.K150N Chr.1: 119042329-119042329 on Build GRCh38
4021238 c.439A>G p.T147A Chr.1: 119042340-119042340 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs576502507 Chr.1: 119042419-119042419 on Build GRCh38
rs757435337 Chr.1: 119042331-119042331 on Build GRCh38
rs149281638 Chr.1: 119042296-119042296 on Build GRCh38
rs60486007 Chr.1: 119042231-119042231 on Build GRCh38
rs1005216761 Chr.1: 119042496-119042496 on Build GRCh38
rs762541487 Chr.1: 119042310-119042310 on Build GRCh38
rs917616593 Chr.1: 119042417-119042417 on Build GRCh38
rs201758984 Chr.1: 119042324-119042324 on Build GRCh38
rs144546478 Chr.1: 119042281-119042281 on Build GRCh38
rs769305019 Chr.1: 119042370-119042370 on Build GRCh38
rs767554000 Chr.1: 119042318-119042318 on Build GRCh38
rs769789160 Chr.1: 119042244-119042244 on Build GRCh38
rs770708427 Chr.1: 119042389-119042389 on Build GRCh38
rs776043900 Chr.1: 119042286-119042286 on Build GRCh38
rs774209060 Chr.1: 119042393-119042393 on Build GRCh38
rs778036621 Chr.1: 119042259-119042259 on Build GRCh38
rs1052634647 Chr.1: 119042409-119042409 on Build GRCh38
rs143494742 Chr.1: 119042366-119042366 on Build GRCh38
rs570439512 Chr.1: 119042292-119042292 on Build GRCh38
rs189518072 Chr.1: 119042479-119042479 on Build GRCh38
rs949125188 Chr.1: 119042466-119042466 on Build GRCh38
rs774297528 Chr.1: 119042314-119042314 on Build GRCh38
rs1044873093 Chr.1: 119042480-119042480 on Build GRCh38
rs868119795 Chr.1: 119042322-119042322 on Build GRCh38
rs759386863 Chr.1: 119042395-119042395 on Build GRCh38
rs376622172 Chr.1: 119042352-119042352 on Build GRCh38
rs904859781 Chr.1: 119042481-119042481 on Build GRCh38
rs369604703 Chr.1: 119042354-119042354 on Build GRCh38
rs185565273 Chr.1: 119042196-119042196 on Build GRCh38
rs752628633 Chr.1: 119042325-119042325 on Build GRCh38
rs765338789 Chr.1: 119042215-119042215 on Build GRCh38
rs757604003 Chr.1: 119042279-119042279 on Build GRCh38
rs33960696 Chr.1: 119042326-119042326 on Build GRCh38
rs759476628 Chr.1: 119042317-119042317 on Build GRCh38
rs779182677 Chr.1: 119042339-119042339 on Build GRCh38
rs780515660 Chr.1: 119042357-119042357 on Build GRCh38
rs750782270 Chr.1: 119042342-119042342 on Build GRCh38
rs140008988 Chr.1: 119042312-119042312 on Build GRCh38
rs754045233 Chr.1: 119042328-119042328 on Build GRCh38
rs149201054 Chr.1: 119042337-119042337 on Build GRCh38
rs888059785 Chr.1: 119042486-119042486 on Build GRCh38
rs749515082 Chr.1: 119042270-119042270 on Build GRCh38
rs1005916308 Chr.1: 119042487-119042487 on Build GRCh38
rs370215087 Chr.1: 119042237-119042237 on Build GRCh38
rs748909829 Chr.1: 119042378-119042378 on Build GRCh38
rs779272688 Chr.1: 119042280-119042280 on Build GRCh38
rs747679577 Chr.1: 119042288-119042288 on Build GRCh38
rs754985501 Chr.1: 119042233-119042233 on Build GRCh38
rs113886394 Chr.1: 119042242-119042242 on Build GRCh38
rs150022801 Chr.1: 119042327-119042327 on Build GRCh38
rs373456845 Chr.1: 119042244-119042244 on Build GRCh38
rs1047993254 Chr.1: 119042457-119042457 on Build GRCh38
rs777166854 Chr.1: 119042373-119042373 on Build GRCh38

View Full Product Details