Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 203394478-203395068 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABI2
Entrez Gene ID
10152
Gene Name
abl-interactor 2
Gene Aliases
ABI-2, ABI2B, AIP-1, AblBP3, SSH3BP2, argBP1, argBPIA, argBPIB
UniGene
Hs.471156
Gene Chromosome Location
Chr.2: 203327874-203432173 on Build GRCh38
Transcripts
NM_001282925.1 NM_001282926.1 NM_001282927.1 NM_001282932.1 NM_005759.5 XM_005246217.2 XM_005246218.2 XM_005246221.2 XM_005246225.2 XM_005246226.2 XM_006712174.2 XM_006712177.2 XM_006712179.2 XM_006712180.2 XM_006712181.2 XM_006712182.2 XM_006712185.1 XM_006712186.2 XM_011510472.2 XM_011510473.2 XM_017003118.1 XM_017003119.1 XM_017003120.1 XM_017003121.1 XM_017003122.1 XM_017003123.1 XM_017003124.1 XM_017003125.1 XM_017003126.1 XM_017003127.1 XM_017003128.1 XM_017003129.1 XM_017003130.1 XM_017003131.1 XM_017003132.1 XM_017003133.1 XM_017003134.1 XM_017003135.1 XM_017003136.1 XM_017003137.1 XM_017003138.1 XM_017003139.1 XM_017003140.1 XM_017003141.1 XM_017003142.1 XM_017003143.1 XM_017003144.1 XM_017003145.1 XM_017003146.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6090033 c.582G>A p.R194R Chr.2: 203394703-203394703 on Build GRCh38
1494779 c.584T>C p.L195P Chr.2: 203394723-203394723 on Build GRCh38
5797241 c.651C>T p.P217P Chr.2: 203394790-203394790 on Build GRCh38
3364509 c.687G>T p.V229V Chr.2: 203394808-203394808 on Build GRCh38
6590973 c.503G>A p.R168H Chr.2: 203394777-203394777 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2123118 Chr.2: 203394833-203394833 on Build GRCh38
rs756308781 Chr.2: 203394676-203394676 on Build GRCh38
rs747339200 Chr.2: 203394869-203394869 on Build GRCh38
rs943780406 Chr.2: 203394966-203394966 on Build GRCh38
rs144970407 Chr.2: 203394782-203394782 on Build GRCh38
rs148405849 Chr.2: 203394801-203394801 on Build GRCh38
rs1046744665 Chr.2: 203394633-203394633 on Build GRCh38
rs890797261 Chr.2: 203394960-203394960 on Build GRCh38
rs769362739 Chr.2: 203394734-203394734 on Build GRCh38
rs910151461 Chr.2: 203394670-203394670 on Build GRCh38
rs35652563 Chr.2: 203395000-203395001 on Build GRCh38
rs114110905 Chr.2: 203394875-203394875 on Build GRCh38
rs778815963 Chr.2: 203395033-203395033 on Build GRCh38
rs929654035 Chr.2: 203394619-203394619 on Build GRCh38
rs370923890 Chr.2: 203394712-203394712 on Build GRCh38
rs772501775 Chr.2: 203394967-203394967 on Build GRCh38
rs367846935 Chr.2: 203394675-203394675 on Build GRCh38
rs540048892 Chr.2: 203394655-203394655 on Build GRCh38
rs370162988 Chr.2: 203394701-203394701 on Build GRCh38
rs185454274 Chr.2: 203394562-203394562 on Build GRCh38
rs755245338 Chr.2: 203394825-203394825 on Build GRCh38
rs1000774078 Chr.2: 203394760-203394760 on Build GRCh38
rs753790334 Chr.2: 203394615-203394615 on Build GRCh38
rs773849369 Chr.2: 203394763-203394763 on Build GRCh38
rs551449375 Chr.2: 203394758-203394758 on Build GRCh38
rs534567946 Chr.2: 203394984-203394984 on Build GRCh38
rs748729529 Chr.2: 203394708-203394708 on Build GRCh38
rs751587341 Chr.2: 203394867-203394888 on Build GRCh38
rs758995568 Chr.2: 203394860-203394860 on Build GRCh38
rs1018079015 Chr.2: 203394657-203394657 on Build GRCh38
rs757585780 Chr.2: 203394873-203394873 on Build GRCh38
rs747350758 Chr.2: 203394721-203394721 on Build GRCh38
rs965137009 Chr.2: 203394550-203394550 on Build GRCh38
rs772675483 Chr.2: 203394714-203394714 on Build GRCh38
rs956628628 Chr.2: 203394792-203394792 on Build GRCh38
rs752801855 Chr.2: 203394839-203394839 on Build GRCh38
rs942284642 Chr.2: 203394541-203394541 on Build GRCh38
rs555096710 Chr.2: 203394900-203394900 on Build GRCh38
rs1017927774 Chr.2: 203394984-203394987 on Build GRCh38
rs760447848 Chr.2: 203394790-203394790 on Build GRCh38
rs951583122 Chr.2: 203394614-203394614 on Build GRCh38
rs374916082 Chr.2: 203394651-203394651 on Build GRCh38
rs114649171 Chr.2: 203394957-203394957 on Build GRCh38
rs781186521 Chr.2: 203394554-203394554 on Build GRCh38
rs373941580 Chr.2: 203394918-203394918 on Build GRCh38
rs753865013 Chr.2: 203394702-203394702 on Build GRCh38
rs528665645 Chr.2: 203394573-203394573 on Build GRCh38
rs971180518 Chr.2: 203394872-203394872 on Build GRCh38
rs1039505596 Chr.2: 203394998-203394998 on Build GRCh38
rs746431938 Chr.2: 203394777-203394777 on Build GRCh38
rs774990502 Chr.2: 203394742-203394742 on Build GRCh38
rs765445473 Chr.2: 203394836-203394836 on Build GRCh38
rs767424735 Chr.2: 203394779-203394779 on Build GRCh38
rs569634751 Chr.2: 203394955-203394956 on Build GRCh38
rs777548696 Chr.2: 203394713-203394713 on Build GRCh38
rs751443126 Chr.2: 203394661-203394661 on Build GRCh38
rs780278269 Chr.2: 203394683-203394683 on Build GRCh38
rs755024902 Chr.2: 203394705-203394705 on Build GRCh38
rs890840194 Chr.2: 203394648-203394648 on Build GRCh38
rs1037951835 Chr.2: 203394544-203394544 on Build GRCh38
rs778386215 Chr.2: 203394864-203394864 on Build GRCh38
rs1006507032 Chr.2: 203394983-203394983 on Build GRCh38
rs754970100 Chr.2: 203395031-203395031 on Build GRCh38
rs190160113 Chr.2: 203394876-203394876 on Build GRCh38
rs899653896 Chr.2: 203395033-203395033 on Build GRCh38
rs761783921 Chr.2: 203394776-203394776 on Build GRCh38
rs893444240 Chr.2: 203394529-203394529 on Build GRCh38
rs984757165 Chr.2: 203394616-203394616 on Build GRCh38
rs929586837 Chr.2: 203394931-203394931 on Build GRCh38
rs747612709 Chr.2: 203395032-203395032 on Build GRCh38
rs773902631 Chr.2: 203394882-203394882 on Build GRCh38
rs775142811 Chr.2: 203394735-203394735 on Build GRCh38
rs767371676 Chr.2: 203394672-203394672 on Build GRCh38
rs919485964 Chr.2: 203394915-203394915 on Build GRCh38
rs376492783 Chr.2: 203394879-203394879 on Build GRCh38

View Full Product Details