Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 51894905-51895485 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAGED1
Entrez Gene ID
9500
Gene Name
MAGE family member D1
Gene Aliases
DLXIN-1, NRAGE
UniGene
Hs.5258
Gene Chromosome Location
Chr.X: 51802998-51902354 on Build GRCh38
Transcripts
NM_001005332.1 NM_001005333.1 NM_006986.3 XM_011530835.2 XM_017029978.1 XM_017029979.1 XM_017029980.1

SNP Detail

SNP ID Chromosome Location
rs144264968 Chr.X: 51895451-51895451 on Build GRCh38
rs931761475 Chr.X: 51895333-51895333 on Build GRCh38
rs995880979 Chr.X: 51895041-51895041 on Build GRCh38
rs782253302 Chr.X: 51895155-51895155 on Build GRCh38
rs782478822 Chr.X: 51895132-51895132 on Build GRCh38
rs782161142 Chr.X: 51895203-51895203 on Build GRCh38
rs781837717 Chr.X: 51895365-51895365 on Build GRCh38
rs887431194 Chr.X: 51895107-51895107 on Build GRCh38
rs1053011035 Chr.X: 51894977-51894977 on Build GRCh38
rs782690282 Chr.X: 51895432-51895432 on Build GRCh38
rs370800331 Chr.X: 51895179-51895179 on Build GRCh38
rs961848179 Chr.X: 51895145-51895145 on Build GRCh38
rs782164609 Chr.X: 51895030-51895030 on Build GRCh38
rs782693253 Chr.X: 51895036-51895036 on Build GRCh38
rs1037699325 Chr.X: 51895421-51895421 on Build GRCh38
rs200403502 Chr.X: 51895065-51895065 on Build GRCh38
rs782232742 Chr.X: 51895094-51895094 on Build GRCh38
rs113823505 Chr.X: 51895351-51895351 on Build GRCh38
rs781822744 Chr.X: 51895035-51895035 on Build GRCh38
rs944530784 Chr.X: 51895027-51895027 on Build GRCh38
rs202085108 Chr.X: 51895237-51895237 on Build GRCh38
rs201938318 Chr.X: 51895221-51895221 on Build GRCh38
rs782336578 Chr.X: 51895339-51895339 on Build GRCh38
rs200269066 Chr.X: 51895352-51895352 on Build GRCh38
rs782090868 Chr.X: 51895035-51895036 on Build GRCh38
rs781969465 Chr.X: 51895112-51895112 on Build GRCh38
rs375273111 Chr.X: 51895182-51895182 on Build GRCh38
rs782707442 Chr.X: 51895183-51895183 on Build GRCh38
rs782763995 Chr.X: 51895277-51895277 on Build GRCh38
rs782630514 Chr.X: 51895086-51895086 on Build GRCh38
rs782072763 Chr.X: 51895402-51895402 on Build GRCh38
rs375817044 Chr.X: 51895009-51895009 on Build GRCh38
rs782224952 Chr.X: 51895335-51895335 on Build GRCh38
rs372774432 Chr.X: 51894976-51894976 on Build GRCh38
rs562698140 Chr.X: 51895247-51895247 on Build GRCh38
rs782015606 Chr.X: 51895369-51895369 on Build GRCh38
rs782782535 Chr.X: 51895204-51895204 on Build GRCh38
rs782524377 Chr.X: 51895320-51895320 on Build GRCh38
rs782376218 Chr.X: 51895178-51895178 on Build GRCh38
rs782107245 Chr.X: 51895033-51895033 on Build GRCh38
rs782777549 Chr.X: 51895029-51895030 on Build GRCh38
rs781908113 Chr.X: 51895447-51895447 on Build GRCh38
rs137856838 Chr.X: 51895353-51895353 on Build GRCh38
rs151112988 Chr.X: 51895282-51895282 on Build GRCh38
rs782486427 Chr.X: 51895215-51895215 on Build GRCh38
rs782184047 Chr.X: 51895146-51895146 on Build GRCh38
rs782423064 Chr.X: 51895147-51895147 on Build GRCh38
rs782685654 Chr.X: 51895303-51895303 on Build GRCh38
rs782818810 Chr.X: 51895210-51895210 on Build GRCh38
rs781915123 Chr.X: 51895199-51895199 on Build GRCh38
rs782744189 Chr.X: 51895034-51895034 on Build GRCh38
rs782783990 Chr.X: 51895462-51895462 on Build GRCh38
rs782622180 Chr.X: 51895325-51895325 on Build GRCh38
rs141080853 Chr.X: 51895314-51895314 on Build GRCh38
rs980161654 Chr.X: 51895208-51895208 on Build GRCh38
rs782790539 Chr.X: 51895015-51895015 on Build GRCh38
rs781868631 Chr.X: 51895031-51895031 on Build GRCh38
rs782449472 Chr.X: 51895040-51895040 on Build GRCh38
rs1004467541 Chr.X: 51895420-51895420 on Build GRCh38
rs782081087 Chr.X: 51895140-51895140 on Build GRCh38
rs782163215 Chr.X: 51895011-51895011 on Build GRCh38
rs781958046 Chr.X: 51895396-51895396 on Build GRCh38
rs781855948 Chr.X: 51895464-51895464 on Build GRCh38
rs989151549 Chr.X: 51895273-51895273 on Build GRCh38
rs781940383 Chr.X: 51895350-51895350 on Build GRCh38
rs782021678 Chr.X: 51895149-51895149 on Build GRCh38
rs944614430 Chr.X: 51895289-51895289 on Build GRCh38
rs900326660 Chr.X: 51895318-51895318 on Build GRCh38
rs782765353 Chr.X: 51895384-51895384 on Build GRCh38
rs1005773945 Chr.X: 51895109-51895109 on Build GRCh38

View Full Product Details