Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 89728825-89729398 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNCA
Entrez Gene ID
6622
Gene Name
synuclein alpha
Gene Aliases
NACP, PARK1, PARK4, PD1
UniGene
Hs.21374
Gene Chromosome Location
Chr.4: 89724099-89838324 on Build GRCh38
Transcripts
NM_000345.3 NM_001146054.1 NM_001146055.1 NM_007308.2 XM_011532208.2 XM_017008562.1 XM_017008563.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6392364 c.344A>C p.D115A Chr.4: 89729240-89729240 on Build GRCh38
5995375 c.321C>T p.A107A Chr.4: 89729263-89729263 on Build GRCh38
3606979 c.358C>T p.P120S Chr.4: 89729226-89729226 on Build GRCh38
5931543 c.350C>T p.P117L Chr.4: 89729234-89729234 on Build GRCh38
6168315 c.334A>T p.I112F Chr.4: 89729250-89729250 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764969376 Chr.4: 89729282-89729282 on Build GRCh38
rs367556974 Chr.4: 89729209-89729209 on Build GRCh38
rs754708820 Chr.4: 89729156-89729156 on Build GRCh38
rs752231442 Chr.4: 89729170-89729170 on Build GRCh38
rs763527339 Chr.4: 89729290-89729290 on Build GRCh38
rs149303475 Chr.4: 89729080-89729080 on Build GRCh38
rs372770165 Chr.4: 89729139-89729139 on Build GRCh38
rs759452333 Chr.4: 89729275-89729275 on Build GRCh38
rs770804878 Chr.4: 89729242-89729242 on Build GRCh38
rs977766243 Chr.4: 89729030-89729030 on Build GRCh38
rs191055637 Chr.4: 89729203-89729203 on Build GRCh38
rs144274752 Chr.4: 89729158-89729158 on Build GRCh38
rs570022321 Chr.4: 89729309-89729309 on Build GRCh38
rs913191658 Chr.4: 89728876-89728876 on Build GRCh38
rs768838511 Chr.4: 89729222-89729222 on Build GRCh38
rs762460433 Chr.4: 89729289-89729289 on Build GRCh38
rs1004834546 Chr.4: 89728850-89728850 on Build GRCh38
rs187036786 Chr.4: 89729141-89729141 on Build GRCh38
rs573271347 Chr.4: 89729371-89729371 on Build GRCh38
rs1019948898 Chr.4: 89729117-89729117 on Build GRCh38
rs945053040 Chr.4: 89729022-89729022 on Build GRCh38
rs113924942 Chr.4: 89728935-89728935 on Build GRCh38
rs753488004 Chr.4: 89729143-89729143 on Build GRCh38
rs971657126 Chr.4: 89728912-89728912 on Build GRCh38
rs1007063698 Chr.4: 89729048-89729048 on Build GRCh38
rs989190766 Chr.4: 89729005-89729005 on Build GRCh38
rs558772669 Chr.4: 89729331-89729331 on Build GRCh38
rs34511254 Chr.4: 89729030-89729030 on Build GRCh38
rs1033273943 Chr.4: 89729146-89729146 on Build GRCh38
rs868044896 Chr.4: 89729236-89729237 on Build GRCh38
rs925791697 Chr.4: 89728949-89728949 on Build GRCh38
rs754183675 Chr.4: 89729318-89729318 on Build GRCh38
rs752360805 Chr.4: 89729285-89729285 on Build GRCh38
rs546366532 Chr.4: 89729183-89729183 on Build GRCh38
rs537692319 Chr.4: 89729011-89729013 on Build GRCh38
rs900303735 Chr.4: 89729091-89729091 on Build GRCh38
rs944609323 Chr.4: 89729053-89729053 on Build GRCh38
rs372101729 Chr.4: 89729238-89729238 on Build GRCh38
rs976674262 Chr.4: 89728865-89728865 on Build GRCh38
rs1034520792 Chr.4: 89729311-89729311 on Build GRCh38
rs143084604 Chr.4: 89729233-89729233 on Build GRCh38
rs764422789 Chr.4: 89728878-89728878 on Build GRCh38
rs750899874 Chr.4: 89729202-89729202 on Build GRCh38
rs780731947 Chr.4: 89729137-89729137 on Build GRCh38
rs77010743 Chr.4: 89729103-89729103 on Build GRCh38
rs200712803 Chr.4: 89729021-89729022 on Build GRCh38
rs563591628 Chr.4: 89729081-89729081 on Build GRCh38
rs1049167065 Chr.4: 89729127-89729127 on Build GRCh38
rs1034495376 Chr.4: 89728971-89728971 on Build GRCh38
rs546025894 Chr.4: 89729140-89729140 on Build GRCh38
rs776535649 Chr.4: 89729270-89729270 on Build GRCh38
rs979032277 Chr.4: 89728930-89728930 on Build GRCh38
rs567122580 Chr.4: 89728999-89728999 on Build GRCh38
rs897443544 Chr.4: 89729022-89729022 on Build GRCh38
rs553372040 Chr.4: 89729019-89729019 on Build GRCh38
rs34638560 Chr.4: 89729321-89729321 on Build GRCh38
rs780419569 Chr.4: 89729218-89729218 on Build GRCh38
rs1024288001 Chr.4: 89729225-89729225 on Build GRCh38
rs574720169 Chr.4: 89729037-89729037 on Build GRCh38
rs138566779 Chr.4: 89728980-89728980 on Build GRCh38
rs760548842 Chr.4: 89729237-89729237 on Build GRCh38
rs370797923 Chr.4: 89729187-89729187 on Build GRCh38
rs916591060 Chr.4: 89729015-89729015 on Build GRCh38
rs145138372 Chr.4: 89729235-89729235 on Build GRCh38
rs756648675 Chr.4: 89729310-89729310 on Build GRCh38
rs749476922 Chr.4: 89729219-89729219 on Build GRCh38
rs1022076790 Chr.4: 89729373-89729373 on Build GRCh38
rs757901993 Chr.4: 89729182-89729182 on Build GRCh38
rs369670877 Chr.4: 89729039-89729039 on Build GRCh38
rs576925735 Chr.4: 89728899-89728899 on Build GRCh38
rs980697399 Chr.4: 89728924-89728924 on Build GRCh38
rs748759477 Chr.4: 89729150-89729150 on Build GRCh38
rs200056149 Chr.4: 89729217-89729217 on Build GRCh38
rs780583054 Chr.4: 89729315-89729315 on Build GRCh38
rs1011738500 Chr.4: 89729340-89729340 on Build GRCh38
rs1001760231 Chr.4: 89729288-89729288 on Build GRCh38
rs779158137 Chr.4: 89729226-89729226 on Build GRCh38

View Full Product Details