Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 25769871-25770467 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC17A1
Entrez Gene ID
6568
Gene Name
solute carrier family 17 member 1
Gene Aliases
NAPI-1, NPT-1, NPT1
UniGene
Hs.205816
Gene Chromosome Location
Chr.6: 25723744-25832108 on Build GRCh38
Transcripts
XM_017011201.1 NM_001286121.1 NM_005495.2 XM_011514211.1 XM_011514213.2 XM_011514214.2 XM_011514215.2 XM_011514217.2 XM_011514218.2 XM_011514219.2 XM_017010150.1 XM_017010151.1

Gene Symbol
SLC17A4
Entrez Gene ID
10050
Gene Name
solute carrier family 17 member 4
Gene Aliases
KAIA2138
UniGene
Hs.282931
Gene Chromosome Location
Chr.6: 25754673-25781175 on Build GRCh38
Transcripts
XM_017011201.1 NM_001286121.1 NM_005495.2 XM_011514211.1 XM_011514213.2 XM_011514214.2 XM_011514215.2 XM_011514217.2 XM_011514218.2 XM_011514219.2 XM_017010150.1 XM_017010151.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6331009 c.369G>T p.L123F Chr.6: 25770138-25770138 on Build GRCh38
7390170 c.614G>C p.G205A Chr.6: 25770466-25770466 on Build GRCh38
4489463 c.347C>T p.S116F Chr.6: 25770116-25770116 on Build GRCh38
3623344 c.580C>T p.P194S Chr.6: 25770432-25770432 on Build GRCh38
5404925 c.466C>T p.P156S Chr.6: 25770235-25770235 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113897781 Chr.6: 25770269-25770269 on Build GRCh38
rs141567358 Chr.6: 25770427-25770427 on Build GRCh38
rs762155902 Chr.6: 25770251-25770251 on Build GRCh38
rs780626791 Chr.6: 25770047-25770047 on Build GRCh38
rs766642305 Chr.6: 25770400-25770400 on Build GRCh38
rs745738447 Chr.6: 25770218-25770218 on Build GRCh38
rs925680389 Chr.6: 25770012-25770012 on Build GRCh38
rs868668864 Chr.6: 25770128-25770128 on Build GRCh38
rs553649027 Chr.6: 25770281-25770281 on Build GRCh38
rs768962705 Chr.6: 25770050-25770051 on Build GRCh38
rs753666751 Chr.6: 25770157-25770157 on Build GRCh38
rs749291828 Chr.6: 25770320-25770320 on Build GRCh38
rs766950278 Chr.6: 25769953-25769953 on Build GRCh38
rs776426818 Chr.6: 25770070-25770070 on Build GRCh38
rs911119771 Chr.6: 25770189-25770189 on Build GRCh38
rs747290493 Chr.6: 25770442-25770442 on Build GRCh38
rs978591318 Chr.6: 25770027-25770027 on Build GRCh38
rs550666765 Chr.6: 25770375-25770376 on Build GRCh38
rs771074520 Chr.6: 25770443-25770443 on Build GRCh38
rs756546634 Chr.6: 25770034-25770034 on Build GRCh38
rs772746932 Chr.6: 25770330-25770330 on Build GRCh38
rs142892037 Chr.6: 25770146-25770146 on Build GRCh38
rs35033101 Chr.6: 25770264-25770265 on Build GRCh38
rs751113057 Chr.6: 25770278-25770278 on Build GRCh38
rs1014434401 Chr.6: 25769978-25769978 on Build GRCh38
rs183805688 Chr.6: 25770327-25770327 on Build GRCh38
rs755291631 Chr.6: 25770270-25770270 on Build GRCh38
rs750336919 Chr.6: 25770355-25770355 on Build GRCh38
rs746740964 Chr.6: 25770208-25770208 on Build GRCh38
rs976785188 Chr.6: 25770078-25770078 on Build GRCh38
rs774433036 Chr.6: 25770249-25770249 on Build GRCh38
rs756706151 Chr.6: 25770168-25770168 on Build GRCh38
rs1056817319 Chr.6: 25770310-25770310 on Build GRCh38
rs756706714 Chr.6: 25770212-25770212 on Build GRCh38
rs759525451 Chr.6: 25770071-25770071 on Build GRCh38
rs750116956 Chr.6: 25770402-25770402 on Build GRCh38
rs373860279 Chr.6: 25770322-25770322 on Build GRCh38
rs755439421 Chr.6: 25770201-25770201 on Build GRCh38
rs779144350 Chr.6: 25770375-25770375 on Build GRCh38
rs750885204 Chr.6: 25770156-25770156 on Build GRCh38
rs760802010 Chr.6: 25770357-25770357 on Build GRCh38
rs769520943 Chr.6: 25770075-25770075 on Build GRCh38
rs371333508 Chr.6: 25770228-25770228 on Build GRCh38
rs774882083 Chr.6: 25770116-25770116 on Build GRCh38
rs779599106 Chr.6: 25770404-25770404 on Build GRCh38
rs762251390 Chr.6: 25770221-25770221 on Build GRCh38
rs759145589 Chr.6: 25770376-25770376 on Build GRCh38
rs745405816 Chr.6: 25770295-25770295 on Build GRCh38
rs2328893 Chr.6: 25770011-25770011 on Build GRCh38
rs755671020 Chr.6: 25770297-25770297 on Build GRCh38
rs199944557 Chr.6: 25770433-25770433 on Build GRCh38
rs150968968 Chr.6: 25770005-25770005 on Build GRCh38
rs772374172 Chr.6: 25770330-25770330 on Build GRCh38
rs768536621 Chr.6: 25770230-25770230 on Build GRCh38
rs773434791 Chr.6: 25770257-25770257 on Build GRCh38
rs897404046 Chr.6: 25769903-25769903 on Build GRCh38
rs754273256 Chr.6: 25770184-25770184 on Build GRCh38
rs773612789 Chr.6: 25770349-25770349 on Build GRCh38
rs199617227 Chr.6: 25770381-25770381 on Build GRCh38
rs955943854 Chr.6: 25770024-25770024 on Build GRCh38
rs764245081 Chr.6: 25770091-25770091 on Build GRCh38
rs777284176 Chr.6: 25770067-25770067 on Build GRCh38
rs955900185 Chr.6: 25770040-25770040 on Build GRCh38
rs780527241 Chr.6: 25770289-25770289 on Build GRCh38
rs779936950 Chr.6: 25770315-25770315 on Build GRCh38
rs550232446 Chr.6: 25769985-25769985 on Build GRCh38
rs140215842 Chr.6: 25770432-25770432 on Build GRCh38
rs766629413 Chr.6: 25770373-25770373 on Build GRCh38
rs539818480 Chr.6: 25769940-25769940 on Build GRCh38
rs766858049 Chr.6: 25769910-25769910 on Build GRCh38
rs780503942 Chr.6: 25770176-25770176 on Build GRCh38
rs1037739188 Chr.6: 25769916-25769916 on Build GRCh38
rs367896029 Chr.6: 25770089-25770089 on Build GRCh38
rs993185083 Chr.6: 25769924-25769924 on Build GRCh38
rs777256679 Chr.6: 25770204-25770204 on Build GRCh38
rs970712794 Chr.6: 25770286-25770286 on Build GRCh38
rs899170866 Chr.6: 25770221-25770221 on Build GRCh38
rs753663996 Chr.6: 25770423-25770423 on Build GRCh38
rs554420645 Chr.6: 25770214-25770214 on Build GRCh38
rs574585650 Chr.6: 25770260-25770260 on Build GRCh38
rs922573323 Chr.6: 25770125-25770125 on Build GRCh38
rs756938602 Chr.6: 25770284-25770284 on Build GRCh38
rs771877983 Chr.6: 25770050-25770050 on Build GRCh38
rs767134718 Chr.6: 25770399-25770399 on Build GRCh38
rs1023154428 Chr.6: 25769993-25769993 on Build GRCh38
rs954055808 Chr.6: 25770194-25770194 on Build GRCh38
rs374333664 Chr.6: 25770028-25770028 on Build GRCh38
rs751858810 Chr.6: 25770096-25770096 on Build GRCh38
rs150895210 Chr.6: 25770431-25770431 on Build GRCh38
rs988568973 Chr.6: 25770035-25770035 on Build GRCh38
rs962735567 Chr.6: 25770428-25770428 on Build GRCh38
rs144232109 Chr.6: 25770277-25770277 on Build GRCh38
rs1007956311 Chr.6: 25769907-25769907 on Build GRCh38
rs754951307 Chr.6: 25770425-25770425 on Build GRCh38
rs868183891 Chr.6: 25770115-25770115 on Build GRCh38
rs960209317 Chr.6: 25769962-25769962 on Build GRCh38
rs148724585 Chr.6: 25770397-25770397 on Build GRCh38
rs767705950 Chr.6: 25770148-25770148 on Build GRCh38
rs763123175 Chr.6: 25770087-25770087 on Build GRCh38
rs754432985 Chr.6: 25770265-25770265 on Build GRCh38
rs765451978 Chr.6: 25770376-25770376 on Build GRCh38
rs200323915 Chr.6: 25770323-25770323 on Build GRCh38
rs767440866 Chr.6: 25770252-25770252 on Build GRCh38
rs769754408 Chr.6: 25770225-25770225 on Build GRCh38
rs920566674 Chr.6: 25770445-25770445 on Build GRCh38
rs376970703 Chr.6: 25770372-25770372 on Build GRCh38
rs746476480 Chr.6: 25770068-25770068 on Build GRCh38
rs1013053685 Chr.6: 25770002-25770002 on Build GRCh38
rs775267322 Chr.6: 25770083-25770083 on Build GRCh38
rs755617921 Chr.6: 25770403-25770403 on Build GRCh38
rs766967507 Chr.6: 25770261-25770261 on Build GRCh38

View Full Product Details