Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 31865113-31865702 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC44A4
Entrez Gene ID
80736
Gene Name
solute carrier family 44 member 4
Gene Aliases
C6orf29, CTL4, NG22, TPPT
UniGene
Hs.335355
Gene Chromosome Location
Chr.6: 31863192-31879046 on Build GRCh38
Transcripts
NM_001178044.1 NM_001178045.1 NM_025257.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1487615 c.1747C>G p.R583G Chr.6: 31865328-31865328 on Build GRCh38
596022 c.1731G>A p.A577A Chr.6: 31865344-31865344 on Build GRCh38
5496511 c.1650A>G p.K550K Chr.6: 31865534-31865534 on Build GRCh38
6452451 c.1721C>T p.A574V Chr.6: 31865354-31865354 on Build GRCh38
741843 c.1698C>T p.Y566Y Chr.6: 31865377-31865377 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs977143069 Chr.6: 31865185-31865185 on Build GRCh38
rs747829201 Chr.6: 31865506-31865506 on Build GRCh38
rs374234974 Chr.6: 31865252-31865252 on Build GRCh38
rs1048307461 Chr.6: 31865273-31865273 on Build GRCh38
rs764434149 Chr.6: 31865548-31865548 on Build GRCh38
rs145759335 Chr.6: 31865377-31865377 on Build GRCh38
rs542943674 Chr.6: 31865186-31865186 on Build GRCh38
rs143476041 Chr.6: 31865323-31865323 on Build GRCh38
rs758266683 Chr.6: 31865598-31865598 on Build GRCh38
rs368786095 Chr.6: 31865514-31865514 on Build GRCh38
rs1003401478 Chr.6: 31865613-31865613 on Build GRCh38
rs56188416 Chr.6: 31865676-31865676 on Build GRCh38
rs796241321 Chr.6: 31865348-31865348 on Build GRCh38
rs188972261 Chr.6: 31865296-31865296 on Build GRCh38
rs1057214009 Chr.6: 31865627-31865627 on Build GRCh38
rs947554837 Chr.6: 31865156-31865156 on Build GRCh38
rs201906809 Chr.6: 31865299-31865299 on Build GRCh38
rs559342304 Chr.6: 31865428-31865428 on Build GRCh38
rs778675612 Chr.6: 31865587-31865587 on Build GRCh38
rs745310362 Chr.6: 31865672-31865672 on Build GRCh38
rs377400490 Chr.6: 31865422-31865422 on Build GRCh38
rs751879751 Chr.6: 31865553-31865553 on Build GRCh38
rs767096721 Chr.6: 31865326-31865326 on Build GRCh38
rs910339854 Chr.6: 31865231-31865231 on Build GRCh38
rs771743354 Chr.6: 31865507-31865507 on Build GRCh38
rs758234324 Chr.6: 31865610-31865610 on Build GRCh38
rs766891240 Chr.6: 31865423-31865423 on Build GRCh38
rs768147694 Chr.6: 31865139-31865139 on Build GRCh38
rs374858833 Chr.6: 31865208-31865208 on Build GRCh38
rs950629713 Chr.6: 31865398-31865398 on Build GRCh38
rs1010454621 Chr.6: 31865570-31865570 on Build GRCh38
rs192589833 Chr.6: 31865298-31865298 on Build GRCh38
rs375543593 Chr.6: 31865297-31865297 on Build GRCh38
rs752994111 Chr.6: 31865456-31865456 on Build GRCh38
rs765697281 Chr.6: 31865455-31865455 on Build GRCh38
rs776111648 Chr.6: 31865525-31865525 on Build GRCh38
rs768475513 Chr.6: 31865493-31865493 on Build GRCh38
rs763686046 Chr.6: 31865307-31865307 on Build GRCh38
rs763532689 Chr.6: 31865675-31865675 on Build GRCh38
rs138442998 Chr.6: 31865472-31865472 on Build GRCh38
rs774006990 Chr.6: 31865318-31865318 on Build GRCh38
rs770464000 Chr.6: 31865515-31865515 on Build GRCh38
rs924329630 Chr.6: 31865198-31865198 on Build GRCh38
rs757363607 Chr.6: 31865475-31865475 on Build GRCh38
rs770368841 Chr.6: 31865673-31865673 on Build GRCh38
rs943725927 Chr.6: 31865233-31865233 on Build GRCh38
rs112257041 Chr.6: 31865202-31865202 on Build GRCh38
rs116224609 Chr.6: 31865501-31865501 on Build GRCh38
rs141954433 Chr.6: 31865580-31865580 on Build GRCh38
rs760710846 Chr.6: 31865294-31865295 on Build GRCh38
rs547352490 Chr.6: 31865376-31865376 on Build GRCh38
rs754945392 Chr.6: 31865581-31865581 on Build GRCh38
rs150931872 Chr.6: 31865330-31865330 on Build GRCh38
rs753113597 Chr.6: 31865530-31865530 on Build GRCh38
rs763471143 Chr.6: 31865589-31865589 on Build GRCh38
rs775424340 Chr.6: 31865271-31865271 on Build GRCh38
rs749290460 Chr.6: 31865480-31865480 on Build GRCh38
rs759927500 Chr.6: 31865447-31865447 on Build GRCh38
rs372077719 Chr.6: 31865328-31865328 on Build GRCh38
rs772713361 Chr.6: 31865405-31865405 on Build GRCh38
rs138964997 Chr.6: 31865381-31865381 on Build GRCh38
rs778079636 Chr.6: 31865467-31865467 on Build GRCh38
rs199617182 Chr.6: 31865295-31865295 on Build GRCh38
rs559523018 Chr.6: 31865511-31865511 on Build GRCh38
rs778059469 Chr.6: 31865366-31865366 on Build GRCh38
rs116475370 Chr.6: 31865147-31865147 on Build GRCh38
rs775815963 Chr.6: 31865674-31865674 on Build GRCh38
rs757523398 Chr.6: 31865572-31865572 on Build GRCh38
rs755317277 Chr.6: 31865338-31865338 on Build GRCh38
rs939852159 Chr.6: 31865583-31865583 on Build GRCh38
rs376770297 Chr.6: 31865285-31865285 on Build GRCh38
rs777509034 Chr.6: 31865646-31865646 on Build GRCh38
rs1041133023 Chr.6: 31865234-31865234 on Build GRCh38
rs760145005 Chr.6: 31865526-31865526 on Build GRCh38
rs746727628 Chr.6: 31865653-31865653 on Build GRCh38
rs888338149 Chr.6: 31865578-31865578 on Build GRCh38
rs142009471 Chr.6: 31865344-31865344 on Build GRCh38
rs747032806 Chr.6: 31865375-31865375 on Build GRCh38
rs763191640 Chr.6: 31865535-31865535 on Build GRCh38
rs770553880 Chr.6: 31865661-31865661 on Build GRCh38
rs781399388 Chr.6: 31865476-31865476 on Build GRCh38
rs267600962 Chr.6: 31865601-31865601 on Build GRCh38
rs899485532 Chr.6: 31865251-31865251 on Build GRCh38
rs546801718 Chr.6: 31865295-31865296 on Build GRCh38
rs745870866 Chr.6: 31865300-31865300 on Build GRCh38
rs1021604144 Chr.6: 31865308-31865308 on Build GRCh38
rs776222148 Chr.6: 31865666-31865666 on Build GRCh38
rs879028015 Chr.6: 31865605-31865605 on Build GRCh38
rs772801407 Chr.6: 31865510-31865510 on Build GRCh38
rs1018374990 Chr.6: 31865337-31865337 on Build GRCh38
rs758542145 Chr.6: 31865360-31865360 on Build GRCh38
rs769843152 Chr.6: 31865301-31865301 on Build GRCh38
rs371746398 Chr.6: 31865527-31865527 on Build GRCh38
rs762774147 Chr.6: 31865304-31865304 on Build GRCh38
rs748439826 Chr.6: 31865345-31865345 on Build GRCh38
rs776954002 Chr.6: 31865296-31865296 on Build GRCh38
rs17207713 Chr.6: 31865444-31865444 on Build GRCh38
rs753117463 Chr.6: 31865302-31865303 on Build GRCh38
rs904496422 Chr.6: 31865626-31865626 on Build GRCh38
rs991740105 Chr.6: 31865146-31865146 on Build GRCh38
rs765451846 Chr.6: 31865299-31865300 on Build GRCh38
rs879077855 Chr.6: 31865604-31865604 on Build GRCh38
rs373514417 Chr.6: 31865385-31865385 on Build GRCh38
rs747996334 Chr.6: 31865608-31865608 on Build GRCh38
rs537294061 Chr.6: 31865577-31865577 on Build GRCh38
rs1043348825 Chr.6: 31865602-31865602 on Build GRCh38
rs749118738 Chr.6: 31865379-31865379 on Build GRCh38
rs539900493 Chr.6: 31865371-31865371 on Build GRCh38
rs771564309 Chr.6: 31865399-31865399 on Build GRCh38
rs758663666 Chr.6: 31865465-31865465 on Build GRCh38
rs760325737 Chr.6: 31865290-31865290 on Build GRCh38

View Full Product Details