Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 89637635-89638212 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
LYRM2
Entrez Gene ID
57226
Gene Name
LYR motif containing 2
Gene Aliases
DJ122O8.2
UniGene
Hs.177275
Gene Chromosome Location
Chr.6: 89632224-89638755 on Build GRCh38
Transcripts
NM_020466.4 NR_028493.1 NR_028494.1 NR_028495.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1082479 c.73_75delCTC p.L25delL Chr.6: 89637853-89637855 on Build GRCh38
5414918 c.46-9C>G p.? Chr.6: 89637891-89637891 on Build GRCh38
6705341 c.186+5G>A p.? Chr.6: 89637737-89637737 on Build GRCh38
6175087 c.169A>G p.K57E Chr.6: 89637759-89637759 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs34012596 Chr.6: 89637792-89637792 on Build GRCh38
rs780981536 Chr.6: 89637880-89637880 on Build GRCh38
rs371667119 Chr.6: 89637923-89637923 on Build GRCh38
rs201314231 Chr.6: 89637734-89637734 on Build GRCh38
rs571445546 Chr.6: 89638083-89638083 on Build GRCh38
rs756422281 Chr.6: 89637851-89637851 on Build GRCh38
rs964477063 Chr.6: 89638163-89638163 on Build GRCh38
rs762238545 Chr.6: 89637676-89637680 on Build GRCh38
rs747750812 Chr.6: 89637903-89637903 on Build GRCh38
rs185146188 Chr.6: 89637957-89637957 on Build GRCh38
rs1055889 Chr.6: 89637750-89637750 on Build GRCh38
rs11553069 Chr.6: 89637828-89637828 on Build GRCh38
rs773514024 Chr.6: 89637676-89637678 on Build GRCh38
rs142068097 Chr.6: 89637831-89637831 on Build GRCh38
rs370217589 Chr.6: 89637696-89637696 on Build GRCh38
rs748820584 Chr.6: 89638095-89638095 on Build GRCh38
rs535797482 Chr.6: 89638172-89638172 on Build GRCh38
rs768340009 Chr.6: 89637745-89637745 on Build GRCh38
rs765566928 Chr.6: 89637778-89637778 on Build GRCh38
rs370758481 Chr.6: 89637859-89637859 on Build GRCh38
rs756193283 Chr.6: 89637710-89637710 on Build GRCh38
rs748800623 Chr.6: 89637929-89637929 on Build GRCh38
rs796421809 Chr.6: 89638010-89638011 on Build GRCh38
rs773244062 Chr.6: 89638108-89638109 on Build GRCh38
rs779911780 Chr.6: 89637872-89637872 on Build GRCh38
rs545532317 Chr.6: 89637856-89637856 on Build GRCh38
rs910278734 Chr.6: 89638011-89638011 on Build GRCh38
rs1017885084 Chr.6: 89638155-89638155 on Build GRCh38
rs114784929 Chr.6: 89638062-89638062 on Build GRCh38
rs60578978 Chr.6: 89638028-89638028 on Build GRCh38
rs1008703314 Chr.6: 89638154-89638154 on Build GRCh38
rs151021837 Chr.6: 89637797-89637797 on Build GRCh38
rs201771180 Chr.6: 89637804-89637804 on Build GRCh38
rs764905689 Chr.6: 89637853-89637855 on Build GRCh38
rs759214824 Chr.6: 89637735-89637736 on Build GRCh38
rs758333754 Chr.6: 89637866-89637866 on Build GRCh38
rs773644699 Chr.6: 89637689-89637689 on Build GRCh38
rs973435454 Chr.6: 89638149-89638149 on Build GRCh38
rs554165462 Chr.6: 89638188-89638188 on Build GRCh38
rs543049656 Chr.6: 89637998-89637998 on Build GRCh38
rs779393677 Chr.6: 89638032-89638032 on Build GRCh38
rs746814594 Chr.6: 89637877-89637877 on Build GRCh38
rs527647342 Chr.6: 89637980-89637980 on Build GRCh38
rs765730789 Chr.6: 89637681-89637681 on Build GRCh38
rs1021358019 Chr.6: 89638067-89638067 on Build GRCh38
rs934899858 Chr.6: 89638043-89638043 on Build GRCh38
rs978440189 Chr.6: 89638022-89638022 on Build GRCh38
rs1025374142 Chr.6: 89637965-89637965 on Build GRCh38
rs943710036 Chr.6: 89638122-89638122 on Build GRCh38
rs1048534232 Chr.6: 89637933-89637933 on Build GRCh38
rs1026148901 Chr.6: 89638132-89638132 on Build GRCh38
rs769238833 Chr.6: 89637774-89637774 on Build GRCh38
rs868649112 Chr.6: 89637748-89637749 on Build GRCh38
rs372777610 Chr.6: 89638041-89638042 on Build GRCh38
rs775181637 Chr.6: 89637857-89637859 on Build GRCh38
rs369897343 Chr.6: 89638020-89638021 on Build GRCh38
rs754703374 Chr.6: 89637879-89637879 on Build GRCh38
rs778010905 Chr.6: 89637727-89637727 on Build GRCh38
rs767996051 Chr.6: 89637822-89637822 on Build GRCh38
rs755622318 Chr.6: 89637987-89637987 on Build GRCh38
rs868732212 Chr.6: 89637698-89637698 on Build GRCh38
rs776331259 Chr.6: 89637747-89637747 on Build GRCh38
rs772438014 Chr.6: 89637676-89637679 on Build GRCh38
rs556752104 Chr.6: 89637817-89637817 on Build GRCh38
rs549692214 Chr.6: 89638070-89638070 on Build GRCh38
rs543524750 Chr.6: 89637675-89637676 on Build GRCh38
rs768767344 Chr.6: 89637676-89637676 on Build GRCh38
rs746828223 Chr.6: 89637743-89637743 on Build GRCh38
rs565445135 Chr.6: 89638164-89638164 on Build GRCh38
rs868772661 Chr.6: 89638112-89638112 on Build GRCh38
rs752814110 Chr.6: 89637709-89637709 on Build GRCh38
rs143603572 Chr.6: 89637674-89637674 on Build GRCh38
rs534210657 Chr.6: 89637759-89637759 on Build GRCh38
rs751904706 Chr.6: 89637700-89637700 on Build GRCh38
rs181413728 Chr.6: 89637781-89637781 on Build GRCh38
rs763382681 Chr.6: 89637693-89637693 on Build GRCh38
rs895220702 Chr.6: 89637939-89637939 on Build GRCh38
rs925635829 Chr.6: 89638040-89638041 on Build GRCh38
rs377405597 Chr.6: 89637848-89637848 on Build GRCh38
rs758296310 Chr.6: 89637739-89637739 on Build GRCh38
rs767902407 Chr.6: 89637703-89637703 on Build GRCh38
rs907123422 Chr.6: 89637982-89637982 on Build GRCh38
rs772731688 Chr.6: 89637777-89637777 on Build GRCh38
rs1037204708 Chr.6: 89637907-89637907 on Build GRCh38
rs964833057 Chr.6: 89638011-89638013 on Build GRCh38
rs953766923 Chr.6: 89638069-89638069 on Build GRCh38
rs950172717 Chr.6: 89638079-89638079 on Build GRCh38
rs764225641 Chr.6: 89637853-89637853 on Build GRCh38
rs189856384 Chr.6: 89638118-89638118 on Build GRCh38
rs931499862 Chr.6: 89638179-89638179 on Build GRCh38
rs201265383 Chr.6: 89637908-89637908 on Build GRCh38
rs115085953 Chr.6: 89637730-89637730 on Build GRCh38
rs759780507 Chr.6: 89637741-89637741 on Build GRCh38
rs188665632 Chr.6: 89637694-89637694 on Build GRCh38
rs772751532 Chr.6: 89637920-89637920 on Build GRCh38
rs868749996 Chr.6: 89637740-89637740 on Build GRCh38
rs375585665 Chr.6: 89637891-89637891 on Build GRCh38
rs532103278 Chr.6: 89638110-89638110 on Build GRCh38

View Full Product Details