Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 149011903-149012483 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PDIA4
Entrez Gene ID
9601
Gene Name
protein disulfide isomerase family A member 4
Gene Aliases
ERP70, ERP72, ERp-72
UniGene
Hs.93659
Gene Chromosome Location
Chr.7: 149003054-149028690 on Build GRCh38
Transcripts
NM_004911.4 XM_006716185.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1187458 c.658G>T p.A220S Chr.7: 149012317-149012317 on Build GRCh38
4164166 c.703G>T p.V235F Chr.7: 149012272-149012272 on Build GRCh38
1235210 c.844C>T p.Q282* Chr.7: 149011981-149011981 on Build GRCh38
4388677 c.802G>A p.G268S Chr.7: 149012173-149012173 on Build GRCh38
1087069 c.709G>A p.A237T Chr.7: 149012266-149012266 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773084572 Chr.7: 149012362-149012362 on Build GRCh38
rs781689302 Chr.7: 149012149-149012149 on Build GRCh38
rs61739280 Chr.7: 149012261-149012261 on Build GRCh38
rs148042089 Chr.7: 149012418-149012418 on Build GRCh38
rs534122693 Chr.7: 149011976-149011976 on Build GRCh38
rs781646528 Chr.7: 149012238-149012238 on Build GRCh38
rs377657545 Chr.7: 149012381-149012381 on Build GRCh38
rs756102159 Chr.7: 149012079-149012080 on Build GRCh38
rs780619263 Chr.7: 149012317-149012317 on Build GRCh38
rs771988421 Chr.7: 149012346-149012346 on Build GRCh38
rs567468691 Chr.7: 149012333-149012333 on Build GRCh38
rs894543063 Chr.7: 149012380-149012380 on Build GRCh38
rs372415891 Chr.7: 149011928-149011928 on Build GRCh38
rs145536019 Chr.7: 149012313-149012313 on Build GRCh38
rs964175581 Chr.7: 149012440-149012440 on Build GRCh38
rs138974987 Chr.7: 149011949-149011949 on Build GRCh38
rs764047250 Chr.7: 149011960-149011960 on Build GRCh38
rs764171372 Chr.7: 149012295-149012295 on Build GRCh38
rs558794904 Chr.7: 149012335-149012335 on Build GRCh38
rs773254296 Chr.7: 149012270-149012270 on Build GRCh38
rs779492257 Chr.7: 149012260-149012260 on Build GRCh38
rs372023035 Chr.7: 149012266-149012266 on Build GRCh38
rs1013121072 Chr.7: 149012400-149012400 on Build GRCh38
rs920451617 Chr.7: 149012225-149012225 on Build GRCh38
rs375136880 Chr.7: 149012269-149012269 on Build GRCh38
rs755138821 Chr.7: 149012105-149012105 on Build GRCh38
rs370665535 Chr.7: 149012037-149012037 on Build GRCh38
rs764379611 Chr.7: 149012222-149012222 on Build GRCh38
rs971628686 Chr.7: 149012189-149012189 on Build GRCh38
rs533578464 Chr.7: 149012332-149012332 on Build GRCh38
rs2290970 Chr.7: 149012415-149012415 on Build GRCh38
rs1018406366 Chr.7: 149012428-149012428 on Build GRCh38
rs770414968 Chr.7: 149012000-149012000 on Build GRCh38
rs145307896 Chr.7: 149012248-149012248 on Build GRCh38
rs1014601793 Chr.7: 149012431-149012431 on Build GRCh38
rs747414354 Chr.7: 149012146-149012146 on Build GRCh38
rs779279777 Chr.7: 149011984-149011984 on Build GRCh38
rs945929299 Chr.7: 149012075-149012075 on Build GRCh38
rs371382515 Chr.7: 149012167-149012167 on Build GRCh38
rs368013782 Chr.7: 149012281-149012281 on Build GRCh38
rs148817386 Chr.7: 149012318-149012318 on Build GRCh38
rs149365261 Chr.7: 149011975-149011975 on Build GRCh38
rs775364359 Chr.7: 149012287-149012287 on Build GRCh38
rs931924104 Chr.7: 149012013-149012013 on Build GRCh38
rs779226690 Chr.7: 149012127-149012127 on Build GRCh38
rs910977714 Chr.7: 149012239-149012239 on Build GRCh38
rs11546292 Chr.7: 149012321-149012321 on Build GRCh38
rs79600968 Chr.7: 149012026-149012026 on Build GRCh38
rs767572743 Chr.7: 149012048-149012048 on Build GRCh38
rs867465730 Chr.7: 149011954-149011954 on Build GRCh38
rs1004430493 Chr.7: 149012426-149012426 on Build GRCh38
rs570306170 Chr.7: 149012051-149012051 on Build GRCh38
rs1020227826 Chr.7: 149012058-149012058 on Build GRCh38
rs751505200 Chr.7: 149011962-149011962 on Build GRCh38
rs371332924 Chr.7: 149012001-149012001 on Build GRCh38
rs762917426 Chr.7: 149012394-149012394 on Build GRCh38
rs759666998 Chr.7: 149012190-149012190 on Build GRCh38
rs370450689 Chr.7: 149011973-149011973 on Build GRCh38
rs906518164 Chr.7: 149012404-149012404 on Build GRCh38
rs763122880 Chr.7: 149012293-149012293 on Build GRCh38
rs191155547 Chr.7: 149012267-149012267 on Build GRCh38
rs768818391 Chr.7: 149012396-149012396 on Build GRCh38
rs762583788 Chr.7: 149012166-149012166 on Build GRCh38
rs754657819 Chr.7: 149011978-149011978 on Build GRCh38
rs752950039 Chr.7: 149012196-149012196 on Build GRCh38
rs1054297876 Chr.7: 149012137-149012137 on Build GRCh38
rs984268183 Chr.7: 149012226-149012226 on Build GRCh38
rs555842203 Chr.7: 149012014-149012014 on Build GRCh38
rs1000685174 Chr.7: 149012134-149012134 on Build GRCh38
rs746804146 Chr.7: 149012375-149012375 on Build GRCh38
rs761711328 Chr.7: 149012044-149012044 on Build GRCh38
rs769339244 Chr.7: 149012156-149012156 on Build GRCh38
rs9918632 Chr.7: 149012442-149012442 on Build GRCh38
rs545639058 Chr.7: 149011989-149011989 on Build GRCh38
rs201276356 Chr.7: 149011971-149011971 on Build GRCh38
rs773962719 Chr.7: 149012170-149012170 on Build GRCh38
rs375265274 Chr.7: 149012136-149012136 on Build GRCh38
rs61739279 Chr.7: 149011985-149011985 on Build GRCh38
rs141864886 Chr.7: 149012157-149012157 on Build GRCh38
rs1046863815 Chr.7: 149012357-149012357 on Build GRCh38
rs758147565 Chr.7: 149012130-149012130 on Build GRCh38
rs961825633 Chr.7: 149012218-149012218 on Build GRCh38
rs200863165 Chr.7: 149012145-149012145 on Build GRCh38
rs757639070 Chr.7: 149012232-149012232 on Build GRCh38
rs767932370 Chr.7: 149012309-149012309 on Build GRCh38
rs971425159 Chr.7: 149012152-149012152 on Build GRCh38
rs374768570 Chr.7: 149012194-149012194 on Build GRCh38
rs745330359 Chr.7: 149012150-149012150 on Build GRCh38
rs762690248 Chr.7: 149011932-149011932 on Build GRCh38
rs150793763 Chr.7: 149012174-149012174 on Build GRCh38
rs760761181 Chr.7: 149012272-149012272 on Build GRCh38
rs201201125 Chr.7: 149011987-149011987 on Build GRCh38
rs757583934 Chr.7: 149012299-149012299 on Build GRCh38
rs747844006 Chr.7: 149012342-149012342 on Build GRCh38
rs973527006 Chr.7: 149012224-149012224 on Build GRCh38
rs367839477 Chr.7: 149012199-149012199 on Build GRCh38
rs971681074 Chr.7: 149012423-149012423 on Build GRCh38
rs147180118 Chr.7: 149012251-149012251 on Build GRCh38
rs773306098 Chr.7: 149012179-149012179 on Build GRCh38
rs762789882 Chr.7: 149012022-149012022 on Build GRCh38
rs775030714 Chr.7: 149012021-149012021 on Build GRCh38
rs752930266 Chr.7: 149012378-149012379 on Build GRCh38
rs765597707 Chr.7: 149012304-149012305 on Build GRCh38
rs112506294 Chr.7: 149012006-149012006 on Build GRCh38
rs776651373 Chr.7: 149012389-149012389 on Build GRCh38
rs775581188 Chr.7: 149012429-149012429 on Build GRCh38
rs148084821 Chr.7: 149011999-149011999 on Build GRCh38
rs766352902 Chr.7: 149012282-149012282 on Build GRCh38
rs371907961 Chr.7: 149012298-149012298 on Build GRCh38
rs139730063 Chr.7: 149012231-149012231 on Build GRCh38
rs771363149 Chr.7: 149012148-149012148 on Build GRCh38
rs756558717 Chr.7: 149012315-149012315 on Build GRCh38
rs140513316 Chr.7: 149012254-149012254 on Build GRCh38
rs372962795 Chr.7: 149012010-149012010 on Build GRCh38
rs369248381 Chr.7: 149012178-149012178 on Build GRCh38

View Full Product Details