Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 192159092-192159526 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS18
Entrez Gene ID
64407
Gene Name
regulator of G-protein signaling 18
Gene Aliases
RGS13
UniGene
Hs.440890
Gene Chromosome Location
Chr.1: 192158462-192187172 on Build GRCh38
Transcripts
NM_130782.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6765856 c.122C>A p.A41D Chr.1: 192159222-192159222 on Build GRCh38
6483252 c.194G>T p.R65I Chr.1: 192159294-192159294 on Build GRCh38
7230880 c.158A>T p.Q53L Chr.1: 192159258-192159258 on Build GRCh38
5960328 c.179A>G p.D60G Chr.1: 192159279-192159279 on Build GRCh38
3480540 c.127G>A p.E43K Chr.1: 192159227-192159227 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs934448021 Chr.1: 192159451-192159451 on Build GRCh38
rs35623527 Chr.1: 192159233-192159233 on Build GRCh38
rs779052527 Chr.1: 192159203-192159203 on Build GRCh38
rs866231531 Chr.1: 192159431-192159431 on Build GRCh38
rs767577887 Chr.1: 192159284-192159284 on Build GRCh38
rs1031841790 Chr.1: 192159460-192159460 on Build GRCh38
rs201783745 Chr.1: 192159285-192159285 on Build GRCh38
rs774253109 Chr.1: 192159266-192159266 on Build GRCh38
rs765096530 Chr.1: 192159287-192159287 on Build GRCh38
rs1026522739 Chr.1: 192159311-192159311 on Build GRCh38
rs772188208 Chr.1: 192159369-192159369 on Build GRCh38
rs928493558 Chr.1: 192159434-192159441 on Build GRCh38
rs755651267 Chr.1: 192159282-192159282 on Build GRCh38
rs947436663 Chr.1: 192159491-192159491 on Build GRCh38
rs984450356 Chr.1: 192159357-192159357 on Build GRCh38
rs979251637 Chr.1: 192159393-192159393 on Build GRCh38
rs547490682 Chr.1: 192159205-192159205 on Build GRCh38
rs34746017 Chr.1: 192159293-192159293 on Build GRCh38
rs562038493 Chr.1: 192159177-192159177 on Build GRCh38
rs770134901 Chr.1: 192159254-192159254 on Build GRCh38
rs948656702 Chr.1: 192159387-192159387 on Build GRCh38
rs893241718 Chr.1: 192159484-192159484 on Build GRCh38
rs372304453 Chr.1: 192159193-192159193 on Build GRCh38
rs981294795 Chr.1: 192159426-192159426 on Build GRCh38
rs527712482 Chr.1: 192159191-192159191 on Build GRCh38
rs1022907322 Chr.1: 192159340-192159340 on Build GRCh38
rs1045563965 Chr.1: 192159133-192159133 on Build GRCh38
rs758365296 Chr.1: 192159291-192159291 on Build GRCh38
rs1052952598 Chr.1: 192159452-192159452 on Build GRCh38
rs746698535 Chr.1: 192159222-192159222 on Build GRCh38
rs751827669 Chr.1: 192159172-192159172 on Build GRCh38
rs764127073 Chr.1: 192159170-192159170 on Build GRCh38
rs879034383 Chr.1: 192159118-192159118 on Build GRCh38
rs749771617 Chr.1: 192159255-192159255 on Build GRCh38
rs748236144 Chr.1: 192159365-192159365 on Build GRCh38
rs780346146 Chr.1: 192159318-192159318 on Build GRCh38
rs907005442 Chr.1: 192159134-192159134 on Build GRCh38
rs777454921 Chr.1: 192159298-192159298 on Build GRCh38
rs529567476 Chr.1: 192159123-192159123 on Build GRCh38
rs756519523 Chr.1: 192159315-192159315 on Build GRCh38
rs12085113 Chr.1: 192159378-192159378 on Build GRCh38
rs368444983 Chr.1: 192159253-192159253 on Build GRCh38
rs762100879 Chr.1: 192159278-192159278 on Build GRCh38
rs750973689 Chr.1: 192159295-192159295 on Build GRCh38
rs752734136 Chr.1: 192159290-192159290 on Build GRCh38
rs145426547 Chr.1: 192159308-192159308 on Build GRCh38
rs752245561 Chr.1: 192159224-192159224 on Build GRCh38
rs768744391 Chr.1: 192159264-192159264 on Build GRCh38
rs201336568 Chr.1: 192159355-192159355 on Build GRCh38
rs890067237 Chr.1: 192159247-192159247 on Build GRCh38
rs769085932 Chr.1: 192159328-192159328 on Build GRCh38
rs376472982 Chr.1: 192159234-192159234 on Build GRCh38
rs112851731 Chr.1: 192159499-192159499 on Build GRCh38
rs745502461 Chr.1: 192159310-192159310 on Build GRCh38
rs756775100 Chr.1: 192159302-192159302 on Build GRCh38
rs1053577044 Chr.1: 192159179-192159179 on Build GRCh38

View Full Product Details