Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 27963504-27963918 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PTHLH
Entrez Gene ID
5744
Gene Name
parathyroid hormone like hormone
Gene Aliases
BDE2, HHM, PLP, PTHR, PTHRP
UniGene
Hs.591159
Gene Chromosome Location
Chr.12: 27958084-27974187 on Build GRCh38
Transcripts
NM_002820.2 NM_198964.1 NM_198965.1 NM_198966.1 XM_011520774.2 XM_011520775.2 XM_017019674.1 XM_017019675.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1476478 c.221C>T p.A74V Chr.12: 27963651-27963651 on Build GRCh38
4041275 c.193G>A p.A65T Chr.12: 27963679-27963679 on Build GRCh38
4892619 c.292G>A p.D98N Chr.12: 27963580-27963580 on Build GRCh38
4146981 c.327C>A p.N109K Chr.12: 27963545-27963545 on Build GRCh38
7168541 c.367G>A p.G123R Chr.12: 27963505-27963505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779114545 Chr.12: 27963622-27963622 on Build GRCh38
rs775088379 Chr.12: 27963651-27963651 on Build GRCh38
rs762580267 Chr.12: 27963654-27963654 on Build GRCh38
rs771833243 Chr.12: 27963542-27963542 on Build GRCh38
rs779090312 Chr.12: 27963782-27963782 on Build GRCh38
rs139412701 Chr.12: 27963611-27963611 on Build GRCh38
rs932755478 Chr.12: 27963645-27963645 on Build GRCh38
rs200580021 Chr.12: 27963722-27963722 on Build GRCh38
rs762237637 Chr.12: 27963679-27963679 on Build GRCh38
rs894039320 Chr.12: 27963794-27963794 on Build GRCh38
rs751930749 Chr.12: 27963585-27963585 on Build GRCh38
rs946948096 Chr.12: 27963823-27963823 on Build GRCh38
rs757041118 Chr.12: 27963821-27963821 on Build GRCh38
rs774207195 Chr.12: 27963672-27963672 on Build GRCh38
rs200164918 Chr.12: 27963758-27963758 on Build GRCh38
rs374868158 Chr.12: 27963894-27963894 on Build GRCh38
rs767816700 Chr.12: 27963708-27963708 on Build GRCh38
rs909791349 Chr.12: 27963552-27963552 on Build GRCh38
rs770689016 Chr.12: 27963626-27963626 on Build GRCh38
rs762739753 Chr.12: 27963577-27963577 on Build GRCh38
rs78410907 Chr.12: 27963530-27963530 on Build GRCh38
rs763783691 Chr.12: 27963668-27963668 on Build GRCh38
rs143336733 Chr.12: 27963834-27963834 on Build GRCh38
rs757711589 Chr.12: 27963594-27963594 on Build GRCh38
rs781557987 Chr.12: 27963524-27963524 on Build GRCh38
rs772943331 Chr.12: 27963549-27963549 on Build GRCh38
rs753187634 Chr.12: 27963639-27963639 on Build GRCh38
rs763857888 Chr.12: 27963584-27963584 on Build GRCh38
rs267606986 Chr.12: 27963741-27963741 on Build GRCh38
rs902838391 Chr.12: 27963833-27963833 on Build GRCh38
rs201319886 Chr.12: 27963705-27963705 on Build GRCh38
rs1039945362 Chr.12: 27963873-27963873 on Build GRCh38
rs267606985 Chr.12: 27963693-27963693 on Build GRCh38
rs752362312 Chr.12: 27963572-27963572 on Build GRCh38
rs769825766 Chr.12: 27963532-27963532 on Build GRCh38
rs756191741 Chr.12: 27963600-27963600 on Build GRCh38
rs1056815666 Chr.12: 27963887-27963887 on Build GRCh38
rs141989276 Chr.12: 27963534-27963534 on Build GRCh38
rs149204360 Chr.12: 27963554-27963554 on Build GRCh38
rs769321368 Chr.12: 27963647-27963647 on Build GRCh38
rs780046816 Chr.12: 27963601-27963601 on Build GRCh38
rs770832859 Chr.12: 27963558-27963558 on Build GRCh38
rs868117405 Chr.12: 27963859-27963859 on Build GRCh38
rs998897168 Chr.12: 27963860-27963860 on Build GRCh38
rs746747626 Chr.12: 27963624-27963624 on Build GRCh38
rs779315594 Chr.12: 27963598-27963598 on Build GRCh38
rs753851601 Chr.12: 27963731-27963731 on Build GRCh38
rs866890264 Chr.12: 27963605-27963605 on Build GRCh38
rs916614518 Chr.12: 27963568-27963568 on Build GRCh38
rs745833158 Chr.12: 27963644-27963644 on Build GRCh38
rs373901355 Chr.12: 27963533-27963533 on Build GRCh38
rs764720667 Chr.12: 27963571-27963571 on Build GRCh38
rs750919798 Chr.12: 27963719-27963719 on Build GRCh38
rs755154380 Chr.12: 27963616-27963616 on Build GRCh38
rs748432222 Chr.12: 27963790-27963790 on Build GRCh38
rs766903878 Chr.12: 27963728-27963728 on Build GRCh38
rs976794916 Chr.12: 27963595-27963595 on Build GRCh38
rs1042684431 Chr.12: 27963824-27963824 on Build GRCh38
rs951205207 Chr.12: 27963528-27963528 on Build GRCh38
rs768058197 Chr.12: 27963596-27963596 on Build GRCh38
rs552484387 Chr.12: 27963567-27963567 on Build GRCh38

View Full Product Details