Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 92107583-92108135 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD8
Entrez Gene ID
284697
Gene Name
BTB domain containing 8
Gene Aliases
-
UniGene
Hs.676102
Gene Chromosome Location
Chr.1: 92080305-92147840 on Build GRCh38
Transcripts
NM_183242.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7252028 c.566C>T p.A189V Chr.1: 92107905-92107905 on Build GRCh38
7328514 c.637G>C p.D213H Chr.1: 92107976-92107976 on Build GRCh38
5801834 c.596delT p.Y200fs*2 Chr.1: 92107935-92107935 on Build GRCh38
7252027 c.566C>T p.A189V Chr.1: 92107905-92107905 on Build GRCh38
2259904 c.661A>T p.R221W Chr.1: 92108000-92108000 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs200905053 Chr.1: 92107712-92107712 on Build GRCh38
rs1027649770 Chr.1: 92108066-92108066 on Build GRCh38
rs781417090 Chr.1: 92107873-92107873 on Build GRCh38
rs138254677 Chr.1: 92108043-92108045 on Build GRCh38
rs569746380 Chr.1: 92108076-92108076 on Build GRCh38
rs150510309 Chr.1: 92107741-92107741 on Build GRCh38
rs754110181 Chr.1: 92107911-92107911 on Build GRCh38
rs139440509 Chr.1: 92108007-92108007 on Build GRCh38
rs767244571 Chr.1: 92107834-92107834 on Build GRCh38
rs954465580 Chr.1: 92107746-92107746 on Build GRCh38
rs778472115 Chr.1: 92107987-92107987 on Build GRCh38
rs781204303 Chr.1: 92107855-92107855 on Build GRCh38
rs763423870 Chr.1: 92107884-92107884 on Build GRCh38
rs941645773 Chr.1: 92107657-92107657 on Build GRCh38
rs778279023 Chr.1: 92107772-92107772 on Build GRCh38
rs759554363 Chr.1: 92108041-92108041 on Build GRCh38
rs201296940 Chr.1: 92108030-92108030 on Build GRCh38
rs1030102173 Chr.1: 92107742-92107742 on Build GRCh38
rs11166163 Chr.1: 92108113-92108113 on Build GRCh38
rs765167143 Chr.1: 92108045-92108045 on Build GRCh38
rs746058441 Chr.1: 92108000-92108000 on Build GRCh38
rs534331398 Chr.1: 92107651-92107651 on Build GRCh38
rs757268870 Chr.1: 92107994-92107994 on Build GRCh38
rs773745854 Chr.1: 92108018-92108018 on Build GRCh38
rs753143083 Chr.1: 92107918-92107920 on Build GRCh38
rs752569392 Chr.1: 92108046-92108046 on Build GRCh38
rs186982005 Chr.1: 92108014-92108014 on Build GRCh38
rs866622504 Chr.1: 92107918-92107918 on Build GRCh38
rs758595751 Chr.1: 92107849-92107849 on Build GRCh38
rs142006093 Chr.1: 92107623-92107623 on Build GRCh38
rs754915147 Chr.1: 92107964-92107964 on Build GRCh38
rs760617811 Chr.1: 92107954-92107954 on Build GRCh38
rs995721126 Chr.1: 92108047-92108047 on Build GRCh38
rs1017237592 Chr.1: 92108043-92108043 on Build GRCh38
rs55792020 Chr.1: 92107882-92107882 on Build GRCh38
rs182850342 Chr.1: 92107703-92107703 on Build GRCh38
rs781542833 Chr.1: 92107995-92107995 on Build GRCh38
rs970036099 Chr.1: 92107854-92107854 on Build GRCh38
rs375512920 Chr.1: 92108035-92108035 on Build GRCh38
rs1052929313 Chr.1: 92107909-92107909 on Build GRCh38
rs769139261 Chr.1: 92108017-92108017 on Build GRCh38
rs746750030 Chr.1: 92107653-92107654 on Build GRCh38
rs897299589 Chr.1: 92108036-92108036 on Build GRCh38
rs373224920 Chr.1: 92107922-92107922 on Build GRCh38
rs114520917 Chr.1: 92107847-92107847 on Build GRCh38
rs777887509 Chr.1: 92107856-92107856 on Build GRCh38
rs756497617 Chr.1: 92107986-92107987 on Build GRCh38
rs1034846545 Chr.1: 92107634-92107634 on Build GRCh38
rs372571918 Chr.1: 92107836-92107836 on Build GRCh38
rs747754395 Chr.1: 92108025-92108025 on Build GRCh38
rs369715107 Chr.1: 92107675-92107678 on Build GRCh38
rs780343926 Chr.1: 92108013-92108013 on Build GRCh38
rs768069019 Chr.1: 92107904-92107904 on Build GRCh38
rs546147329 Chr.1: 92107822-92107822 on Build GRCh38
rs771145690 Chr.1: 92107865-92107865 on Build GRCh38
rs548239298 Chr.1: 92108032-92108032 on Build GRCh38
rs766250060 Chr.1: 92107958-92107958 on Build GRCh38
rs746287892 Chr.1: 92107875-92107875 on Build GRCh38
rs201802045 Chr.1: 92107836-92107836 on Build GRCh38
rs776017178 Chr.1: 92107883-92107883 on Build GRCh38
rs980066663 Chr.1: 92108092-92108092 on Build GRCh38
rs750033221 Chr.1: 92107940-92107940 on Build GRCh38
rs373753138 Chr.1: 92108058-92108058 on Build GRCh38
rs756791621 Chr.1: 92107761-92107761 on Build GRCh38
rs376250311 Chr.1: 92107985-92107985 on Build GRCh38
rs757394987 Chr.1: 92108050-92108050 on Build GRCh38
rs762912503 Chr.1: 92108070-92108070 on Build GRCh38
rs747218663 Chr.1: 92107859-92107859 on Build GRCh38
rs369473873 Chr.1: 92107908-92107908 on Build GRCh38
rs1016191620 Chr.1: 92107820-92107820 on Build GRCh38
rs770138731 Chr.1: 92107880-92107880 on Build GRCh38
rs995833457 Chr.1: 92107716-92107716 on Build GRCh38
rs1042705741 Chr.1: 92107639-92107639 on Build GRCh38
rs745575136 Chr.1: 92107841-92107841 on Build GRCh38
rs753875792 Chr.1: 92107962-92107962 on Build GRCh38
rs369895599 Chr.1: 92108029-92108029 on Build GRCh38
rs370358765 Chr.1: 92107986-92107986 on Build GRCh38
rs1007227516 Chr.1: 92107750-92107750 on Build GRCh38
rs376352986 Chr.1: 92107837-92107837 on Build GRCh38
rs1051384964 Chr.1: 92108021-92108021 on Build GRCh38
rs767142009 Chr.1: 92107932-92107932 on Build GRCh38
rs961628214 Chr.1: 92107825-92107825 on Build GRCh38
rs747769430 Chr.1: 92107802-92107802 on Build GRCh38
rs1010800975 Chr.1: 92108068-92108068 on Build GRCh38

View Full Product Details