Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50349257-50349851 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NPRL2
Entrez Gene ID
10641
Gene Name
NPR2-like, GATOR1 complex subunit
Gene Aliases
NPR2, NPR2L, TUSC4
UniGene
Hs.437083
Gene Chromosome Location
Chr.3: 50347487-50351055 on Build GRCh38
Transcripts
NM_006545.4 XM_005264808.4 XM_011533288.2 XM_017005555.1 XM_017005556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7133370 c.412G>A p.E138K Chr.3: 50349422-50349422 on Build GRCh38
5899440 c.409G>A p.E137K Chr.3: 50349425-50349425 on Build GRCh38
4707881 c.355G>A p.V119M Chr.3: 50349479-50349479 on Build GRCh38
1424180 c.324T>C p.Y108Y Chr.3: 50349680-50349680 on Build GRCh38
1670221 c.352T>G p.F118V Chr.3: 50349482-50349482 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138670017 Chr.3: 50349398-50349398 on Build GRCh38
rs752423985 Chr.3: 50349791-50349791 on Build GRCh38
rs368301992 Chr.3: 50349639-50349639 on Build GRCh38
rs759397803 Chr.3: 50349631-50349631 on Build GRCh38
rs759328601 Chr.3: 50349776-50349776 on Build GRCh38
rs748413371 Chr.3: 50349721-50349721 on Build GRCh38
rs761147330 Chr.3: 50349655-50349656 on Build GRCh38
rs766389272 Chr.3: 50349524-50349524 on Build GRCh38
rs181469895 Chr.3: 50349437-50349437 on Build GRCh38
rs1047956744 Chr.3: 50349585-50349585 on Build GRCh38
rs372216543 Chr.3: 50349656-50349656 on Build GRCh38
rs1003793255 Chr.3: 50349651-50349651 on Build GRCh38
rs760942051 Chr.3: 50349347-50349347 on Build GRCh38
rs371354092 Chr.3: 50349714-50349714 on Build GRCh38
rs869137991 Chr.3: 50349597-50349597 on Build GRCh38
rs1055732039 Chr.3: 50349566-50349566 on Build GRCh38
rs767181174 Chr.3: 50349634-50349634 on Build GRCh38
rs774253805 Chr.3: 50349412-50349412 on Build GRCh38
rs938208371 Chr.3: 50349529-50349529 on Build GRCh38
rs959606803 Chr.3: 50349366-50349366 on Build GRCh38
rs766337806 Chr.3: 50349403-50349403 on Build GRCh38
rs886780729 Chr.3: 50349585-50349585 on Build GRCh38
rs201475488 Chr.3: 50349379-50349379 on Build GRCh38
rs751450458 Chr.3: 50349536-50349536 on Build GRCh38
rs769094641 Chr.3: 50349715-50349715 on Build GRCh38
rs1027597260 Chr.3: 50349352-50349352 on Build GRCh38
rs587609327 Chr.3: 50349688-50349688 on Build GRCh38
rs750758549 Chr.3: 50349439-50349439 on Build GRCh38
rs746390246 Chr.3: 50349442-50349442 on Build GRCh38
rs886037965 Chr.3: 50349690-50349690 on Build GRCh38
rs933977683 Chr.3: 50349465-50349465 on Build GRCh38
rs145092948 Chr.3: 50349733-50349733 on Build GRCh38
rs1022386642 Chr.3: 50349388-50349388 on Build GRCh38
rs990046470 Chr.3: 50349377-50349377 on Build GRCh38
rs776610216 Chr.3: 50349376-50349376 on Build GRCh38
rs780702056 Chr.3: 50349512-50349512 on Build GRCh38
rs910455804 Chr.3: 50349683-50349683 on Build GRCh38
rs768804808 Chr.3: 50349479-50349479 on Build GRCh38
rs773365420 Chr.3: 50349513-50349513 on Build GRCh38
rs750415908 Chr.3: 50349392-50349392 on Build GRCh38
rs370615963 Chr.3: 50349390-50349390 on Build GRCh38
rs930938706 Chr.3: 50349579-50349579 on Build GRCh38
rs777201017 Chr.3: 50349440-50349440 on Build GRCh38
rs898935413 Chr.3: 50349799-50349799 on Build GRCh38
rs587705764 Chr.3: 50349592-50349592 on Build GRCh38
rs2233480 Chr.3: 50349752-50349752 on Build GRCh38
rs374673532 Chr.3: 50349404-50349404 on Build GRCh38
rs747710630 Chr.3: 50349438-50349438 on Build GRCh38
rs374673037 Chr.3: 50349716-50349716 on Build GRCh38
rs755667864 Chr.3: 50349643-50349643 on Build GRCh38
rs201388042 Chr.3: 50349459-50349459 on Build GRCh38
rs753339148 Chr.3: 50349674-50349674 on Build GRCh38
rs771505565 Chr.3: 50349449-50349449 on Build GRCh38
rs928016141 Chr.3: 50349480-50349480 on Build GRCh38
rs758940779 Chr.3: 50349691-50349691 on Build GRCh38
rs761929873 Chr.3: 50349389-50349389 on Build GRCh38
rs771086596 Chr.3: 50349746-50349746 on Build GRCh38
rs73082913 Chr.3: 50349826-50349826 on Build GRCh38
rs920757984 Chr.3: 50349578-50349578 on Build GRCh38
rs144456183 Chr.3: 50349458-50349458 on Build GRCh38
rs776934492 Chr.3: 50349491-50349491 on Build GRCh38
rs760439764 Chr.3: 50349809-50349809 on Build GRCh38
rs767360726 Chr.3: 50349783-50349783 on Build GRCh38
rs763052544 Chr.3: 50349516-50349516 on Build GRCh38
rs371149676 Chr.3: 50349505-50349505 on Build GRCh38
rs770138304 Chr.3: 50349725-50349725 on Build GRCh38
rs775772015 Chr.3: 50349336-50349336 on Build GRCh38
rs763703092 Chr.3: 50349815-50349815 on Build GRCh38
rs776081495 Chr.3: 50349535-50349535 on Build GRCh38
rs1033913832 Chr.3: 50349302-50349302 on Build GRCh38
rs747558141 Chr.3: 50349635-50349635 on Build GRCh38
rs747364938 Chr.3: 50349473-50349473 on Build GRCh38
rs780511845 Chr.3: 50349712-50349712 on Build GRCh38
rs764369246 Chr.3: 50349356-50349356 on Build GRCh38
rs772395363 Chr.3: 50349465-50349467 on Build GRCh38
rs763103438 Chr.3: 50349740-50349740 on Build GRCh38
rs375739985 Chr.3: 50349630-50349630 on Build GRCh38
rs869283779 Chr.3: 50349641-50349641 on Build GRCh38
rs377733182 Chr.3: 50349405-50349405 on Build GRCh38
rs2233482 Chr.3: 50349279-50349279 on Build GRCh38
rs2233481 Chr.3: 50349300-50349300 on Build GRCh38

View Full Product Details