Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 72241740-72242305 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PARP6
Entrez Gene ID
56965
Gene Name
poly(ADP-ribose) polymerase family member 6
Gene Aliases
ARTD17, PARP-6-B1, PARP-6-C, pART17
UniGene
Hs.270244
Gene Chromosome Location
Chr.15: 72241181-72272575 on Build GRCh38
Transcripts
NM_001323515.1 NM_001323516.1 NM_001323519.1 NM_001323521.1 NM_001323522.1 NM_001323523.1 NM_001323524.1 NM_001323525.1 NM_001323526.1 NM_001323528.1 NM_001323530.1 NM_001323531.1 NM_001323532.1 NM_020214.3 NR_136594.1 NR_136596.1 NR_136599.1 NR_136603.1 NR_136604.1 NR_136605.1 NR_136606.1 NR_136607.1 NR_136608.1 NR_136609.1 NR_136610.1 NR_136611.1 XM_011521805.1 XM_017022418.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1219320 c.1762G>A p.D588N Chr.15: 72241929-72241929 on Build GRCh38
964587 c.1719G>T p.K573N Chr.15: 72241972-72241972 on Build GRCh38
6241430 c.1649C>A p.S550Y Chr.15: 72242213-72242213 on Build GRCh38
3503694 c.1694C>T p.A565V Chr.15: 72242168-72242168 on Build GRCh38
4995598 c.1715C>G p.S572C Chr.15: 72241976-72241976 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372234832 Chr.15: 72241891-72241891 on Build GRCh38
rs755596368 Chr.15: 72241994-72241994 on Build GRCh38
rs539524202 Chr.15: 72242089-72242089 on Build GRCh38
rs1057196555 Chr.15: 72241820-72241820 on Build GRCh38
rs749117710 Chr.15: 72242267-72242267 on Build GRCh38
rs770503133 Chr.15: 72241869-72241869 on Build GRCh38
rs749857624 Chr.15: 72241852-72241852 on Build GRCh38
rs936525259 Chr.15: 72242006-72242006 on Build GRCh38
rs1049556734 Chr.15: 72242104-72242104 on Build GRCh38
rs756528183 Chr.15: 72241936-72241936 on Build GRCh38
rs11542893 Chr.15: 72241912-72241912 on Build GRCh38
rs768589193 Chr.15: 72242139-72242139 on Build GRCh38
rs771846731 Chr.15: 72241924-72241924 on Build GRCh38
rs761824034 Chr.15: 72242175-72242175 on Build GRCh38
rs756698453 Chr.15: 72242249-72242249 on Build GRCh38
rs546830227 Chr.15: 72241862-72241862 on Build GRCh38
rs769669071 Chr.15: 72241887-72241887 on Build GRCh38
rs1054071215 Chr.15: 72242009-72242009 on Build GRCh38
rs767341174 Chr.15: 72241825-72241826 on Build GRCh38
rs762032065 Chr.15: 72241921-72241921 on Build GRCh38
rs958980122 Chr.15: 72242274-72242274 on Build GRCh38
rs762819841 Chr.15: 72242137-72242137 on Build GRCh38
rs776726674 Chr.15: 72242037-72242037 on Build GRCh38
rs773475754 Chr.15: 72242264-72242264 on Build GRCh38
rs955121576 Chr.15: 72241767-72241767 on Build GRCh38
rs777275830 Chr.15: 72241999-72241999 on Build GRCh38
rs372747393 Chr.15: 72242217-72242217 on Build GRCh38
rs986972354 Chr.15: 72241819-72241820 on Build GRCh38
rs995789600 Chr.15: 72242081-72242081 on Build GRCh38
rs774452482 Chr.15: 72242153-72242153 on Build GRCh38
rs926805417 Chr.15: 72241894-72241894 on Build GRCh38
rs374658326 Chr.15: 72242111-72242111 on Build GRCh38
rs763645997 Chr.15: 72242142-72242142 on Build GRCh38
rs770577160 Chr.15: 72242015-72242015 on Build GRCh38
rs550689621 Chr.15: 72242063-72242063 on Build GRCh38
rs534229327 Chr.15: 72242251-72242251 on Build GRCh38
rs754389702 Chr.15: 72241944-72241944 on Build GRCh38
rs754264929 Chr.15: 72242216-72242216 on Build GRCh38
rs528407843 Chr.15: 72241777-72241777 on Build GRCh38
rs574243739 Chr.15: 72242270-72242270 on Build GRCh38
rs368978644 Chr.15: 72242186-72242186 on Build GRCh38
rs780729301 Chr.15: 72242026-72242026 on Build GRCh38
rs190535304 Chr.15: 72242194-72242194 on Build GRCh38
rs761041673 Chr.15: 72242197-72242197 on Build GRCh38
rs750754082 Chr.15: 72241935-72241935 on Build GRCh38
rs75194249 Chr.15: 72242256-72242256 on Build GRCh38
rs201968521 Chr.15: 72242215-72242215 on Build GRCh38
rs745507701 Chr.15: 72242252-72242252 on Build GRCh38
rs1007946821 Chr.15: 72242220-72242220 on Build GRCh38
rs776419660 Chr.15: 72241870-72241870 on Build GRCh38
rs548978018 Chr.15: 72242262-72242262 on Build GRCh38
rs185432952 Chr.15: 72242087-72242087 on Build GRCh38
rs780602299 Chr.15: 72241942-72241942 on Build GRCh38
rs908667699 Chr.15: 72241949-72241949 on Build GRCh38
rs751604054 Chr.15: 72241902-72241902 on Build GRCh38
rs769786852 Chr.15: 72242200-72242200 on Build GRCh38
rs777488275 Chr.15: 72241859-72241859 on Build GRCh38
rs746505272 Chr.15: 72242011-72242011 on Build GRCh38
rs775322615 Chr.15: 72242128-72242128 on Build GRCh38
rs1038357902 Chr.15: 72242040-72242040 on Build GRCh38
rs779757166 Chr.15: 72242266-72242266 on Build GRCh38
rs143896290 Chr.15: 72242030-72242030 on Build GRCh38
rs753260181 Chr.15: 72242225-72242226 on Build GRCh38
rs890185939 Chr.15: 72242092-72242092 on Build GRCh38
rs751030534 Chr.15: 72242242-72242242 on Build GRCh38
rs771675237 Chr.15: 72241855-72241855 on Build GRCh38
rs989847794 Chr.15: 72241856-72241856 on Build GRCh38
rs561936594 Chr.15: 72241892-72241892 on Build GRCh38
rs765693706 Chr.15: 72242237-72242237 on Build GRCh38
rs767667723 Chr.15: 72241930-72241930 on Build GRCh38

View Full Product Details