Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 19534189-19534772 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SH3KBP1
Entrez Gene ID
30011
Gene Name
SH3 domain containing kinase binding protein 1
Gene Aliases
CD2BP3, CIN85, GIG10, HSB-1, HSB1, MIG18
UniGene
Hs.726365
Gene Chromosome Location
Chr.X: 19533965-19887626 on Build GRCh38
Transcripts
NM_001024666.2 NM_001184960.1 NM_031892.2 XM_011545500.2 XM_017029462.1 XM_017029464.1 XM_017029466.1 XM_017029467.1 XM_017029468.1 XM_017029470.1

SNP Detail

SNP ID Chromosome Location
rs1008032909 Chr.X: 19534532-19534532 on Build GRCh38
rs1015339792 Chr.X: 19534562-19534562 on Build GRCh38
rs965494106 Chr.X: 19534341-19534341 on Build GRCh38
rs1049523537 Chr.X: 19534517-19534517 on Build GRCh38
rs899073887 Chr.X: 19534693-19534693 on Build GRCh38
rs777157272 Chr.X: 19534696-19534696 on Build GRCh38
rs917983130 Chr.X: 19534288-19534288 on Build GRCh38
rs775821458 Chr.X: 19534580-19534580 on Build GRCh38
rs189460900 Chr.X: 19534208-19534208 on Build GRCh38
rs959626466 Chr.X: 19534260-19534260 on Build GRCh38
rs368683863 Chr.X: 19534507-19534507 on Build GRCh38
rs1026853954 Chr.X: 19534606-19534606 on Build GRCh38
rs1042558195 Chr.X: 19534323-19534323 on Build GRCh38
rs1043448792 Chr.X: 19534390-19534390 on Build GRCh38
rs761044532 Chr.X: 19534420-19534420 on Build GRCh38
rs771782128 Chr.X: 19534404-19534404 on Build GRCh38
rs1054034996 Chr.X: 19534403-19534403 on Build GRCh38
rs985673794 Chr.X: 19534214-19534214 on Build GRCh38
rs749211351 Chr.X: 19534217-19534217 on Build GRCh38
rs947412670 Chr.X: 19534359-19534359 on Build GRCh38
rs1029061930 Chr.X: 19534728-19534728 on Build GRCh38
rs772019732 Chr.X: 19534575-19534575 on Build GRCh38
rs180969967 Chr.X: 19534405-19534405 on Build GRCh38
rs868782725 Chr.X: 19534213-19534213 on Build GRCh38
rs995415588 Chr.X: 19534605-19534605 on Build GRCh38
rs879144445 Chr.X: 19534364-19534364 on Build GRCh38
rs927436174 Chr.X: 19534394-19534394 on Build GRCh38
rs919891259 Chr.X: 19534252-19534252 on Build GRCh38
rs770448355 Chr.X: 19534219-19534219 on Build GRCh38
rs990946491 Chr.X: 19534261-19534261 on Build GRCh38
rs971182111 Chr.X: 19534692-19534692 on Build GRCh38
rs959432269 Chr.X: 19534620-19534620 on Build GRCh38
rs936820870 Chr.X: 19534707-19534707 on Build GRCh38
rs890840099 Chr.X: 19534531-19534531 on Build GRCh38
rs924140627 Chr.X: 19534702-19534702 on Build GRCh38
rs778448080 Chr.X: 19534351-19534351 on Build GRCh38
rs768925139 Chr.X: 19534459-19534459 on Build GRCh38
rs867856503 Chr.X: 19534739-19534739 on Build GRCh38
rs1025077628 Chr.X: 19534653-19534653 on Build GRCh38
rs949594145 Chr.X: 19534299-19534299 on Build GRCh38
rs745509512 Chr.X: 19534396-19534396 on Build GRCh38
rs974045201 Chr.X: 19534237-19534237 on Build GRCh38
rs936874888 Chr.X: 19534352-19534352 on Build GRCh38
rs888894357 Chr.X: 19534745-19534745 on Build GRCh38
rs991201088 Chr.X: 19534652-19534652 on Build GRCh38

View Full Product Details