Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 26440068-26440622 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTN3A2
Entrez Gene ID
11118
Gene Name
butyrophilin subfamily 3 member A2
Gene Aliases
BT3.2, BTF4, BTN3.2, CD277
UniGene
Hs.376046
Gene Chromosome Location
Chr.6: 26365159-26451697 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

Gene Symbol
BTN3A3
Entrez Gene ID
10384
Gene Name
butyrophilin subfamily 3 member A3
Gene Aliases
BTF3, BTN3.3
UniGene
Hs.167741
Gene Chromosome Location
Chr.6: 26440472-26453415 on Build GRCh38
Transcripts
XM_011514268.2 NM_001242803.1 NM_006994.4 NM_197974.2

SNP Detail

SNP ID Chromosome Location
rs899748264 Chr.6: 26440304-26440304 on Build GRCh38
rs958256905 Chr.6: 26440426-26440426 on Build GRCh38
rs1048262530 Chr.6: 26440575-26440575 on Build GRCh38
rs779236465 Chr.6: 26440162-26440162 on Build GRCh38
rs576078689 Chr.6: 26440309-26440309 on Build GRCh38
rs1024685696 Chr.6: 26440396-26440396 on Build GRCh38
rs757830776 Chr.6: 26440119-26440119 on Build GRCh38
rs184460009 Chr.6: 26440094-26440094 on Build GRCh38
rs181211814 Chr.6: 26440298-26440298 on Build GRCh38
rs117832370 Chr.6: 26440592-26440592 on Build GRCh38
rs188436444 Chr.6: 26440219-26440219 on Build GRCh38
rs995378024 Chr.6: 26440570-26440570 on Build GRCh38
rs72500816 Chr.6: 26440418-26440418 on Build GRCh38
rs558993965 Chr.6: 26440556-26440556 on Build GRCh38
rs9379874 Chr.6: 26440201-26440201 on Build GRCh38
rs747868265 Chr.6: 26440296-26440296 on Build GRCh38
rs932277745 Chr.6: 26440430-26440430 on Build GRCh38
rs759671641 Chr.6: 26440548-26440548 on Build GRCh38
rs942186622 Chr.6: 26440531-26440531 on Build GRCh38
rs572696264 Chr.6: 26440362-26440362 on Build GRCh38
rs563342415 Chr.6: 26440520-26440520 on Build GRCh38
rs138389459 Chr.6: 26440363-26440363 on Build GRCh38
rs980936927 Chr.6: 26440170-26440170 on Build GRCh38
rs991795640 Chr.6: 26440385-26440385 on Build GRCh38
rs1039495237 Chr.6: 26440562-26440562 on Build GRCh38
rs1002554952 Chr.6: 26440326-26440326 on Build GRCh38
rs539110449 Chr.6: 26440241-26440241 on Build GRCh38
rs982594951 Chr.6: 26440110-26440110 on Build GRCh38
rs1052218685 Chr.6: 26440535-26440535 on Build GRCh38
rs991348982 Chr.6: 26440101-26440101 on Build GRCh38
rs17539219 Chr.6: 26440314-26440314 on Build GRCh38
rs1004292043 Chr.6: 26440382-26440382 on Build GRCh38
rs897589096 Chr.6: 26440557-26440557 on Build GRCh38
rs769570141 Chr.6: 26440297-26440297 on Build GRCh38
rs556115914 Chr.6: 26440264-26440264 on Build GRCh38
rs375242400 Chr.6: 26440442-26440442 on Build GRCh38
rs367923360 Chr.6: 26440460-26440460 on Build GRCh38
rs12214444 Chr.6: 26440424-26440424 on Build GRCh38
rs935341799 Chr.6: 26440232-26440232 on Build GRCh38
rs989769475 Chr.6: 26440427-26440427 on Build GRCh38
rs527447679 Chr.6: 26440096-26440096 on Build GRCh38
rs945358492 Chr.6: 26440501-26440501 on Build GRCh38
rs114195506 Chr.6: 26440564-26440564 on Build GRCh38
rs781006637 Chr.6: 26440255-26440255 on Build GRCh38
rs887926775 Chr.6: 26440282-26440282 on Build GRCh38
rs1051823519 Chr.6: 26440274-26440274 on Build GRCh38
rs764509180 Chr.6: 26440095-26440095 on Build GRCh38
rs773452661 Chr.6: 26440137-26440137 on Build GRCh38
rs887981298 Chr.6: 26440530-26440530 on Build GRCh38
rs970899523 Chr.6: 26440405-26440405 on Build GRCh38
rs946825486 Chr.6: 26440132-26440132 on Build GRCh38
rs147073759 Chr.6: 26440279-26440279 on Build GRCh38
rs754169718 Chr.6: 26440112-26440112 on Build GRCh38

View Full Product Details