Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 17793274-17793801 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SNX13
Entrez Gene ID
23161
Gene Name
sorting nexin 13
Gene Aliases
RGS-PX1
UniGene
Hs.487648
Gene Chromosome Location
Chr.7: 17790761-17940518 on Build GRCh38
Transcripts
NM_015132.4 XM_005249672.1 XM_011515229.1 XM_011515230.2 XM_011515231.2 XM_011515233.2 XM_017011893.1 XM_017011894.1 XM_017011895.1 XM_017011896.1 XM_017011897.1

SNP Detail

SNP ID Chromosome Location
rs368412139 Chr.7: 17793314-17793314 on Build GRCh38
rs956951712 Chr.7: 17793670-17793670 on Build GRCh38
rs956692625 Chr.7: 17793431-17793431 on Build GRCh38
rs1004037400 Chr.7: 17793640-17793640 on Build GRCh38
rs192346168 Chr.7: 17793599-17793599 on Build GRCh38
rs781182713 Chr.7: 17793371-17793371 on Build GRCh38
rs995709118 Chr.7: 17793597-17793597 on Build GRCh38
rs988428159 Chr.7: 17793432-17793432 on Build GRCh38
rs922362473 Chr.7: 17793768-17793768 on Build GRCh38
rs986347477 Chr.7: 17793735-17793735 on Build GRCh38
rs1031326027 Chr.7: 17793662-17793662 on Build GRCh38
rs572893546 Chr.7: 17793755-17793755 on Build GRCh38
rs928676477 Chr.7: 17793514-17793514 on Build GRCh38
rs115673149 Chr.7: 17793462-17793462 on Build GRCh38
rs570806806 Chr.7: 17793345-17793345 on Build GRCh38
rs1017734466 Chr.7: 17793367-17793367 on Build GRCh38
rs539398482 Chr.7: 17793505-17793505 on Build GRCh38
rs565949958 Chr.7: 17793434-17793434 on Build GRCh38
rs1037028095 Chr.7: 17793732-17793732 on Build GRCh38
rs780186683 Chr.7: 17793663-17793663 on Build GRCh38
rs980314409 Chr.7: 17793726-17793726 on Build GRCh38
rs922299873 Chr.7: 17793561-17793561 on Build GRCh38
rs1049079557 Chr.7: 17793606-17793606 on Build GRCh38
rs914433448 Chr.7: 17793696-17793696 on Build GRCh38
rs1035438244 Chr.7: 17793405-17793405 on Build GRCh38
rs955920905 Chr.7: 17793429-17793429 on Build GRCh38
rs1041121531 Chr.7: 17793302-17793302 on Build GRCh38
rs115060981 Chr.7: 17793438-17793438 on Build GRCh38
rs910714339 Chr.7: 17793745-17793745 on Build GRCh38
rs756315335 Chr.7: 17793554-17793554 on Build GRCh38
rs888312564 Chr.7: 17793357-17793357 on Build GRCh38
rs371702024 Chr.7: 17793728-17793728 on Build GRCh38
rs184930989 Chr.7: 17793399-17793399 on Build GRCh38
rs1036975782 Chr.7: 17793581-17793581 on Build GRCh38
rs889966259 Chr.7: 17793612-17793612 on Build GRCh38
rs1007079685 Chr.7: 17793781-17793781 on Build GRCh38
rs946893946 Chr.7: 17793446-17793446 on Build GRCh38
rs1027141140 Chr.7: 17793340-17793340 on Build GRCh38
rs557213165 Chr.7: 17793385-17793385 on Build GRCh38
rs910692201 Chr.7: 17793477-17793477 on Build GRCh38
rs528466683 Chr.7: 17793396-17793396 on Build GRCh38
rs955973064 Chr.7: 17793671-17793671 on Build GRCh38
rs189541040 Chr.7: 17793566-17793566 on Build GRCh38
rs900804148 Chr.7: 17793306-17793306 on Build GRCh38
rs890007451 Chr.7: 17793765-17793765 on Build GRCh38
rs771721391 Chr.7: 17793710-17793710 on Build GRCh38
rs1045312229 Chr.7: 17793635-17793635 on Build GRCh38
rs1046995248 Chr.7: 17793573-17793573 on Build GRCh38
rs535517062 Chr.7: 17793756-17793756 on Build GRCh38
rs1035104694 Chr.7: 17793655-17793655 on Build GRCh38
rs914401929 Chr.7: 17793459-17793459 on Build GRCh38
rs367803434 Chr.7: 17793474-17793474 on Build GRCh38
rs536451745 Chr.7: 17793435-17793435 on Build GRCh38
rs1005328907 Chr.7: 17793358-17793358 on Build GRCh38
rs1005399373 Chr.7: 17793634-17793634 on Build GRCh38
rs75078641 Chr.7: 17793720-17793720 on Build GRCh38
rs778806263 Chr.7: 17793690-17793690 on Build GRCh38
rs904989743 Chr.7: 17793639-17793639 on Build GRCh38
rs888345203 Chr.7: 17793589-17793589 on Build GRCh38
rs968303727 Chr.7: 17793727-17793727 on Build GRCh38

View Full Product Details