Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 37906084-37906647 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SFRP4
Entrez Gene ID
6424
Gene Name
secreted frizzled related protein 4
Gene Aliases
FRP-4, FRPHE, sFRP-4
UniGene
Hs.658169
Gene Chromosome Location
Chr.7: 37905932-37916923 on Build GRCh38
Transcripts
NM_003014.3

SNP Detail

SNP ID Chromosome Location
rs202233158 Chr.7: 37906589-37906589 on Build GRCh38
rs964115650 Chr.7: 37906282-37906282 on Build GRCh38
rs919986521 Chr.7: 37906579-37906579 on Build GRCh38
rs545170794 Chr.7: 37906431-37906431 on Build GRCh38
rs911195217 Chr.7: 37906264-37906264 on Build GRCh38
rs891497828 Chr.7: 37906197-37906197 on Build GRCh38
rs984237280 Chr.7: 37906364-37906364 on Build GRCh38
rs930438535 Chr.7: 37906333-37906333 on Build GRCh38
rs537443461 Chr.7: 37906151-37906151 on Build GRCh38
rs557351351 Chr.7: 37906190-37906190 on Build GRCh38
rs1011950437 Chr.7: 37906435-37906435 on Build GRCh38
rs906794864 Chr.7: 37906268-37906268 on Build GRCh38
rs999671621 Chr.7: 37906465-37906465 on Build GRCh38
rs925818851 Chr.7: 37906514-37906514 on Build GRCh38
rs964355201 Chr.7: 37906558-37906558 on Build GRCh38
rs769501048 Chr.7: 37906192-37906192 on Build GRCh38
rs905505984 Chr.7: 37906595-37906595 on Build GRCh38
rs955627340 Chr.7: 37906230-37906230 on Build GRCh38
rs939255294 Chr.7: 37906402-37906402 on Build GRCh38
rs577824584 Chr.7: 37906569-37906569 on Build GRCh38
rs1008893699 Chr.7: 37906525-37906525 on Build GRCh38
rs553557973 Chr.7: 37906395-37906395 on Build GRCh38
rs935831107 Chr.7: 37906553-37906553 on Build GRCh38
rs747849389 Chr.7: 37906126-37906126 on Build GRCh38
rs573373764 Chr.7: 37906421-37906421 on Build GRCh38
rs760465419 Chr.7: 37906557-37906557 on Build GRCh38
rs955589481 Chr.7: 37906513-37906513 on Build GRCh38
rs907767174 Chr.7: 37906387-37906387 on Build GRCh38
rs1043462287 Chr.7: 37906437-37906437 on Build GRCh38
rs576625640 Chr.7: 37906125-37906125 on Build GRCh38
rs567787422 Chr.7: 37906304-37906304 on Build GRCh38
rs974554304 Chr.7: 37906292-37906292 on Build GRCh38
rs985763871 Chr.7: 37906509-37906509 on Build GRCh38
rs1013875292 Chr.7: 37906239-37906239 on Build GRCh38
rs1049595398 Chr.7: 37906110-37906110 on Build GRCh38
rs920017561 Chr.7: 37906305-37906305 on Build GRCh38
rs772734319 Chr.7: 37906200-37906200 on Build GRCh38
rs904331477 Chr.7: 37906111-37906111 on Build GRCh38
rs762609001 Chr.7: 37906273-37906273 on Build GRCh38
rs575493144 Chr.7: 37906583-37906583 on Build GRCh38
rs370610522 Chr.7: 37906359-37906359 on Build GRCh38
rs17171199 Chr.7: 37906438-37906438 on Build GRCh38
rs1031188792 Chr.7: 37906502-37906502 on Build GRCh38
rs987099753 Chr.7: 37906249-37906249 on Build GRCh38

View Full Product Details