Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144453327-144453870 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPK1
Entrez Gene ID
27010
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
HTPK1, PP20, THMD5
UniGene
Hs.660232
Gene Chromosome Location
Chr.7: 144451941-144836424 on Build GRCh38
Transcripts
NM_001042482.1 NM_022445.3 XM_005249970.1 XM_011516031.1 XM_011516032.2 XM_011516033.2 XM_011516034.2 XM_011516035.2 XM_011516037.2 XM_011516039.2 XM_011516041.2 XM_011516043.1 XM_011516044.1 XM_011516047.2 XM_011516048.1 XM_011516050.1 XM_017011970.1 XM_017011971.1 XM_017011973.1 XM_017011974.1 XM_017011975.1 XM_017011976.1 XM_017011977.1 XM_017011978.1 XM_017011979.1 XM_017011980.1 XM_017011981.1 XM_017011982.1 XM_017011983.1 XM_017011984.1 XM_017011985.1 XM_017011986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5791517 c.630T>A p.A210A Chr.7: 144453647-144453647 on Build GRCh38
3832204 c.670T>G p.S224A Chr.7: 144453607-144453607 on Build GRCh38
6334412 c.638C>A p.T213K Chr.7: 144453639-144453639 on Build GRCh38
3259235 c.642G>T p.L214F Chr.7: 144453635-144453635 on Build GRCh38
3259234 c.644T>C p.V215A Chr.7: 144453633-144453633 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747262651 Chr.7: 144453657-144453657 on Build GRCh38
rs993615445 Chr.7: 144453689-144453689 on Build GRCh38
rs112713879 Chr.7: 144453723-144453723 on Build GRCh38
rs779048294 Chr.7: 144453635-144453635 on Build GRCh38
rs373578497 Chr.7: 144453705-144453705 on Build GRCh38
rs879046139 Chr.7: 144453836-144453836 on Build GRCh38
rs183936021 Chr.7: 144453783-144453783 on Build GRCh38
rs764746135 Chr.7: 144453609-144453609 on Build GRCh38
rs770269972 Chr.7: 144453678-144453678 on Build GRCh38
rs750322523 Chr.7: 144453615-144453615 on Build GRCh38
rs763805803 Chr.7: 144453697-144453697 on Build GRCh38
rs899305639 Chr.7: 144453405-144453405 on Build GRCh38
rs748289097 Chr.7: 144453642-144453642 on Build GRCh38
rs778011281 Chr.7: 144453656-144453656 on Build GRCh38
rs766932422 Chr.7: 144453494-144453494 on Build GRCh38
rs548115884 Chr.7: 144453653-144453653 on Build GRCh38
rs1026508154 Chr.7: 144453432-144453432 on Build GRCh38
rs772262824 Chr.7: 144453643-144453643 on Build GRCh38
rs568804217 Chr.7: 144453610-144453610 on Build GRCh38
rs758136518 Chr.7: 144453522-144453522 on Build GRCh38
rs757245567 Chr.7: 144453537-144453537 on Build GRCh38
rs748307256 Chr.7: 144453577-144453577 on Build GRCh38
rs113056357 Chr.7: 144453608-144453608 on Build GRCh38
rs773756971 Chr.7: 144453697-144453698 on Build GRCh38
rs925712502 Chr.7: 144453604-144453604 on Build GRCh38
rs753388291 Chr.7: 144453449-144453449 on Build GRCh38
rs763896351 Chr.7: 144453611-144453611 on Build GRCh38
rs867075548 Chr.7: 144453472-144453472 on Build GRCh38
rs1044707304 Chr.7: 144453655-144453655 on Build GRCh38
rs748672196 Chr.7: 144453681-144453681 on Build GRCh38
rs532067918 Chr.7: 144453516-144453516 on Build GRCh38
rs771280369 Chr.7: 144453658-144453658 on Build GRCh38
rs746865416 Chr.7: 144453529-144453529 on Build GRCh38
rs368458768 Chr.7: 144453613-144453613 on Build GRCh38
rs773555092 Chr.7: 144453587-144453587 on Build GRCh38
rs919953383 Chr.7: 144453738-144453738 on Build GRCh38
rs375718551 Chr.7: 144453513-144453513 on Build GRCh38
rs188268966 Chr.7: 144453796-144453796 on Build GRCh38
rs781237117 Chr.7: 144453545-144453545 on Build GRCh38
rs754558149 Chr.7: 144453509-144453509 on Build GRCh38
rs539336947 Chr.7: 144453676-144453676 on Build GRCh38
rs761078676 Chr.7: 144453592-144453592 on Build GRCh38
rs543389713 Chr.7: 144453436-144453436 on Build GRCh38
rs749295013 Chr.7: 144453576-144453576 on Build GRCh38
rs540800145 Chr.7: 144453462-144453462 on Build GRCh38
rs775127014 Chr.7: 144453681-144453681 on Build GRCh38
rs113719353 Chr.7: 144453456-144453456 on Build GRCh38
rs145466113 Chr.7: 144453730-144453730 on Build GRCh38
rs938025274 Chr.7: 144453666-144453666 on Build GRCh38
rs751374816 Chr.7: 144453612-144453612 on Build GRCh38
rs978505783 Chr.7: 144453564-144453564 on Build GRCh38
rs550639617 Chr.7: 144453600-144453600 on Build GRCh38
rs772523726 Chr.7: 144453698-144453698 on Build GRCh38
rs113536847 Chr.7: 144453614-144453614 on Build GRCh38
rs776874412 Chr.7: 144453664-144453664 on Build GRCh38
rs777155582 Chr.7: 144453602-144453602 on Build GRCh38
rs779514047 Chr.7: 144453682-144453682 on Build GRCh38
rs1016771607 Chr.7: 144453353-144453353 on Build GRCh38
rs6953807 Chr.7: 144453745-144453745 on Build GRCh38
rs1032748361 Chr.7: 144453826-144453826 on Build GRCh38
rs564993869 Chr.7: 144453515-144453515 on Build GRCh38
rs115576202 Chr.7: 144453441-144453441 on Build GRCh38
rs760020447 Chr.7: 144453676-144453676 on Build GRCh38
rs772352353 Chr.7: 144453581-144453581 on Build GRCh38
rs575636592 Chr.7: 144453473-144453473 on Build GRCh38
rs371271054 Chr.7: 144453621-144453621 on Build GRCh38
rs753267970 Chr.7: 144453507-144453507 on Build GRCh38
rs778438891 Chr.7: 144453652-144453652 on Build GRCh38
rs777643659 Chr.7: 144453527-144453527 on Build GRCh38
rs113934415 Chr.7: 144453691-144453691 on Build GRCh38
rs146756933 Chr.7: 144453430-144453430 on Build GRCh38

View Full Product Details