Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 23255216-23255661 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHMP7
Entrez Gene ID
91782
Gene Name
charged multivesicular body protein 7
Gene Aliases
-
UniGene
Hs.5019
Gene Chromosome Location
Chr.8: 23243637-23262000 on Build GRCh38
Transcripts
NM_001317899.1 NM_152272.4 XM_006716415.2 XM_017013961.1 XM_017013962.1 XM_017013963.1 XM_017013964.1 XM_017013965.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5685017 c.640G>T p.E214* Chr.8: 23255415-23255415 on Build GRCh38
4821647 c.580G>A p.E194K Chr.8: 23255355-23255355 on Build GRCh38
1201076 c.634G>T p.V212F Chr.8: 23255409-23255409 on Build GRCh38
7414739 c.612G>T p.L204L Chr.8: 23255387-23255387 on Build GRCh38
6483949 c.526C>A p.P176T Chr.8: 23255301-23255301 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372133284 Chr.8: 23255474-23255474 on Build GRCh38
rs146285644 Chr.8: 23255264-23255264 on Build GRCh38
rs887132508 Chr.8: 23255547-23255547 on Build GRCh38
rs749757566 Chr.8: 23255291-23255291 on Build GRCh38
rs765613038 Chr.8: 23255410-23255410 on Build GRCh38
rs1044309910 Chr.8: 23255629-23255629 on Build GRCh38
rs781545438 Chr.8: 23255445-23255445 on Build GRCh38
rs987320101 Chr.8: 23255285-23255285 on Build GRCh38
rs750638100 Chr.8: 23255411-23255411 on Build GRCh38
rs528528209 Chr.8: 23255641-23255641 on Build GRCh38
rs201176959 Chr.8: 23255304-23255304 on Build GRCh38
rs761313590 Chr.8: 23255499-23255499 on Build GRCh38
rs370704599 Chr.8: 23255293-23255293 on Build GRCh38
rs995865545 Chr.8: 23255535-23255535 on Build GRCh38
rs1005404565 Chr.8: 23255433-23255433 on Build GRCh38
rs144111266 Chr.8: 23255423-23255423 on Build GRCh38
rs752655432 Chr.8: 23255248-23255248 on Build GRCh38
rs956548726 Chr.8: 23255608-23255608 on Build GRCh38
rs898373960 Chr.8: 23255505-23255505 on Build GRCh38
rs767808320 Chr.8: 23255638-23255638 on Build GRCh38
rs968990063 Chr.8: 23255586-23255586 on Build GRCh38
rs772969108 Chr.8: 23255476-23255477 on Build GRCh38
rs759170647 Chr.8: 23255333-23255333 on Build GRCh38
rs1008248463 Chr.8: 23255549-23255549 on Build GRCh38
rs757496234 Chr.8: 23255268-23255268 on Build GRCh38
rs767408139 Chr.8: 23255242-23255242 on Build GRCh38
rs769042906 Chr.8: 23255294-23255294 on Build GRCh38
rs760199715 Chr.8: 23255414-23255414 on Build GRCh38
rs544155332 Chr.8: 23255550-23255550 on Build GRCh38
rs896253394 Chr.8: 23255495-23255495 on Build GRCh38
rs554721092 Chr.8: 23255348-23255348 on Build GRCh38
rs77467372 Chr.8: 23255555-23255555 on Build GRCh38
rs1016325995 Chr.8: 23255436-23255436 on Build GRCh38
rs139408800 Chr.8: 23255340-23255340 on Build GRCh38
rs748566933 Chr.8: 23255458-23255458 on Build GRCh38
rs763994535 Chr.8: 23255361-23255361 on Build GRCh38
rs779138390 Chr.8: 23255286-23255286 on Build GRCh38
rs184706231 Chr.8: 23255441-23255441 on Build GRCh38
rs924392697 Chr.8: 23255273-23255273 on Build GRCh38
rs542779994 Chr.8: 23255347-23255347 on Build GRCh38
rs774904114 Chr.8: 23255403-23255403 on Build GRCh38
rs770633533 Chr.8: 23255313-23255313 on Build GRCh38
rs1021787500 Chr.8: 23255577-23255577 on Build GRCh38
rs888374410 Chr.8: 23255634-23255634 on Build GRCh38
rs759015296 Chr.8: 23255327-23255327 on Build GRCh38
rs540436626 Chr.8: 23255485-23255485 on Build GRCh38
rs975514227 Chr.8: 23255556-23255556 on Build GRCh38
rs57455008 Chr.8: 23255601-23255601 on Build GRCh38
rs756444175 Chr.8: 23255461-23255461 on Build GRCh38
rs367547668 Chr.8: 23255350-23255350 on Build GRCh38
rs75619283 Chr.8: 23255540-23255540 on Build GRCh38
rs199739181 Chr.8: 23255303-23255303 on Build GRCh38
rs750536723 Chr.8: 23255288-23255288 on Build GRCh38
rs777211878 Chr.8: 23255470-23255473 on Build GRCh38
rs78884645 Chr.8: 23255617-23255617 on Build GRCh38
rs375158904 Chr.8: 23255477-23255477 on Build GRCh38
rs189147669 Chr.8: 23255488-23255488 on Build GRCh38
rs776502129 Chr.8: 23255368-23255368 on Build GRCh38
rs200178384 Chr.8: 23255312-23255312 on Build GRCh38
rs1030359663 Chr.8: 23255453-23255453 on Build GRCh38
rs771504529 Chr.8: 23255478-23255478 on Build GRCh38
rs753780033 Chr.8: 23255498-23255498 on Build GRCh38
rs530827320 Chr.8: 23255592-23255592 on Build GRCh38
rs761697485 Chr.8: 23255369-23255369 on Build GRCh38
rs774036693 Chr.8: 23255318-23255318 on Build GRCh38
rs748957186 Chr.8: 23255311-23255311 on Build GRCh38
rs867518147 Chr.8: 23255263-23255263 on Build GRCh38
rs377356281 Chr.8: 23255269-23255269 on Build GRCh38
rs755424068 Chr.8: 23255444-23255444 on Build GRCh38

View Full Product Details