Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 22737441-22737927 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GAS2
Entrez Gene ID
2620
Gene Name
growth arrest specific 2
Gene Aliases
-
UniGene
Hs.711297
Gene Chromosome Location
Chr.11: 22626113-22885930 on Build GRCh38
Transcripts
NM_001143830.1 NM_005256.3 NM_177553.2 XM_011519971.2 XM_011519972.2 XM_011519975.1 XM_011519978.2 XM_017017528.1 XM_017017529.1 XM_017017530.1 XM_017017531.1 XM_017017532.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2112698 c.412C>A p.L138I Chr.11: 22737707-22737707 on Build GRCh38
4848986 c.430G>C p.E144Q Chr.11: 22737725-22737725 on Build GRCh38
1135163 c.449T>C p.L150P Chr.11: 22737744-22737744 on Build GRCh38
2112700 c.439C>G p.L147V Chr.11: 22737734-22737734 on Build GRCh38
1297929 c.470C>G p.A157G Chr.11: 22737765-22737765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985168557 Chr.11: 22737804-22737804 on Build GRCh38
rs773021267 Chr.11: 22737687-22737687 on Build GRCh38
rs752997248 Chr.11: 22737755-22737755 on Build GRCh38
rs199567310 Chr.11: 22737661-22737661 on Build GRCh38
rs1045879713 Chr.11: 22737828-22737828 on Build GRCh38
rs201829982 Chr.11: 22737806-22737806 on Build GRCh38
rs754122927 Chr.11: 22737766-22737766 on Build GRCh38
rs981927699 Chr.11: 22737539-22737539 on Build GRCh38
rs749016095 Chr.11: 22737812-22737812 on Build GRCh38
rs762324906 Chr.11: 22737696-22737696 on Build GRCh38
rs781291046 Chr.11: 22737801-22737801 on Build GRCh38
rs757116824 Chr.11: 22737520-22737520 on Build GRCh38
rs1013303809 Chr.11: 22737850-22737850 on Build GRCh38
rs536896531 Chr.11: 22737485-22737485 on Build GRCh38
rs374996114 Chr.11: 22737790-22737790 on Build GRCh38
rs1001775998 Chr.11: 22737549-22737549 on Build GRCh38
rs370785219 Chr.11: 22737685-22737685 on Build GRCh38
rs147075339 Chr.11: 22737551-22737551 on Build GRCh38
rs970561063 Chr.11: 22737526-22737526 on Build GRCh38
rs757254542 Chr.11: 22737782-22737782 on Build GRCh38
rs894494217 Chr.11: 22737835-22737835 on Build GRCh38
rs760149502 Chr.11: 22737716-22737716 on Build GRCh38
rs770331465 Chr.11: 22737815-22737815 on Build GRCh38
rs893388217 Chr.11: 22737590-22737590 on Build GRCh38
rs765949417 Chr.11: 22737711-22737711 on Build GRCh38
rs754794140 Chr.11: 22737881-22737881 on Build GRCh38
rs779815834 Chr.11: 22737658-22737658 on Build GRCh38
rs764251796 Chr.11: 22737764-22737764 on Build GRCh38
rs147985883 Chr.11: 22737718-22737718 on Build GRCh38
rs7934119 Chr.11: 22737622-22737622 on Build GRCh38
rs201086358 Chr.11: 22737682-22737682 on Build GRCh38
rs907918927 Chr.11: 22737465-22737465 on Build GRCh38
rs774695678 Chr.11: 22737715-22737715 on Build GRCh38
rs907395518 Chr.11: 22737522-22737522 on Build GRCh38
rs180803897 Chr.11: 22737817-22737817 on Build GRCh38
rs1011623311 Chr.11: 22737618-22737618 on Build GRCh38
rs919804537 Chr.11: 22737748-22737748 on Build GRCh38
rs368409840 Chr.11: 22737805-22737805 on Build GRCh38
rs777510325 Chr.11: 22737464-22737464 on Build GRCh38
rs749116868 Chr.11: 22737484-22737484 on Build GRCh38
rs770029870 Chr.11: 22737885-22737885 on Build GRCh38
rs576375630 Chr.11: 22737601-22737601 on Build GRCh38
rs763063566 Chr.11: 22737905-22737905 on Build GRCh38
rs779101145 Chr.11: 22737786-22737786 on Build GRCh38
rs756488991 Chr.11: 22737759-22737759 on Build GRCh38
rs768315965 Chr.11: 22737672-22737672 on Build GRCh38
rs113652439 Chr.11: 22737649-22737649 on Build GRCh38
rs767867012 Chr.11: 22737736-22737736 on Build GRCh38
rs747959153 Chr.11: 22737684-22737684 on Build GRCh38

View Full Product Details