Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 26572701-26573273 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYYR1
Entrez Gene ID
116159
Gene Name
cysteine and tyrosine rich 1
Gene Aliases
C21orf95
UniGene
Hs.37445
Gene Chromosome Location
Chr.21: 26466209-26573404 on Build GRCh38
Transcripts
NM_001320768.1 NM_052954.3 NR_135472.1 XM_011529450.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5437464 c.393C>T p.Y131Y Chr.21: 26572872-26572872 on Build GRCh38
6094408 c.344C>A p.P115H Chr.21: 26572921-26572921 on Build GRCh38
5437463 c.69C>T p.Y23Y Chr.21: 26572872-26572872 on Build GRCh38
6161090 c.330C>T p.D110D Chr.21: 26572935-26572935 on Build GRCh38
3933935 c.323G>T p.W108L Chr.21: 26572942-26572942 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992870234 Chr.21: 26572987-26572987 on Build GRCh38
rs759162634 Chr.21: 26572911-26572911 on Build GRCh38
rs757702759 Chr.21: 26572930-26572930 on Build GRCh38
rs917336679 Chr.21: 26573221-26573221 on Build GRCh38
rs769914396 Chr.21: 26572881-26572881 on Build GRCh38
rs768829358 Chr.21: 26572940-26572940 on Build GRCh38
rs766477801 Chr.21: 26573019-26573019 on Build GRCh38
rs576436644 Chr.21: 26572818-26572818 on Build GRCh38
rs765500303 Chr.21: 26573021-26573022 on Build GRCh38
rs936222096 Chr.21: 26573074-26573074 on Build GRCh38
rs980224906 Chr.21: 26573251-26573251 on Build GRCh38
rs747913196 Chr.21: 26573028-26573028 on Build GRCh38
rs146452127 Chr.21: 26572937-26572937 on Build GRCh38
rs952763494 Chr.21: 26572897-26572897 on Build GRCh38
rs751249082 Chr.21: 26572910-26572910 on Build GRCh38
rs1046652353 Chr.21: 26573110-26573110 on Build GRCh38
rs772794233 Chr.21: 26572892-26572892 on Build GRCh38
rs996675405 Chr.21: 26573149-26573149 on Build GRCh38
rs541639134 Chr.21: 26572901-26572901 on Build GRCh38
rs912460270 Chr.21: 26573215-26573215 on Build GRCh38
rs904805802 Chr.21: 26572777-26572777 on Build GRCh38
rs762195618 Chr.21: 26572986-26572986 on Build GRCh38
rs751595053 Chr.21: 26573112-26573112 on Build GRCh38
rs1027170306 Chr.21: 26572773-26572773 on Build GRCh38
rs1039503756 Chr.21: 26572749-26572749 on Build GRCh38
rs140942198 Chr.21: 26572941-26572941 on Build GRCh38
rs548464418 Chr.21: 26573143-26573143 on Build GRCh38
rs770545016 Chr.21: 26572961-26572961 on Build GRCh38
rs1019317489 Chr.21: 26573114-26573114 on Build GRCh38
rs755995987 Chr.21: 26573033-26573033 on Build GRCh38
rs948839480 Chr.21: 26573228-26573228 on Build GRCh38
rs749208475 Chr.21: 26573045-26573045 on Build GRCh38
rs1053319472 Chr.21: 26573096-26573096 on Build GRCh38
rs977399124 Chr.21: 26573005-26573005 on Build GRCh38
rs752820529 Chr.21: 26573152-26573174 on Build GRCh38
rs778094973 Chr.21: 26572820-26572820 on Build GRCh38
rs892093088 Chr.21: 26573103-26573103 on Build GRCh38
rs758940931 Chr.21: 26573013-26573013 on Build GRCh38
rs772215473 Chr.21: 26572842-26572842 on Build GRCh38
rs1049004285 Chr.21: 26573100-26573100 on Build GRCh38
rs569358844 Chr.21: 26573025-26573025 on Build GRCh38
rs1021868952 Chr.21: 26573178-26573178 on Build GRCh38
rs1033699297 Chr.21: 26572850-26572850 on Build GRCh38
rs746729718 Chr.21: 26572866-26572866 on Build GRCh38
rs952586973 Chr.21: 26573171-26573171 on Build GRCh38
rs1028602261 Chr.21: 26573156-26573156 on Build GRCh38
rs776333672 Chr.21: 26572868-26572868 on Build GRCh38
rs984012632 Chr.21: 26573172-26573172 on Build GRCh38
rs992862046 Chr.21: 26573213-26573213 on Build GRCh38
rs1039406706 Chr.21: 26573044-26573044 on Build GRCh38
rs778747021 Chr.21: 26573250-26573250 on Build GRCh38
rs766812419 Chr.21: 26573106-26573106 on Build GRCh38
rs1019325883 Chr.21: 26572858-26572858 on Build GRCh38
rs763440110 Chr.21: 26573094-26573094 on Build GRCh38
rs900960083 Chr.21: 26573121-26573121 on Build GRCh38
rs568653719 Chr.21: 26573190-26573190 on Build GRCh38
rs761808985 Chr.21: 26572877-26572877 on Build GRCh38
rs745373706 Chr.21: 26573053-26573053 on Build GRCh38
rs372128791 Chr.21: 26573088-26573088 on Build GRCh38
rs769791881 Chr.21: 26572949-26572949 on Build GRCh38
rs764514093 Chr.21: 26572913-26572913 on Build GRCh38
rs773852667 Chr.21: 26573052-26573052 on Build GRCh38
rs200978810 Chr.21: 26572935-26572935 on Build GRCh38
rs967266417 Chr.21: 26573181-26573181 on Build GRCh38
rs576125420 Chr.21: 26572867-26572867 on Build GRCh38
rs765679917 Chr.21: 26572992-26572992 on Build GRCh38
rs940019409 Chr.21: 26572984-26572984 on Build GRCh38
rs750458342 Chr.21: 26572934-26572934 on Build GRCh38
rs866099992 Chr.21: 26572865-26572865 on Build GRCh38
rs753953923 Chr.21: 26572969-26572970 on Build GRCh38
rs373284632 Chr.21: 26572920-26572920 on Build GRCh38
rs534529050 Chr.21: 26572803-26572803 on Build GRCh38
rs532547845 Chr.21: 26572974-26572974 on Build GRCh38
rs977724482 Chr.21: 26573187-26573188 on Build GRCh38
rs920936675 Chr.21: 26573222-26573222 on Build GRCh38
rs113556917 Chr.21: 26573142-26573142 on Build GRCh38
rs1009248078 Chr.21: 26572855-26572855 on Build GRCh38
rs562239375 Chr.21: 26572830-26572830 on Build GRCh38
rs767730490 Chr.21: 26573139-26573139 on Build GRCh38
rs762504954 Chr.21: 26572958-26572958 on Build GRCh38
rs538362698 Chr.21: 26573127-26573127 on Build GRCh38
rs560478891 Chr.21: 26572968-26572968 on Build GRCh38
rs532591284 Chr.21: 26573186-26573186 on Build GRCh38
rs749106796 Chr.21: 26573194-26573194 on Build GRCh38
rs892145796 Chr.21: 26572841-26572841 on Build GRCh38
rs747912821 Chr.21: 26572942-26572942 on Build GRCh38
rs1028240234 Chr.21: 26572893-26572893 on Build GRCh38
rs1002225046 Chr.21: 26572840-26572840 on Build GRCh38
rs1022050534 Chr.21: 26572750-26572763 on Build GRCh38
rs751651024 Chr.21: 26573021-26573021 on Build GRCh38
rs773314119 Chr.21: 26572955-26572955 on Build GRCh38
rs759475710 Chr.21: 26572993-26572994 on Build GRCh38
rs906810433 Chr.21: 26573132-26573132 on Build GRCh38
rs752946614 Chr.21: 26573164-26573164 on Build GRCh38
rs369765568 Chr.21: 26572827-26572827 on Build GRCh38
rs759211669 Chr.21: 26572966-26572966 on Build GRCh38
rs553317321 Chr.21: 26572816-26572816 on Build GRCh38
rs113151255 Chr.21: 26572788-26572788 on Build GRCh38
rs781279668 Chr.21: 26573027-26573027 on Build GRCh38
rs756446566 Chr.21: 26573180-26573180 on Build GRCh38
rs931055135 Chr.21: 26573223-26573223 on Build GRCh38
rs771047962 Chr.21: 26573051-26573051 on Build GRCh38
rs755048541 Chr.21: 26573135-26573135 on Build GRCh38
rs926091525 Chr.21: 26573252-26573252 on Build GRCh38
rs775022750 Chr.21: 26573071-26573071 on Build GRCh38
rs535278986 Chr.21: 26573061-26573061 on Build GRCh38
rs965209899 Chr.21: 26572861-26572861 on Build GRCh38
rs760365559 Chr.21: 26573075-26573075 on Build GRCh38
rs773630649 Chr.21: 26573090-26573090 on Build GRCh38
rs777761841 Chr.21: 26573042-26573042 on Build GRCh38
rs534185584 Chr.21: 26573244-26573244 on Build GRCh38
rs750904627 Chr.21: 26573010-26573010 on Build GRCh38
rs745960184 Chr.21: 26572831-26572831 on Build GRCh38
rs762557964 Chr.21: 26572896-26572896 on Build GRCh38
rs931086980 Chr.21: 26573076-26573076 on Build GRCh38
rs996056486 Chr.21: 26572754-26572754 on Build GRCh38
rs780282523 Chr.21: 26572863-26572863 on Build GRCh38
rs200490811 Chr.21: 26572977-26572977 on Build GRCh38
rs62216535 Chr.21: 26572822-26572822 on Build GRCh38
rs561942992 Chr.21: 26572931-26572931 on Build GRCh38
rs773776527 Chr.21: 26572964-26572964 on Build GRCh38

View Full Product Details