Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99129050-99129639 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SYNM
Entrez Gene ID
23336
Gene Name
synemin
Gene Aliases
DMN, SYN
UniGene
Hs.207106
Gene Chromosome Location
Chr.15: 99105069-99136586 on Build GRCh38
Transcripts
NM_015286.5 NM_145728.2 XM_017022035.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3887843 c.1173T>G p.G391G Chr.15: 99129536-99129536 on Build GRCh38
3402041 c.1109G>A p.S370N Chr.15: 99129472-99129472 on Build GRCh38
6795224 c.1243G>A p.G415R Chr.15: 99129606-99129606 on Build GRCh38
3999697 c.1259G>A p.S420N Chr.15: 99129622-99129622 on Build GRCh38
7275740 c.1217C>T p.A406V Chr.15: 99129580-99129580 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs887397798 Chr.15: 99129341-99129341 on Build GRCh38
rs782772728 Chr.15: 99129425-99129425 on Build GRCh38
rs782324812 Chr.15: 99129475-99129475 on Build GRCh38
rs971108707 Chr.15: 99129427-99129427 on Build GRCh38
rs547691414 Chr.15: 99129210-99129210 on Build GRCh38
rs763067856 Chr.15: 99129410-99129410 on Build GRCh38
rs782224745 Chr.15: 99129471-99129471 on Build GRCh38
rs782165312 Chr.15: 99129577-99129577 on Build GRCh38
rs782209355 Chr.15: 99129564-99129564 on Build GRCh38
rs782193575 Chr.15: 99129568-99129568 on Build GRCh38
rs782217745 Chr.15: 99129351-99129351 on Build GRCh38
rs571481653 Chr.15: 99129161-99129161 on Build GRCh38
rs782400600 Chr.15: 99129491-99129491 on Build GRCh38
rs558695170 Chr.15: 99129547-99129547 on Build GRCh38
rs782600325 Chr.15: 99129429-99129429 on Build GRCh38
rs782348919 Chr.15: 99129366-99129366 on Build GRCh38
rs782527382 Chr.15: 99129614-99129614 on Build GRCh38
rs373024133 Chr.15: 99129111-99129111 on Build GRCh38
rs782705010 Chr.15: 99129413-99129413 on Build GRCh38
rs374368974 Chr.15: 99129407-99129407 on Build GRCh38
rs914470407 Chr.15: 99129528-99129528 on Build GRCh38
rs575103038 Chr.15: 99129248-99129248 on Build GRCh38
rs782144896 Chr.15: 99129421-99129421 on Build GRCh38
rs781881876 Chr.15: 99129606-99129606 on Build GRCh38
rs988143921 Chr.15: 99129140-99129140 on Build GRCh38
rs782543837 Chr.15: 99129551-99129551 on Build GRCh38
rs971223295 Chr.15: 99129078-99129078 on Build GRCh38
rs781927247 Chr.15: 99129571-99129571 on Build GRCh38
rs565868015 Chr.15: 99129215-99129215 on Build GRCh38
rs536759583 Chr.15: 99129524-99129524 on Build GRCh38
rs782487161 Chr.15: 99129535-99129535 on Build GRCh38
rs371251399 Chr.15: 99129396-99129396 on Build GRCh38
rs782602174 Chr.15: 99129586-99129586 on Build GRCh38
rs367716061 Chr.15: 99129517-99129517 on Build GRCh38
rs782811370 Chr.15: 99129605-99129605 on Build GRCh38
rs782603809 Chr.15: 99129542-99129542 on Build GRCh38
rs782282943 Chr.15: 99129457-99129457 on Build GRCh38
rs781827727 Chr.15: 99129591-99129591 on Build GRCh38
rs782392945 Chr.15: 99129466-99129466 on Build GRCh38
rs782507566 Chr.15: 99129346-99129346 on Build GRCh38
rs955941252 Chr.15: 99129126-99129126 on Build GRCh38
rs781918612 Chr.15: 99129476-99129476 on Build GRCh38
rs782255135 Chr.15: 99129561-99129561 on Build GRCh38
rs782635417 Chr.15: 99129348-99129348 on Build GRCh38
rs532419864 Chr.15: 99129167-99129167 on Build GRCh38
rs371133008 Chr.15: 99129373-99129373 on Build GRCh38
rs372377561 Chr.15: 99129578-99129578 on Build GRCh38
rs1051244248 Chr.15: 99129306-99129306 on Build GRCh38
rs782062605 Chr.15: 99129412-99129412 on Build GRCh38
rs373616613 Chr.15: 99129514-99129514 on Build GRCh38
rs10444901 Chr.15: 99129148-99129148 on Build GRCh38
rs540983762 Chr.15: 99129563-99129563 on Build GRCh38
rs782637067 Chr.15: 99129469-99129469 on Build GRCh38
rs543392968 Chr.15: 99129445-99129445 on Build GRCh38
rs781884023 Chr.15: 99129338-99129338 on Build GRCh38
rs781915200 Chr.15: 99129190-99129190 on Build GRCh38
rs781902368 Chr.15: 99129420-99129420 on Build GRCh38
rs782671153 Chr.15: 99129555-99129555 on Build GRCh38
rs569686388 Chr.15: 99129504-99129504 on Build GRCh38
rs782069904 Chr.15: 99129505-99129505 on Build GRCh38
rs782115557 Chr.15: 99129586-99129586 on Build GRCh38
rs376775614 Chr.15: 99129562-99129562 on Build GRCh38
rs1038583772 Chr.15: 99129364-99129364 on Build GRCh38
rs782620136 Chr.15: 99129615-99129615 on Build GRCh38
rs782371078 Chr.15: 99129404-99129404 on Build GRCh38
rs782252958 Chr.15: 99129424-99129424 on Build GRCh38
rs782374281 Chr.15: 99129574-99129574 on Build GRCh38
rs782113960 Chr.15: 99129503-99129503 on Build GRCh38
rs372221449 Chr.15: 99129518-99129518 on Build GRCh38
rs921327146 Chr.15: 99129223-99129223 on Build GRCh38
rs782719964 Chr.15: 99129512-99129512 on Build GRCh38
rs1051299106 Chr.15: 99129566-99129566 on Build GRCh38
rs782435411 Chr.15: 99129603-99129603 on Build GRCh38
rs981212972 Chr.15: 99129089-99129089 on Build GRCh38
rs3743242 Chr.15: 99129423-99129423 on Build GRCh38
rs782022101 Chr.15: 99129402-99129402 on Build GRCh38
rs782489690 Chr.15: 99129428-99129428 on Build GRCh38
rs202151579 Chr.15: 99129450-99129450 on Build GRCh38
rs782004737 Chr.15: 99129583-99129583 on Build GRCh38
rs1040209430 Chr.15: 99129195-99129195 on Build GRCh38
rs946706611 Chr.15: 99129169-99129169 on Build GRCh38
rs898156647 Chr.15: 99129372-99129372 on Build GRCh38
rs782157651 Chr.15: 99129490-99129490 on Build GRCh38
rs376683602 Chr.15: 99129395-99129395 on Build GRCh38

View Full Product Details