Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 54173609-54174197 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MBOAT7
Entrez Gene ID
79143
Gene Name
membrane bound O-acyltransferase domain containing 7
Gene Aliases
BB1, LENG4, LPIAT, LRC4, MBOA7, OACT7, hMBOA-7
UniGene
Hs.467279
Gene Chromosome Location
Chr.19: 54173412-54190050 on Build GRCh38
Transcripts
NM_001146056.2 NM_001146083.2 NM_024298.4 XM_011527299.2 XM_011527300.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3538613 c.1283C>T p.S428F Chr.19: 54174180-54174180 on Build GRCh38
4081211 c.1377C>T p.S459S Chr.19: 54174086-54174086 on Build GRCh38
4845074 c.1361G>A p.R454Q Chr.19: 54174102-54174102 on Build GRCh38
1305076 c.1344T>G p.G448G Chr.19: 54174119-54174119 on Build GRCh38
3933170 c.1351A>C p.S451R Chr.19: 54174112-54174112 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs964114026 Chr.19: 54173987-54173987 on Build GRCh38
rs774674992 Chr.19: 54174170-54174170 on Build GRCh38
rs372068130 Chr.19: 54173970-54173970 on Build GRCh38
rs753504958 Chr.19: 54174055-54174055 on Build GRCh38
rs1002252855 Chr.19: 54173653-54173656 on Build GRCh38
rs753922894 Chr.19: 54173759-54173759 on Build GRCh38
rs949381184 Chr.19: 54173806-54173806 on Build GRCh38
rs767033347 Chr.19: 54174035-54174035 on Build GRCh38
rs972708873 Chr.19: 54173770-54173770 on Build GRCh38
rs374689587 Chr.19: 54174010-54174010 on Build GRCh38
rs4806721 Chr.19: 54174065-54174065 on Build GRCh38
rs756906134 Chr.19: 54174079-54174080 on Build GRCh38
rs1025602485 Chr.19: 54173695-54173695 on Build GRCh38
rs1020228855 Chr.19: 54173668-54173668 on Build GRCh38
rs926598101 Chr.19: 54173991-54173991 on Build GRCh38
rs548203729 Chr.19: 54174081-54174102 on Build GRCh38
rs781513873 Chr.19: 54174068-54174068 on Build GRCh38
rs892948124 Chr.19: 54173885-54173886 on Build GRCh38
rs140333674 Chr.19: 54173836-54173839 on Build GRCh38
rs746991748 Chr.19: 54174067-54174067 on Build GRCh38
rs560947151 Chr.19: 54173942-54173942 on Build GRCh38
rs79199039 Chr.19: 54174099-54174099 on Build GRCh38
rs770522908 Chr.19: 54173801-54173801 on Build GRCh38
rs542729679 Chr.19: 54173886-54173886 on Build GRCh38
rs914177362 Chr.19: 54173817-54173817 on Build GRCh38
rs906840198 Chr.19: 54173644-54173644 on Build GRCh38
rs938074510 Chr.19: 54173861-54173861 on Build GRCh38
rs375191001 Chr.19: 54173993-54173993 on Build GRCh38
rs771972635 Chr.19: 54173982-54173982 on Build GRCh38
rs936875730 Chr.19: 54173779-54173779 on Build GRCh38
rs1025466329 Chr.19: 54173956-54173956 on Build GRCh38
rs760122418 Chr.19: 54174098-54174098 on Build GRCh38
rs925354400 Chr.19: 54173763-54173763 on Build GRCh38
rs763878395 Chr.19: 54174131-54174131 on Build GRCh38
rs73062690 Chr.19: 54173954-54173954 on Build GRCh38
rs377278786 Chr.19: 54173661-54173661 on Build GRCh38
rs759920987 Chr.19: 54174028-54174028 on Build GRCh38
rs750495395 Chr.19: 54174085-54174085 on Build GRCh38
rs995339457 Chr.19: 54173694-54173694 on Build GRCh38
rs577015014 Chr.19: 54173846-54173846 on Build GRCh38
rs780464028 Chr.19: 54174149-54174149 on Build GRCh38
rs1034106753 Chr.19: 54173656-54173656 on Build GRCh38
rs759231014 Chr.19: 54174102-54174102 on Build GRCh38
rs58694079 Chr.19: 54173838-54173841 on Build GRCh38
rs754722210 Chr.19: 54174060-54174060 on Build GRCh38
rs2556359 Chr.19: 54173662-54173662 on Build GRCh38
rs909347206 Chr.19: 54173733-54173733 on Build GRCh38
rs1041517822 Chr.19: 54174080-54174080 on Build GRCh38
rs912575734 Chr.19: 54173753-54173753 on Build GRCh38
rs746122975 Chr.19: 54174071-54174071 on Build GRCh38
rs142954889 Chr.19: 54174100-54174100 on Build GRCh38
rs756463106 Chr.19: 54174143-54174143 on Build GRCh38
rs187952084 Chr.19: 54174091-54174091 on Build GRCh38
rs372936043 Chr.19: 54174138-54174138 on Build GRCh38
rs554225371 Chr.19: 54174174-54174174 on Build GRCh38
rs533980097 Chr.19: 54174153-54174153 on Build GRCh38
rs774928119 Chr.19: 54174097-54174097 on Build GRCh38
rs769080314 Chr.19: 54174169-54174169 on Build GRCh38
rs1001569486 Chr.19: 54173891-54173891 on Build GRCh38
rs905322373 Chr.19: 54173652-54173652 on Build GRCh38
rs560327525 Chr.19: 54174011-54174011 on Build GRCh38
rs958096059 Chr.19: 54174015-54174015 on Build GRCh38
rs962539130 Chr.19: 54173679-54173679 on Build GRCh38
rs368279422 Chr.19: 54173915-54173915 on Build GRCh38
rs763410948 Chr.19: 54174033-54174033 on Build GRCh38
rs770304033 Chr.19: 54174029-54174029 on Build GRCh38
rs748735134 Chr.19: 54173835-54173836 on Build GRCh38
rs988160383 Chr.19: 54174016-54174016 on Build GRCh38
rs563420117 Chr.19: 54173760-54173760 on Build GRCh38
rs781462426 Chr.19: 54174141-54174141 on Build GRCh38
rs761850542 Chr.19: 54174061-54174061 on Build GRCh38
rs765952180 Chr.19: 54174050-54174050 on Build GRCh38
rs574800081 Chr.19: 54173683-54173683 on Build GRCh38
rs371687177 Chr.19: 54173704-54173704 on Build GRCh38
rs963878633 Chr.19: 54173966-54173966 on Build GRCh38
rs1053889570 Chr.19: 54173879-54173879 on Build GRCh38
rs1046463888 Chr.19: 54173649-54173649 on Build GRCh38
rs926865984 Chr.19: 54174058-54174058 on Build GRCh38
rs1001306327 Chr.19: 54173653-54173653 on Build GRCh38
rs752497719 Chr.19: 54174127-54174127 on Build GRCh38
rs202068416 Chr.19: 54174036-54174036 on Build GRCh38
rs906876637 Chr.19: 54173890-54173890 on Build GRCh38
rs1030077665 Chr.19: 54173698-54173698 on Build GRCh38
rs746065377 Chr.19: 54174142-54174142 on Build GRCh38
rs949412164 Chr.19: 54173637-54173637 on Build GRCh38
rs1008151880 Chr.19: 54173979-54173979 on Build GRCh38
rs144074025 Chr.19: 54173722-54173722 on Build GRCh38
rs769359319 Chr.19: 54174126-54174127 on Build GRCh38
rs373348958 Chr.19: 54174032-54174032 on Build GRCh38
rs554021557 Chr.19: 54173815-54173815 on Build GRCh38
rs770252619 Chr.19: 54174084-54174084 on Build GRCh38
rs758215036 Chr.19: 54174130-54174130 on Build GRCh38
rs951265060 Chr.19: 54173705-54173705 on Build GRCh38
rs760318435 Chr.19: 54174038-54174038 on Build GRCh38
rs765063268 Chr.19: 54174103-54174103 on Build GRCh38
rs867456958 Chr.19: 54174156-54174156 on Build GRCh38
rs148708963 Chr.19: 54174066-54174066 on Build GRCh38
rs777760704 Chr.19: 54174004-54174004 on Build GRCh38
rs758110672 Chr.19: 54173971-54173972 on Build GRCh38
rs776995662 Chr.19: 54174027-54174027 on Build GRCh38
rs562820980 Chr.19: 54173895-54173895 on Build GRCh38
rs183211366 Chr.19: 54173921-54173921 on Build GRCh38
rs548244158 Chr.19: 54174096-54174096 on Build GRCh38
rs745362588 Chr.19: 54174105-54174106 on Build GRCh38
rs749789404 Chr.19: 54174094-54174094 on Build GRCh38
rs1029961533 Chr.19: 54173911-54173911 on Build GRCh38

View Full Product Details