Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98069064-98069659 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NANS
Entrez Gene ID
54187
Gene Name
N-acetylneuraminate synthase
Gene Aliases
HEL-S-100, SAS, SEMDG
UniGene
Hs.522310
Gene Chromosome Location
Chr.9: 98056677-98083083 on Build GRCh38
Transcripts
NM_018946.3 XM_011518787.2 XM_017014811.1

SNP Detail

SNP ID Chromosome Location
rs751603834 Chr.9: 98069140-98069140 on Build GRCh38
rs145156141 Chr.9: 98069297-98069297 on Build GRCh38
rs952042204 Chr.9: 98069136-98069136 on Build GRCh38
rs1007488636 Chr.9: 98069466-98069466 on Build GRCh38
rs546194502 Chr.9: 98069481-98069481 on Build GRCh38
rs752948004 Chr.9: 98069455-98069455 on Build GRCh38
rs1017848214 Chr.9: 98069229-98069229 on Build GRCh38
rs554408575 Chr.9: 98069310-98069310 on Build GRCh38
rs538731795 Chr.9: 98069122-98069122 on Build GRCh38
rs997695930 Chr.9: 98069537-98069537 on Build GRCh38
rs559702291 Chr.9: 98069514-98069514 on Build GRCh38
rs183690320 Chr.9: 98069192-98069192 on Build GRCh38
rs74813260 Chr.9: 98069143-98069143 on Build GRCh38
rs543670255 Chr.9: 98069339-98069339 on Build GRCh38
rs949928512 Chr.9: 98069420-98069420 on Build GRCh38
rs767696541 Chr.9: 98069435-98069435 on Build GRCh38
rs980409882 Chr.9: 98069562-98069562 on Build GRCh38
rs976246759 Chr.9: 98069240-98069240 on Build GRCh38
rs887851479 Chr.9: 98069490-98069490 on Build GRCh38
rs78988385 Chr.9: 98069160-98069160 on Build GRCh38
rs562085832 Chr.9: 98069591-98069591 on Build GRCh38
rs935644659 Chr.9: 98069338-98069338 on Build GRCh38
rs904077181 Chr.9: 98069165-98069165 on Build GRCh38
rs368830234 Chr.9: 98069366-98069366 on Build GRCh38
rs377011919 Chr.9: 98069594-98069594 on Build GRCh38
rs188660026 Chr.9: 98069296-98069296 on Build GRCh38
rs114143001 Chr.9: 98069434-98069434 on Build GRCh38
rs915869650 Chr.9: 98069416-98069416 on Build GRCh38
rs998041541 Chr.9: 98069359-98069359 on Build GRCh38
rs897926124 Chr.9: 98069110-98069110 on Build GRCh38
rs183572826 Chr.9: 98069518-98069518 on Build GRCh38
rs1025933607 Chr.9: 98069534-98069534 on Build GRCh38
rs951244574 Chr.9: 98069535-98069535 on Build GRCh38
rs993993367 Chr.9: 98069449-98069449 on Build GRCh38
rs1049110540 Chr.9: 98069486-98069486 on Build GRCh38
rs892665201 Chr.9: 98069628-98069628 on Build GRCh38
rs545718149 Chr.9: 98069552-98069552 on Build GRCh38
rs917662980 Chr.9: 98069542-98069542 on Build GRCh38
rs192558691 Chr.9: 98069316-98069316 on Build GRCh38
rs947684114 Chr.9: 98069153-98069153 on Build GRCh38
rs368807074 Chr.9: 98069239-98069239 on Build GRCh38
rs914060389 Chr.9: 98069279-98069279 on Build GRCh38
rs1018167636 Chr.9: 98069505-98069505 on Build GRCh38
rs925527030 Chr.9: 98069564-98069564 on Build GRCh38
rs376004451 Chr.9: 98069251-98069281 on Build GRCh38
rs76206620 Chr.9: 98069292-98069292 on Build GRCh38
rs992341646 Chr.9: 98069536-98069536 on Build GRCh38
rs115954384 Chr.9: 98069133-98069133 on Build GRCh38
rs186744621 Chr.9: 98069523-98069523 on Build GRCh38
rs977580406 Chr.9: 98069576-98069576 on Build GRCh38

View Full Product Details