Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 62474043-62474637 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO11
Entrez Gene ID
51194
Gene Name
importin 11
Gene Aliases
RanBP11
UniGene
Hs.482269
Gene Chromosome Location
Chr.5: 62412746-62628589 on Build GRCh38
Transcripts
NM_001134779.1 NM_016338.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6686638 c.711delT p.L239fs*8 Chr.5: 62474418-62474418 on Build GRCh38
5344958 c.859A>G p.K287E Chr.5: 62474446-62474446 on Build GRCh38
4692811 c.724A>G p.I242V Chr.5: 62474431-62474431 on Build GRCh38
4692813 c.745T>C p.F249L Chr.5: 62474452-62474452 on Build GRCh38
6686633 c.829-8delT p.? Chr.5: 62474408-62474408 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374309016 Chr.5: 62474472-62474472 on Build GRCh38
rs777245828 Chr.5: 62474506-62474509 on Build GRCh38
rs757293970 Chr.5: 62474228-62474228 on Build GRCh38
rs576287654 Chr.5: 62474152-62474152 on Build GRCh38
rs757106213 Chr.5: 62474403-62474403 on Build GRCh38
rs532857463 Chr.5: 62474431-62474431 on Build GRCh38
rs759141926 Chr.5: 62474446-62474446 on Build GRCh38
rs563098431 Chr.5: 62474425-62474425 on Build GRCh38
rs1039138695 Chr.5: 62474204-62474204 on Build GRCh38
rs140147182 Chr.5: 62474429-62474429 on Build GRCh38
rs376181935 Chr.5: 62474500-62474500 on Build GRCh38
rs751556732 Chr.5: 62474494-62474494 on Build GRCh38
rs112241089 Chr.5: 62474266-62474266 on Build GRCh38
rs770021355 Chr.5: 62474418-62474418 on Build GRCh38
rs552234408 Chr.5: 62474582-62474582 on Build GRCh38
rs138205751 Chr.5: 62474304-62474304 on Build GRCh38
rs78289519 Chr.5: 62474128-62474128 on Build GRCh38
rs944447264 Chr.5: 62474294-62474294 on Build GRCh38
rs26633 Chr.5: 62474158-62474158 on Build GRCh38
rs1016369137 Chr.5: 62474291-62474291 on Build GRCh38
rs1033549204 Chr.5: 62474353-62474353 on Build GRCh38
rs373466825 Chr.5: 62474521-62474524 on Build GRCh38
rs767881366 Chr.5: 62474442-62474442 on Build GRCh38
rs769080623 Chr.5: 62474430-62474430 on Build GRCh38
rs766024933 Chr.5: 62474305-62474305 on Build GRCh38
rs746061516 Chr.5: 62474262-62474262 on Build GRCh38
rs142408263 Chr.5: 62474443-62474443 on Build GRCh38
rs557073869 Chr.5: 62474130-62474130 on Build GRCh38
rs187511215 Chr.5: 62474111-62474111 on Build GRCh38
rs757922963 Chr.5: 62474476-62474476 on Build GRCh38
rs1013264622 Chr.5: 62474563-62474563 on Build GRCh38
rs917605004 Chr.5: 62474110-62474110 on Build GRCh38
rs956715364 Chr.5: 62474224-62474224 on Build GRCh38
rs969472299 Chr.5: 62474192-62474192 on Build GRCh38
rs925005501 Chr.5: 62474151-62474151 on Build GRCh38
rs910723823 Chr.5: 62474282-62474282 on Build GRCh38
rs115288756 Chr.5: 62474578-62474578 on Build GRCh38
rs749399568 Chr.5: 62474172-62474172 on Build GRCh38
rs192575399 Chr.5: 62474209-62474209 on Build GRCh38
rs927932279 Chr.5: 62474214-62474214 on Build GRCh38
rs371332789 Chr.5: 62474404-62474404 on Build GRCh38
rs543410791 Chr.5: 62474347-62474347 on Build GRCh38
rs762219763 Chr.5: 62474440-62474440 on Build GRCh38
rs1024870329 Chr.5: 62474493-62474493 on Build GRCh38
rs776290688 Chr.5: 62474475-62474475 on Build GRCh38
rs544932016 Chr.5: 62474156-62474156 on Build GRCh38
rs547961587 Chr.5: 62474190-62474190 on Build GRCh38
rs750648596 Chr.5: 62474211-62474211 on Build GRCh38
rs993375044 Chr.5: 62474098-62474098 on Build GRCh38
rs988134169 Chr.5: 62474225-62474225 on Build GRCh38
rs559727178 Chr.5: 62474584-62474584 on Build GRCh38
rs570549952 Chr.5: 62474495-62474495 on Build GRCh38
rs560693591 Chr.5: 62474259-62474259 on Build GRCh38
rs1003551250 Chr.5: 62474606-62474606 on Build GRCh38
rs548790954 Chr.5: 62474571-62474571 on Build GRCh38
rs373781717 Chr.5: 62474336-62474336 on Build GRCh38
rs755683024 Chr.5: 62474390-62474391 on Build GRCh38
rs781076408 Chr.5: 62474412-62474412 on Build GRCh38
rs779241470 Chr.5: 62474246-62474246 on Build GRCh38
rs149573335 Chr.5: 62474485-62474485 on Build GRCh38
rs992084181 Chr.5: 62474082-62474082 on Build GRCh38
rs764892995 Chr.5: 62474447-62474447 on Build GRCh38
rs781166969 Chr.5: 62474498-62474498 on Build GRCh38
rs936463761 Chr.5: 62474602-62474602 on Build GRCh38
rs144232729 Chr.5: 62474536-62474536 on Build GRCh38
rs897460349 Chr.5: 62474426-62474426 on Build GRCh38
rs540813908 Chr.5: 62474245-62474245 on Build GRCh38
rs746869318 Chr.5: 62474383-62474383 on Build GRCh38

View Full Product Details