Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 134857307-134857835 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5orf24
Entrez Gene ID
134553
Gene Name
chromosome 5 open reading frame 24
Gene Aliases
-
UniGene
Hs.708536
Gene Chromosome Location
Chr.5: 134833603-134859735 on Build GRCh38
Transcripts
NM_001135586.1 NM_001300894.1 NM_152409.3 XM_005271889.3 XM_017009049.1 XM_017009050.1

SNP Detail

SNP ID Chromosome Location
rs1003918874 Chr.5: 134857696-134857696 on Build GRCh38
rs1011313586 Chr.5: 134857458-134857458 on Build GRCh38
rs751950966 Chr.5: 134857719-134857719 on Build GRCh38
rs148502660 Chr.5: 134857784-134857784 on Build GRCh38
rs747628225 Chr.5: 134857356-134857356 on Build GRCh38
rs907859387 Chr.5: 134857551-134857551 on Build GRCh38
rs115396590 Chr.5: 134857474-134857474 on Build GRCh38
rs116661206 Chr.5: 134857794-134857794 on Build GRCh38
rs1053772409 Chr.5: 134857543-134857543 on Build GRCh38
rs996759228 Chr.5: 134857797-134857797 on Build GRCh38
rs925927246 Chr.5: 134857516-134857516 on Build GRCh38
rs938550327 Chr.5: 134857664-134857664 on Build GRCh38
rs373114001 Chr.5: 134857372-134857372 on Build GRCh38
rs551228259 Chr.5: 134857626-134857626 on Build GRCh38
rs952442123 Chr.5: 134857349-134857349 on Build GRCh38
rs559805106 Chr.5: 134857578-134857578 on Build GRCh38
rs1031307708 Chr.5: 134857384-134857384 on Build GRCh38
rs888109963 Chr.5: 134857688-134857688 on Build GRCh38
rs955785801 Chr.5: 134857451-134857451 on Build GRCh38
rs761850116 Chr.5: 134857530-134857533 on Build GRCh38
rs537119206 Chr.5: 134857708-134857708 on Build GRCh38
rs548760168 Chr.5: 134857686-134857689 on Build GRCh38
rs1021832111 Chr.5: 134857473-134857473 on Build GRCh38
rs755437349 Chr.5: 134857749-134857749 on Build GRCh38
rs545929215 Chr.5: 134857421-134857421 on Build GRCh38
rs552406540 Chr.5: 134857346-134857346 on Build GRCh38
rs1011366231 Chr.5: 134857734-134857734 on Build GRCh38
rs1015675862 Chr.5: 134857702-134857702 on Build GRCh38
rs569364389 Chr.5: 134857655-134857655 on Build GRCh38
rs895711358 Chr.5: 134857704-134857704 on Build GRCh38
rs1014136846 Chr.5: 134857737-134857737 on Build GRCh38
rs932378470 Chr.5: 134857682-134857682 on Build GRCh38
rs973444540 Chr.5: 134857612-134857612 on Build GRCh38
rs1044198279 Chr.5: 134857658-134857659 on Build GRCh38
rs934717882 Chr.5: 134857517-134857517 on Build GRCh38
rs78987868 Chr.5: 134857353-134857353 on Build GRCh38
rs868807537 Chr.5: 134857683-134857683 on Build GRCh38
rs528481653 Chr.5: 134857393-134857393 on Build GRCh38
rs978843610 Chr.5: 134857512-134857512 on Build GRCh38
rs528617744 Chr.5: 134857652-134857652 on Build GRCh38
rs145780106 Chr.5: 134857497-134857497 on Build GRCh38
rs377231492 Chr.5: 134857449-134857449 on Build GRCh38
rs775983826 Chr.5: 134857479-134857479 on Build GRCh38
rs979251510 Chr.5: 134857799-134857799 on Build GRCh38
rs907121652 Chr.5: 134857660-134857660 on Build GRCh38
rs1048168437 Chr.5: 134857684-134857684 on Build GRCh38
rs777113122 Chr.5: 134857417-134857417 on Build GRCh38
rs114887018 Chr.5: 134857633-134857633 on Build GRCh38
rs563305516 Chr.5: 134857371-134857371 on Build GRCh38
rs1031004965 Chr.5: 134857505-134857505 on Build GRCh38

View Full Product Details