Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 44360344-44360722 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATS1
Entrez Gene ID
221409
Gene Name
spermatogenesis associated serine rich 1
Gene Aliases
DDIP, SPATA8, SRSP1
UniGene
Hs.135283
Gene Chromosome Location
Chr.6: 44342660-44377167 on Build GRCh38
Transcripts
NM_145026.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3627844 c.370G>A p.D124N Chr.6: 44360528-44360528 on Build GRCh38
5405429 c.369G>A p.K123K Chr.6: 44360527-44360527 on Build GRCh38
1697298 c.364C>T p.Q122* Chr.6: 44360522-44360522 on Build GRCh38
211035 c.326A>G p.H109R Chr.6: 44360484-44360484 on Build GRCh38
3627843 c.370G>A p.D124N Chr.6: 44360528-44360528 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749267646 Chr.6: 44360413-44360413 on Build GRCh38
rs549801826 Chr.6: 44360651-44360651 on Build GRCh38
rs1042957998 Chr.6: 44360644-44360644 on Build GRCh38
rs765738411 Chr.6: 44360472-44360472 on Build GRCh38
rs2274832 Chr.6: 44360662-44360662 on Build GRCh38
rs372441506 Chr.6: 44360427-44360427 on Build GRCh38
rs948530438 Chr.6: 44360700-44360700 on Build GRCh38
rs374774066 Chr.6: 44360445-44360445 on Build GRCh38
rs756022447 Chr.6: 44360396-44360396 on Build GRCh38
rs772325141 Chr.6: 44360428-44360428 on Build GRCh38
rs190235137 Chr.6: 44360375-44360375 on Build GRCh38
rs576317319 Chr.6: 44360406-44360406 on Build GRCh38
rs146592592 Chr.6: 44360373-44360373 on Build GRCh38
rs377248636 Chr.6: 44360573-44360573 on Build GRCh38
rs752437754 Chr.6: 44360546-44360546 on Build GRCh38
rs1041359817 Chr.6: 44360407-44360407 on Build GRCh38
rs368808979 Chr.6: 44360587-44360587 on Build GRCh38
rs768953076 Chr.6: 44360594-44360594 on Build GRCh38
rs759023355 Chr.6: 44360508-44360508 on Build GRCh38
rs962015968 Chr.6: 44360553-44360553 on Build GRCh38
rs141936435 Chr.6: 44360559-44360559 on Build GRCh38
rs371515009 Chr.6: 44360674-44360674 on Build GRCh38
rs775737609 Chr.6: 44360449-44360449 on Build GRCh38
rs758452319 Chr.6: 44360577-44360577 on Build GRCh38
rs553780650 Chr.6: 44360690-44360690 on Build GRCh38
rs770317693 Chr.6: 44360606-44360606 on Build GRCh38
rs370191775 Chr.6: 44360425-44360425 on Build GRCh38
rs551694146 Chr.6: 44360588-44360588 on Build GRCh38
rs747348091 Chr.6: 44360453-44360453 on Build GRCh38
rs535582125 Chr.6: 44360678-44360678 on Build GRCh38
rs947567832 Chr.6: 44360638-44360638 on Build GRCh38
rs914778665 Chr.6: 44360630-44360630 on Build GRCh38
rs777076411 Chr.6: 44360465-44360465 on Build GRCh38
rs369103191 Chr.6: 44360551-44360551 on Build GRCh38
rs760505431 Chr.6: 44360548-44360548 on Build GRCh38
rs149190490 Chr.6: 44360463-44360463 on Build GRCh38
rs551476340 Chr.6: 44360417-44360417 on Build GRCh38
rs762399024 Chr.6: 44360466-44360466 on Build GRCh38
rs757233870 Chr.6: 44360416-44360416 on Build GRCh38
rs757215357 Chr.6: 44360569-44360569 on Build GRCh38
rs150235811 Chr.6: 44360568-44360568 on Build GRCh38
rs1057253188 Chr.6: 44360423-44360423 on Build GRCh38
rs73439562 Chr.6: 44360649-44360649 on Build GRCh38
rs921286456 Chr.6: 44360560-44360560 on Build GRCh38
rs569881327 Chr.6: 44360605-44360605 on Build GRCh38
rs137992604 Chr.6: 44360571-44360571 on Build GRCh38
rs986345381 Chr.6: 44360374-44360374 on Build GRCh38
rs373506829 Chr.6: 44360612-44360612 on Build GRCh38
rs767172409 Chr.6: 44360509-44360509 on Build GRCh38
rs1015522436 Chr.6: 44360550-44360550 on Build GRCh38
rs367976426 Chr.6: 44360411-44360411 on Build GRCh38
rs781539195 Chr.6: 44360603-44360603 on Build GRCh38
rs773904097 Chr.6: 44360506-44360506 on Build GRCh38

View Full Product Details