Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 215577774-215578337 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD3
Entrez Gene ID
51133
Gene Name
potassium channel tetramerization domain containing 3
Gene Aliases
NY-REN-45
UniGene
Hs.335139
Gene Chromosome Location
Chr.1: 215567185-215621821 on Build GRCh38
Transcripts
NM_001319294.1 NM_001319295.1 NM_016121.4 XM_005273158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6692083 c.267G>A p.E89E Chr.1: 215578034-215578034 on Build GRCh38
4028061 c.344T>C p.L115S Chr.1: 215578028-215578028 on Build GRCh38
240319 c.350G>A p.R117H Chr.1: 215578034-215578034 on Build GRCh38
169107 c.340G>T p.E114* Chr.1: 215578024-215578024 on Build GRCh38
392055 c.379delG p.G127fs*13 Chr.1: 215578063-215578063 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs949937969 Chr.1: 215578040-215578040 on Build GRCh38
rs878878908 Chr.1: 215577848-215577848 on Build GRCh38
rs772495411 Chr.1: 215577942-215577942 on Build GRCh38
rs763895825 Chr.1: 215578074-215578074 on Build GRCh38
rs565705517 Chr.1: 215578129-215578129 on Build GRCh38
rs1029103609 Chr.1: 215578146-215578146 on Build GRCh38
rs967137779 Chr.1: 215577873-215577873 on Build GRCh38
rs752499134 Chr.1: 215578150-215578150 on Build GRCh38
rs199535715 Chr.1: 215577967-215577967 on Build GRCh38
rs937182688 Chr.1: 215577969-215577969 on Build GRCh38
rs75328598 Chr.1: 215578256-215578256 on Build GRCh38
rs768258816 Chr.1: 215577833-215577833 on Build GRCh38
rs372012064 Chr.1: 215578076-215578076 on Build GRCh38
rs1045860175 Chr.1: 215578287-215578287 on Build GRCh38
rs571468862 Chr.1: 215578204-215578204 on Build GRCh38
rs978420661 Chr.1: 215578265-215578265 on Build GRCh38
rs761279951 Chr.1: 215578254-215578254 on Build GRCh38
rs1033192423 Chr.1: 215578292-215578292 on Build GRCh38
rs750393753 Chr.1: 215578104-215578104 on Build GRCh38
rs996772161 Chr.1: 215578142-215578142 on Build GRCh38
rs375833467 Chr.1: 215577977-215577977 on Build GRCh38
rs753479757 Chr.1: 215578114-215578114 on Build GRCh38
rs368679009 Chr.1: 215578038-215578038 on Build GRCh38
rs535478393 Chr.1: 215578176-215578176 on Build GRCh38
rs372331747 Chr.1: 215577970-215577970 on Build GRCh38
rs925690130 Chr.1: 215577931-215577931 on Build GRCh38
rs776739978 Chr.1: 215577965-215577965 on Build GRCh38
rs140073774 Chr.1: 215578159-215578159 on Build GRCh38
rs779710590 Chr.1: 215578110-215578110 on Build GRCh38
rs771406569 Chr.1: 215577953-215577953 on Build GRCh38
rs773461881 Chr.1: 215577948-215577948 on Build GRCh38
rs951095658 Chr.1: 215578176-215578176 on Build GRCh38
rs781505014 Chr.1: 215578133-215578133 on Build GRCh38
rs1054425962 Chr.1: 215578033-215578033 on Build GRCh38
rs1020451934 Chr.1: 215578214-215578214 on Build GRCh38
rs909550476 Chr.1: 215578185-215578185 on Build GRCh38
rs1019898817 Chr.1: 215577819-215577819 on Build GRCh38
rs950192147 Chr.1: 215577930-215577930 on Build GRCh38
rs747536577 Chr.1: 215577950-215577950 on Build GRCh38
rs567951649 Chr.1: 215578034-215578034 on Build GRCh38
rs962956420 Chr.1: 215578234-215578234 on Build GRCh38
rs899220479 Chr.1: 215578138-215578138 on Build GRCh38
rs761807245 Chr.1: 215578097-215578097 on Build GRCh38
rs958466057 Chr.1: 215578297-215578297 on Build GRCh38
rs751372529 Chr.1: 215578094-215578094 on Build GRCh38
rs1038200978 Chr.1: 215578136-215578136 on Build GRCh38
rs187978963 Chr.1: 215577906-215577906 on Build GRCh38
rs747371933 Chr.1: 215578126-215578126 on Build GRCh38
rs1029384247 Chr.1: 215578171-215578171 on Build GRCh38
rs754735094 Chr.1: 215578124-215578124 on Build GRCh38
rs151156017 Chr.1: 215577861-215577861 on Build GRCh38
rs998862152 Chr.1: 215577995-215577995 on Build GRCh38
rs759506092 Chr.1: 215577858-215577858 on Build GRCh38
rs771007497 Chr.1: 215578106-215578107 on Build GRCh38
rs924700685 Chr.1: 215578309-215578309 on Build GRCh38
rs755643626 Chr.1: 215578109-215578109 on Build GRCh38
rs770786917 Chr.1: 215577878-215577878 on Build GRCh38
rs376336661 Chr.1: 215578102-215578102 on Build GRCh38
rs916047548 Chr.1: 215577881-215577881 on Build GRCh38
rs950284505 Chr.1: 215578282-215578282 on Build GRCh38
rs991761851 Chr.1: 215578313-215578313 on Build GRCh38

View Full Product Details