Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 105188103-105188615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APPL2
Entrez Gene ID
55198
Gene Name
adaptor protein, phosphotyrosine interacting with PH domain and leucine zipper 2
Gene Aliases
DIP13B
UniGene
Hs.506603
Gene Chromosome Location
Chr.12: 105173297-105236230 on Build GRCh38
Transcripts
NM_001251904.1 NM_001251905.1 NM_018171.3 XM_006719472.1 XM_011538530.2 XM_011538531.2 XM_011538532.2 XM_017019551.1 XM_017019552.1 XM_017019553.1 XM_017019554.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2064878 c.1591C>G p.R531G Chr.12: 105188316-105188316 on Build GRCh38
7356040 c.1585A>G p.I529V Chr.12: 105188322-105188322 on Build GRCh38
7356041 c.1603A>G p.I535V Chr.12: 105188322-105188322 on Build GRCh38
3455416 c.1460-1G>A p.? Chr.12: 105188448-105188448 on Build GRCh38
3455415 c.1491G>A p.R497R Chr.12: 105188416-105188416 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748340674 Chr.12: 105188444-105188445 on Build GRCh38
rs1005534868 Chr.12: 105188343-105188343 on Build GRCh38
rs559602828 Chr.12: 105188241-105188241 on Build GRCh38
rs144240861 Chr.12: 105188417-105188417 on Build GRCh38
rs754194004 Chr.12: 105188291-105188291 on Build GRCh38
rs892277628 Chr.12: 105188152-105188152 on Build GRCh38
rs973987506 Chr.12: 105188553-105188553 on Build GRCh38
rs767454859 Chr.12: 105188275-105188275 on Build GRCh38
rs974721303 Chr.12: 105188175-105188175 on Build GRCh38
rs954974637 Chr.12: 105188255-105188255 on Build GRCh38
rs536236110 Chr.12: 105188418-105188418 on Build GRCh38
rs947804781 Chr.12: 105188592-105188592 on Build GRCh38
rs779226550 Chr.12: 105188245-105188245 on Build GRCh38
rs1196852 Chr.12: 105188467-105188467 on Build GRCh38
rs770766801 Chr.12: 105188459-105188459 on Build GRCh38
rs1054472851 Chr.12: 105188139-105188139 on Build GRCh38
rs1010351647 Chr.12: 105188154-105188154 on Build GRCh38
rs777371226 Chr.12: 105188451-105188451 on Build GRCh38
rs565347239 Chr.12: 105188315-105188315 on Build GRCh38
rs774133852 Chr.12: 105188514-105188514 on Build GRCh38
rs968732539 Chr.12: 105188170-105188170 on Build GRCh38
rs748355709 Chr.12: 105188264-105188264 on Build GRCh38
rs746718259 Chr.12: 105188330-105188330 on Build GRCh38
rs759682022 Chr.12: 105188269-105188269 on Build GRCh38
rs768687003 Chr.12: 105188480-105188480 on Build GRCh38
rs757933124 Chr.12: 105188316-105188316 on Build GRCh38
rs746595601 Chr.12: 105188225-105188225 on Build GRCh38
rs779790593 Chr.12: 105188223-105188223 on Build GRCh38
rs762272581 Chr.12: 105188390-105188390 on Build GRCh38
rs913233986 Chr.12: 105188428-105188428 on Build GRCh38
rs954695680 Chr.12: 105188480-105188481 on Build GRCh38
rs757740468 Chr.12: 105188292-105188292 on Build GRCh38
rs900374435 Chr.12: 105188562-105188562 on Build GRCh38
rs769461870 Chr.12: 105188422-105188422 on Build GRCh38
rs962281133 Chr.12: 105188527-105188527 on Build GRCh38
rs747934315 Chr.12: 105188337-105188337 on Build GRCh38
rs762712188 Chr.12: 105188336-105188336 on Build GRCh38
rs776963128 Chr.12: 105188441-105188442 on Build GRCh38
rs754727159 Chr.12: 105188430-105188430 on Build GRCh38
rs778280004 Chr.12: 105188285-105188285 on Build GRCh38
rs750956406 Chr.12: 105188416-105188416 on Build GRCh38
rs112527494 Chr.12: 105188289-105188289 on Build GRCh38
rs775024469 Chr.12: 105188312-105188312 on Build GRCh38
rs377184900 Chr.12: 105188403-105188403 on Build GRCh38
rs201637640 Chr.12: 105188466-105188466 on Build GRCh38
rs140290194 Chr.12: 105188349-105188349 on Build GRCh38
rs780080660 Chr.12: 105188265-105188265 on Build GRCh38
rs1044765682 Chr.12: 105188339-105188339 on Build GRCh38
rs1003433564 Chr.12: 105188133-105188133 on Build GRCh38
rs763932081 Chr.12: 105188497-105188497 on Build GRCh38
rs751084513 Chr.12: 105188421-105188421 on Build GRCh38
rs769739514 Chr.12: 105188253-105188253 on Build GRCh38
rs147119491 Chr.12: 105188503-105188503 on Build GRCh38
rs550424373 Chr.12: 105188346-105188346 on Build GRCh38
rs755939958 Chr.12: 105188450-105188450 on Build GRCh38
rs771411345 Chr.12: 105188354-105188354 on Build GRCh38
rs138511304 Chr.12: 105188578-105188578 on Build GRCh38
rs746857142 Chr.12: 105188282-105188285 on Build GRCh38
rs368682496 Chr.12: 105188178-105188179 on Build GRCh38
rs779500057 Chr.12: 105188295-105188295 on Build GRCh38
rs368033064 Chr.12: 105188268-105188268 on Build GRCh38
rs528801003 Chr.12: 105188281-105188281 on Build GRCh38
rs529271842 Chr.12: 105188174-105188174 on Build GRCh38
rs768541477 Chr.12: 105188380-105188381 on Build GRCh38
rs145831831 Chr.12: 105188360-105188360 on Build GRCh38
rs747714689 Chr.12: 105188441-105188441 on Build GRCh38
rs761201093 Chr.12: 105188193-105188193 on Build GRCh38
rs200469851 Chr.12: 105188438-105188438 on Build GRCh38
rs764265959 Chr.12: 105188373-105188373 on Build GRCh38
rs754014477 Chr.12: 105188388-105188388 on Build GRCh38
rs79951915 Chr.12: 105188206-105188206 on Build GRCh38
rs749302957 Chr.12: 105188454-105188454 on Build GRCh38
rs945258002 Chr.12: 105188180-105188180 on Build GRCh38
rs775531118 Chr.12: 105188359-105188359 on Build GRCh38
rs768419500 Chr.12: 105188332-105188332 on Build GRCh38
rs147539487 Chr.12: 105188322-105188322 on Build GRCh38
rs770019091 Chr.12: 105188495-105188497 on Build GRCh38
rs906457766 Chr.12: 105188129-105188132 on Build GRCh38
rs752919519 Chr.12: 105188277-105188277 on Build GRCh38
rs776555960 Chr.12: 105188242-105188242 on Build GRCh38
rs990256499 Chr.12: 105188423-105188423 on Build GRCh38
rs1022122136 Chr.12: 105188168-105188168 on Build GRCh38

View Full Product Details