Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 26942743-26943153 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NUDC
Entrez Gene ID
10726
Gene Name
nuclear distribution C, dynein complex regulator
Gene Aliases
HNUDC, MNUDC, NPD011
UniGene
Hs.263812
Gene Chromosome Location
Chr.1: 26911128-26946871 on Build GRCh38
Transcripts
NM_006600.3 XM_011540529.1 XM_011540530.1 XM_017000094.1 XM_017000095.1 XM_017000096.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1639825 c.536C>T p.S179L Chr.1: 26942766-26942766 on Build GRCh38
3741205 c.614G>A p.R205Q Chr.1: 26942938-26942938 on Build GRCh38
907794 c.616C>T p.H206Y Chr.1: 26942940-26942940 on Build GRCh38
5039770 c.712G>A p.G238S Chr.1: 26943036-26943036 on Build GRCh38
3487618 c.621C>T p.L207L Chr.1: 26942945-26942945 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752670310 Chr.1: 26942819-26942819 on Build GRCh38
rs747001801 Chr.1: 26942921-26942921 on Build GRCh38
rs780325348 Chr.1: 26942918-26942918 on Build GRCh38
rs771899838 Chr.1: 26943081-26943081 on Build GRCh38
rs375024836 Chr.1: 26942825-26942825 on Build GRCh38
rs578246670 Chr.1: 26943119-26943119 on Build GRCh38
rs755665481 Chr.1: 26942866-26942866 on Build GRCh38
rs759453469 Chr.1: 26943080-26943080 on Build GRCh38
rs373529231 Chr.1: 26942855-26942855 on Build GRCh38
rs990536856 Chr.1: 26942805-26942805 on Build GRCh38
rs781397057 Chr.1: 26943019-26943019 on Build GRCh38
rs748037809 Chr.1: 26943089-26943089 on Build GRCh38
rs757800953 Chr.1: 26942882-26942882 on Build GRCh38
rs758803019 Chr.1: 26942987-26942987 on Build GRCh38
rs201546227 Chr.1: 26942810-26942810 on Build GRCh38
rs763039829 Chr.1: 26942858-26942858 on Build GRCh38
rs142314176 Chr.1: 26942811-26942811 on Build GRCh38
rs775680543 Chr.1: 26942764-26942764 on Build GRCh38
rs752086600 Chr.1: 26942865-26942865 on Build GRCh38
rs926634344 Chr.1: 26943122-26943122 on Build GRCh38
rs751240291 Chr.1: 26942792-26942792 on Build GRCh38
rs376755295 Chr.1: 26942891-26942891 on Build GRCh38
rs747246668 Chr.1: 26943006-26943006 on Build GRCh38
rs754341066 Chr.1: 26942875-26942875 on Build GRCh38
rs146502618 Chr.1: 26942766-26942766 on Build GRCh38
rs988371609 Chr.1: 26943020-26943020 on Build GRCh38
rs772144546 Chr.1: 26942890-26942890 on Build GRCh38
rs1007200326 Chr.1: 26942935-26942935 on Build GRCh38
rs772387172 Chr.1: 26942841-26942841 on Build GRCh38
rs769036067 Chr.1: 26943017-26943017 on Build GRCh38
rs770927596 Chr.1: 26943044-26943044 on Build GRCh38
rs142020224 Chr.1: 26943035-26943035 on Build GRCh38
rs147512061 Chr.1: 26942972-26942972 on Build GRCh38
rs940137202 Chr.1: 26942838-26942838 on Build GRCh38
rs957514841 Chr.1: 26942803-26942803 on Build GRCh38
rs775382781 Chr.1: 26942843-26942843 on Build GRCh38
rs558519759 Chr.1: 26942968-26942968 on Build GRCh38
rs773295610 Chr.1: 26942857-26942857 on Build GRCh38
rs780339137 Chr.1: 26942995-26942995 on Build GRCh38
rs776915568 Chr.1: 26942928-26942928 on Build GRCh38
rs778260046 Chr.1: 26942816-26942816 on Build GRCh38
rs771037654 Chr.1: 26942934-26942934 on Build GRCh38
rs779227569 Chr.1: 26942886-26942886 on Build GRCh38
rs746047226 Chr.1: 26942827-26942827 on Build GRCh38
rs897568442 Chr.1: 26942852-26942852 on Build GRCh38
rs766069056 Chr.1: 26942791-26942791 on Build GRCh38
rs887403061 Chr.1: 26942945-26942945 on Build GRCh38
rs905290537 Chr.1: 26942879-26942879 on Build GRCh38
rs369768223 Chr.1: 26942930-26942930 on Build GRCh38
rs150079522 Chr.1: 26942922-26942922 on Build GRCh38
rs751406686 Chr.1: 26942860-26942860 on Build GRCh38
rs377621982 Chr.1: 26943091-26943091 on Build GRCh38
rs767233944 Chr.1: 26942863-26942863 on Build GRCh38
rs868227978 Chr.1: 26943037-26943037 on Build GRCh38
rs199748967 Chr.1: 26942985-26942985 on Build GRCh38
rs765242799 Chr.1: 26942767-26942767 on Build GRCh38
rs766270565 Chr.1: 26942859-26942859 on Build GRCh38
rs774766076 Chr.1: 26942826-26942826 on Build GRCh38
rs537966346 Chr.1: 26942946-26942946 on Build GRCh38
rs756541904 Chr.1: 26942812-26942812 on Build GRCh38
rs777325642 Chr.1: 26942873-26942873 on Build GRCh38
rs773981016 Chr.1: 26942937-26942937 on Build GRCh38
rs1056280909 Chr.1: 26942883-26942883 on Build GRCh38
rs774362052 Chr.1: 26943046-26943046 on Build GRCh38
rs746265049 Chr.1: 26942889-26942889 on Build GRCh38
rs760839405 Chr.1: 26942847-26942847 on Build GRCh38
rs753396660 Chr.1: 26942971-26942971 on Build GRCh38
rs749599733 Chr.1: 26942824-26942824 on Build GRCh38
rs778703597 Chr.1: 26943042-26943042 on Build GRCh38
rs777974372 Chr.1: 26942784-26942784 on Build GRCh38
rs1031525845 Chr.1: 26943007-26943007 on Build GRCh38
rs756180278 Chr.1: 26942847-26942848 on Build GRCh38
rs913871571 Chr.1: 26942823-26942823 on Build GRCh38
rs202205518 Chr.1: 26942799-26942799 on Build GRCh38
rs777888059 Chr.1: 26942916-26942918 on Build GRCh38
rs372594675 Chr.1: 26942834-26942834 on Build GRCh38
rs760387987 Chr.1: 26942950-26942950 on Build GRCh38
rs981448496 Chr.1: 26943120-26943120 on Build GRCh38
rs938106483 Chr.1: 26943124-26943124 on Build GRCh38
rs201662428 Chr.1: 26943026-26943026 on Build GRCh38
rs754721688 Chr.1: 26942798-26942798 on Build GRCh38
rs752385591 Chr.1: 26942947-26942947 on Build GRCh38
rs369137943 Chr.1: 26942854-26942854 on Build GRCh38
rs554502070 Chr.1: 26943028-26943028 on Build GRCh38
rs148845287 Chr.1: 26942938-26942938 on Build GRCh38
rs201624714 Chr.1: 26942776-26942776 on Build GRCh38
rs139238058 Chr.1: 26942953-26942953 on Build GRCh38

View Full Product Details