Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 223602144-223602696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SCG2
Entrez Gene ID
7857
Gene Name
secretogranin II
Gene Aliases
CHGC, EM66, SN, SgII
UniGene
Hs.516726
Gene Chromosome Location
Chr.2: 223596940-223602499 on Build GRCh38
Transcripts
NM_003469.4

SNP Detail

SNP ID Chromosome Location
rs529906473 Chr.2: 223602542-223602542 on Build GRCh38
rs540401883 Chr.2: 223602308-223602308 on Build GRCh38
rs533271421 Chr.2: 223602180-223602180 on Build GRCh38
rs560417373 Chr.2: 223602355-223602355 on Build GRCh38
rs777497551 Chr.2: 223602281-223602281 on Build GRCh38
rs369394830 Chr.2: 223602435-223602435 on Build GRCh38
rs189026148 Chr.2: 223602660-223602660 on Build GRCh38
rs1051912042 Chr.2: 223602394-223602394 on Build GRCh38
rs376342367 Chr.2: 223602600-223602600 on Build GRCh38
rs949481965 Chr.2: 223602431-223602431 on Build GRCh38
rs74272275 Chr.2: 223602614-223602614 on Build GRCh38
rs960117523 Chr.2: 223602500-223602501 on Build GRCh38
rs796813016 Chr.2: 223602513-223602515 on Build GRCh38
rs1042415160 Chr.2: 223602448-223602448 on Build GRCh38
rs1026041843 Chr.2: 223602551-223602551 on Build GRCh38
rs184375877 Chr.2: 223602508-223602508 on Build GRCh38
rs60875758 Chr.2: 223602513-223602513 on Build GRCh38
rs373057461 Chr.2: 223602632-223602632 on Build GRCh38
rs1045346498 Chr.2: 223602269-223602269 on Build GRCh38
rs950836611 Chr.2: 223602313-223602313 on Build GRCh38
rs997286879 Chr.2: 223602382-223602382 on Build GRCh38
rs1039118758 Chr.2: 223602520-223602520 on Build GRCh38
rs35882594 Chr.2: 223602537-223602538 on Build GRCh38
rs918037835 Chr.2: 223602423-223602423 on Build GRCh38
rs866322279 Chr.2: 223602274-223602274 on Build GRCh38
rs965778508 Chr.2: 223602275-223602275 on Build GRCh38
rs150862313 Chr.2: 223602617-223602617 on Build GRCh38
rs993810522 Chr.2: 223602458-223602458 on Build GRCh38
rs991225489 Chr.2: 223602364-223602364 on Build GRCh38
rs1007259662 Chr.2: 223602518-223602518 on Build GRCh38
rs991054679 Chr.2: 223602628-223602628 on Build GRCh38
rs11459758 Chr.2: 223602517-223602518 on Build GRCh38
rs530767887 Chr.2: 223602491-223602491 on Build GRCh38
rs950890947 Chr.2: 223602575-223602575 on Build GRCh38
rs550964560 Chr.2: 223602504-223602504 on Build GRCh38
rs33968250 Chr.2: 223602534-223602534 on Build GRCh38
rs997232784 Chr.2: 223602299-223602299 on Build GRCh38
rs997284278 Chr.2: 223602549-223602549 on Build GRCh38
rs1017390408 Chr.2: 223602239-223602239 on Build GRCh38
rs888904067 Chr.2: 223602604-223602604 on Build GRCh38
rs528581715 Chr.2: 223602379-223602379 on Build GRCh38
rs898808211 Chr.2: 223602652-223602655 on Build GRCh38
rs1006783707 Chr.2: 223602576-223602576 on Build GRCh38
rs985074471 Chr.2: 223602325-223602325 on Build GRCh38
rs932993355 Chr.2: 223602519-223602519 on Build GRCh38
rs769319586 Chr.2: 223602279-223602279 on Build GRCh38
rs1026162058 Chr.2: 223602478-223602478 on Build GRCh38
rs200034848 Chr.2: 223602515-223602516 on Build GRCh38
rs925255417 Chr.2: 223602516-223602516 on Build GRCh38
rs1007616737 Chr.2: 223602177-223602177 on Build GRCh38
rs745768847 Chr.2: 223602641-223602641 on Build GRCh38
rs117215799 Chr.2: 223602455-223602455 on Build GRCh38
rs1050090934 Chr.2: 223602532-223602532 on Build GRCh38
rs992625050 Chr.2: 223602220-223602220 on Build GRCh38
rs909442197 Chr.2: 223602331-223602331 on Build GRCh38
rs892614698 Chr.2: 223602381-223602381 on Build GRCh38
rs575306615 Chr.2: 223602622-223602622 on Build GRCh38
rs567428623 Chr.2: 223602515-223602515 on Build GRCh38
rs566762109 Chr.2: 223602587-223602587 on Build GRCh38
rs1052640719 Chr.2: 223602343-223602343 on Build GRCh38
rs947995235 Chr.2: 223602252-223602252 on Build GRCh38
rs1015878535 Chr.2: 223602454-223602454 on Build GRCh38
rs992255911 Chr.2: 223602501-223602501 on Build GRCh38
rs888790961 Chr.2: 223602395-223602395 on Build GRCh38
rs1051930807 Chr.2: 223602564-223602567 on Build GRCh38

View Full Product Details