Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 66816139-66816705 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
NAE1
Entrez Gene ID
8883
Gene Name
NEDD8 activating enzyme E1 subunit 1
Gene Aliases
A-116A10.1, APPBP1, HPP1, ula-1
UniGene
Hs.460978
Gene Chromosome Location
Chr.16: 66802878-66830977 on Build GRCh38
Transcripts
NM_001018159.1 NM_001018160.1 NM_001286500.1 NM_003905.3 XM_005256215.1 XM_011523422.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3937113 c.789G>C p.E263D Chr.16: 66816632-66816632 on Build GRCh38
7312266 c.819C>A p.N273K Chr.16: 66816602-66816602 on Build GRCh38
972223 c.768T>C p.N256N Chr.16: 66816653-66816653 on Build GRCh38
2993783 c.761delA p.N254fs*19 Chr.16: 66816660-66816660 on Build GRCh38
124659 c.760A>T p.N254Y Chr.16: 66816661-66816661 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs955620179 Chr.16: 66816676-66816676 on Build GRCh38
rs987533725 Chr.16: 66816452-66816452 on Build GRCh38
rs1039348972 Chr.16: 66816290-66816290 on Build GRCh38
rs987438815 Chr.16: 66816188-66816188 on Build GRCh38
rs201813040 Chr.16: 66816637-66816637 on Build GRCh38
rs547813758 Chr.16: 66816425-66816425 on Build GRCh38
rs902237658 Chr.16: 66816355-66816355 on Build GRCh38
rs570034391 Chr.16: 66816209-66816209 on Build GRCh38
rs760747466 Chr.16: 66816583-66816583 on Build GRCh38
rs943356705 Chr.16: 66816575-66816575 on Build GRCh38
rs374399521 Chr.16: 66816412-66816412 on Build GRCh38
rs556560716 Chr.16: 66816522-66816522 on Build GRCh38
rs537114946 Chr.16: 66816506-66816506 on Build GRCh38
rs1017731900 Chr.16: 66816408-66816408 on Build GRCh38
rs1023632882 Chr.16: 66816405-66816405 on Build GRCh38
rs1044188291 Chr.16: 66816426-66816426 on Build GRCh38
rs1013198510 Chr.16: 66816385-66816385 on Build GRCh38
rs375619119 Chr.16: 66816568-66816568 on Build GRCh38
rs1051496670 Chr.16: 66816399-66816399 on Build GRCh38
rs1042122806 Chr.16: 66816354-66816354 on Build GRCh38
rs574481822 Chr.16: 66816477-66816477 on Build GRCh38
rs772588768 Chr.16: 66816571-66816571 on Build GRCh38
rs981508574 Chr.16: 66816294-66816294 on Build GRCh38
rs986833469 Chr.16: 66816677-66816677 on Build GRCh38
rs898356021 Chr.16: 66816415-66816415 on Build GRCh38
rs751496617 Chr.16: 66816665-66816665 on Build GRCh38
rs138888796 Chr.16: 66816411-66816411 on Build GRCh38
rs956926792 Chr.16: 66816322-66816322 on Build GRCh38
rs1023119463 Chr.16: 66816232-66816232 on Build GRCh38
rs749950514 Chr.16: 66816537-66816538 on Build GRCh38
rs762532232 Chr.16: 66816413-66816413 on Build GRCh38
rs950753544 Chr.16: 66816445-66816445 on Build GRCh38
rs774917716 Chr.16: 66816565-66816565 on Build GRCh38
rs908474179 Chr.16: 66816406-66816406 on Build GRCh38
rs1035289319 Chr.16: 66816307-66816307 on Build GRCh38
rs185259091 Chr.16: 66816189-66816189 on Build GRCh38
rs189537573 Chr.16: 66816384-66816384 on Build GRCh38
rs974726699 Chr.16: 66816647-66816647 on Build GRCh38
rs971128719 Chr.16: 66816269-66816269 on Build GRCh38
rs750822466 Chr.16: 66816430-66816430 on Build GRCh38
rs766527852 Chr.16: 66816598-66816598 on Build GRCh38
rs988279346 Chr.16: 66816330-66816330 on Build GRCh38
rs773235354 Chr.16: 66816577-66816577 on Build GRCh38
rs893673979 Chr.16: 66816314-66816314 on Build GRCh38
rs538955961 Chr.16: 66816356-66816356 on Build GRCh38
rs758796257 Chr.16: 66816534-66816534 on Build GRCh38
rs565891714 Chr.16: 66816409-66816409 on Build GRCh38
rs763831583 Chr.16: 66816653-66816653 on Build GRCh38
rs946538653 Chr.16: 66816318-66816318 on Build GRCh38
rs929297486 Chr.16: 66816163-66816163 on Build GRCh38
rs762800147 Chr.16: 66816638-66816638 on Build GRCh38
rs534255692 Chr.16: 66816375-66816375 on Build GRCh38
rs943322213 Chr.16: 66816206-66816206 on Build GRCh38
rs768960066 Chr.16: 66816543-66816543 on Build GRCh38
rs200655956 Chr.16: 66816614-66816614 on Build GRCh38
rs1022500272 Chr.16: 66816562-66816562 on Build GRCh38
rs915437978 Chr.16: 66816344-66816344 on Build GRCh38
rs1012701845 Chr.16: 66816201-66816201 on Build GRCh38
rs776894137 Chr.16: 66816635-66816635 on Build GRCh38

View Full Product Details