Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 69863868-69864456 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CD226
Entrez Gene ID
10666
Gene Name
CD226 molecule
Gene Aliases
DNAM-1, DNAM1, PTA1, TLiSA1
UniGene
Hs.660130
Gene Chromosome Location
Chr.18: 69860228-69962086 on Build GRCh38
Transcripts
NM_001303618.1 NM_001303619.1 NM_006566.3 XM_005266642.3 XM_005266643.3 XM_006722374.3 XM_017025524.1 XM_017025525.1 XM_017025526.1 XM_017025527.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4644150 c.997A>C p.K333Q Chr.18: 69864328-69864328 on Build GRCh38
989715 c.957G>A p.E319E Chr.18: 69864368-69864368 on Build GRCh38
6623591 c.973T>C p.Y325H Chr.18: 69864352-69864352 on Build GRCh38
6038050 c.934C>T p.Q312* Chr.18: 69864391-69864391 on Build GRCh38
3527352 c.908C>T p.P303L Chr.18: 69864417-69864417 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752790330 Chr.18: 69864328-69864328 on Build GRCh38
rs909119943 Chr.18: 69863935-69863935 on Build GRCh38
rs897263728 Chr.18: 69863963-69863963 on Build GRCh38
rs200072703 Chr.18: 69864272-69864272 on Build GRCh38
rs558593252 Chr.18: 69864393-69864393 on Build GRCh38
rs751135209 Chr.18: 69864289-69864289 on Build GRCh38
rs765664676 Chr.18: 69864407-69864407 on Build GRCh38
rs1023921347 Chr.18: 69864035-69864035 on Build GRCh38
rs752902848 Chr.18: 69863933-69863933 on Build GRCh38
rs761550474 Chr.18: 69864396-69864396 on Build GRCh38
rs758955798 Chr.18: 69864432-69864432 on Build GRCh38
rs757450081 Chr.18: 69864191-69864191 on Build GRCh38
rs931901690 Chr.18: 69863898-69863898 on Build GRCh38
rs1030650239 Chr.18: 69864093-69864093 on Build GRCh38
rs141282007 Chr.18: 69864337-69864337 on Build GRCh38
rs563225231 Chr.18: 69864188-69864188 on Build GRCh38
rs1056589871 Chr.18: 69864334-69864334 on Build GRCh38
rs968690619 Chr.18: 69864064-69864064 on Build GRCh38
rs183726227 Chr.18: 69864383-69864383 on Build GRCh38
rs994182263 Chr.18: 69864004-69864004 on Build GRCh38
rs948325607 Chr.18: 69864177-69864177 on Build GRCh38
rs79913944 Chr.18: 69863922-69863922 on Build GRCh38
rs748271269 Chr.18: 69864299-69864299 on Build GRCh38
rs796640397 Chr.18: 69864226-69864226 on Build GRCh38
rs776426087 Chr.18: 69864267-69864267 on Build GRCh38
rs199638156 Chr.18: 69864281-69864281 on Build GRCh38
rs762422878 Chr.18: 69864279-69864279 on Build GRCh38
rs923102763 Chr.18: 69864123-69864123 on Build GRCh38
rs530682432 Chr.18: 69864202-69864202 on Build GRCh38
rs540725482 Chr.18: 69864023-69864023 on Build GRCh38
rs892572451 Chr.18: 69864245-69864245 on Build GRCh38
rs757477134 Chr.18: 69864348-69864348 on Build GRCh38
rs576874246 Chr.18: 69863997-69863997 on Build GRCh38
rs1010094418 Chr.18: 69863913-69863913 on Build GRCh38
rs755524346 Chr.18: 69864301-69864301 on Build GRCh38
rs763361 Chr.18: 69864406-69864406 on Build GRCh38
rs558557634 Chr.18: 69863930-69863930 on Build GRCh38
rs1025856718 Chr.18: 69864023-69864025 on Build GRCh38
rs138476878 Chr.18: 69864292-69864292 on Build GRCh38
rs780961348 Chr.18: 69864351-69864351 on Build GRCh38
rs754111980 Chr.18: 69863956-69863956 on Build GRCh38
rs896613059 Chr.18: 69864011-69864011 on Build GRCh38
rs1006787354 Chr.18: 69863974-69863974 on Build GRCh38
rs1000555224 Chr.18: 69864083-69864083 on Build GRCh38
rs190695222 Chr.18: 69864319-69864319 on Build GRCh38
rs753094424 Chr.18: 69864423-69864423 on Build GRCh38
rs1017944165 Chr.18: 69863994-69863994 on Build GRCh38
rs186207803 Chr.18: 69864265-69864265 on Build GRCh38
rs549767892 Chr.18: 69864242-69864242 on Build GRCh38
rs1012825028 Chr.18: 69864022-69864022 on Build GRCh38
rs928228842 Chr.18: 69864217-69864217 on Build GRCh38
rs984283339 Chr.18: 69864073-69864073 on Build GRCh38
rs574543275 Chr.18: 69864119-69864119 on Build GRCh38
rs769700135 Chr.18: 69864375-69864375 on Build GRCh38
rs1045621268 Chr.18: 69864367-69864367 on Build GRCh38
rs555283543 Chr.18: 69863914-69863914 on Build GRCh38
rs146733994 Chr.18: 69864336-69864336 on Build GRCh38
rs941400957 Chr.18: 69863936-69863936 on Build GRCh38
rs749282215 Chr.18: 69864386-69864386 on Build GRCh38
rs57613010 Chr.18: 69864269-69864269 on Build GRCh38
rs768005426 Chr.18: 69864389-69864389 on Build GRCh38
rs1005237225 Chr.18: 69864385-69864385 on Build GRCh38
rs747285269 Chr.18: 69864263-69864263 on Build GRCh38
rs542135604 Chr.18: 69864153-69864153 on Build GRCh38
rs200959714 Chr.18: 69864300-69864300 on Build GRCh38
rs767333276 Chr.18: 69864401-69864401 on Build GRCh38
rs763085746 Chr.18: 69864005-69864005 on Build GRCh38
rs201880773 Chr.18: 69864288-69864288 on Build GRCh38
rs766780605 Chr.18: 69864296-69864296 on Build GRCh38
rs956458266 Chr.18: 69864115-69864115 on Build GRCh38
rs576142392 Chr.18: 69863916-69863916 on Build GRCh38
rs909598621 Chr.18: 69864033-69864045 on Build GRCh38
rs559536978 Chr.18: 69864027-69864027 on Build GRCh38
rs745722083 Chr.18: 69864368-69864368 on Build GRCh38
rs953277385 Chr.18: 69864215-69864215 on Build GRCh38

View Full Product Details