Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 110173345-110173887 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM164
Entrez Gene ID
84187
Gene Name
transmembrane protein 164
Gene Aliases
bB360B22.3
UniGene
Hs.496572
Gene Chromosome Location
Chr.X: 110002413-110184438 on Build GRCh38
Transcripts
NM_017698.2 NM_032227.3 XM_005262205.4 XM_005262206.3 XM_005262208.4 XM_011531051.2 XM_011531054.1 XM_011531055.2 XM_017029894.1 XM_017029896.1 XM_017029897.1 XM_017029899.1 XM_017029900.1

SNP Detail

SNP ID Chromosome Location
rs765437500 Chr.X: 110173661-110173661 on Build GRCh38
rs377487696 Chr.X: 110173835-110173835 on Build GRCh38
rs766150763 Chr.X: 110173416-110173416 on Build GRCh38
rs746973766 Chr.X: 110173438-110173438 on Build GRCh38
rs878873454 Chr.X: 110173462-110173462 on Build GRCh38
rs775102460 Chr.X: 110173460-110173461 on Build GRCh38
rs931788181 Chr.X: 110173791-110173791 on Build GRCh38
rs775218538 Chr.X: 110173480-110173480 on Build GRCh38
rs550157052 Chr.X: 110173524-110173524 on Build GRCh38
rs34813944 Chr.X: 110173461-110173461 on Build GRCh38
rs184994773 Chr.X: 110173495-110173495 on Build GRCh38
rs922084117 Chr.X: 110173755-110173755 on Build GRCh38
rs753646007 Chr.X: 110173422-110173422 on Build GRCh38
rs757813829 Chr.X: 110173466-110173466 on Build GRCh38
rs751025142 Chr.X: 110173752-110173752 on Build GRCh38
rs769642174 Chr.X: 110173426-110173426 on Build GRCh38
rs893608233 Chr.X: 110173724-110173724 on Build GRCh38
rs397895707 Chr.X: 110173474-110173474 on Build GRCh38
rs935218036 Chr.X: 110173691-110173691 on Build GRCh38
rs373836050 Chr.X: 110173723-110173723 on Build GRCh38
rs1015477892 Chr.X: 110173425-110173425 on Build GRCh38
rs764192243 Chr.X: 110173367-110173367 on Build GRCh38
rs966243746 Chr.X: 110173750-110173750 on Build GRCh38
rs765031945 Chr.X: 110173449-110173449 on Build GRCh38
rs762074290 Chr.X: 110173394-110173394 on Build GRCh38
rs1054592013 Chr.X: 110173843-110173843 on Build GRCh38
rs199995701 Chr.X: 110173474-110173474 on Build GRCh38
rs763269134 Chr.X: 110173774-110173774 on Build GRCh38
rs374331197 Chr.X: 110173388-110173388 on Build GRCh38
rs1057115934 Chr.X: 110173817-110173817 on Build GRCh38
rs896083006 Chr.X: 110173842-110173842 on Build GRCh38
rs905897552 Chr.X: 110173778-110173778 on Build GRCh38
rs754809574 Chr.X: 110173424-110173424 on Build GRCh38
rs908142320 Chr.X: 110173743-110173743 on Build GRCh38
rs987385542 Chr.X: 110173686-110173686 on Build GRCh38
rs911810640 Chr.X: 110173689-110173689 on Build GRCh38
rs1002681504 Chr.X: 110173456-110173456 on Build GRCh38
rs921644289 Chr.X: 110173627-110173627 on Build GRCh38
rs892788890 Chr.X: 110173382-110173382 on Build GRCh38
rs952128965 Chr.X: 110173662-110173662 on Build GRCh38
rs774485434 Chr.X: 110173461-110173463 on Build GRCh38
rs200859909 Chr.X: 110173494-110173494 on Build GRCh38
rs60563871 Chr.X: 110173609-110173609 on Build GRCh38
rs752706773 Chr.X: 110173461-110173461 on Build GRCh38
rs745550955 Chr.X: 110173460-110173460 on Build GRCh38
rs762808338 Chr.X: 110173461-110173462 on Build GRCh38

View Full Product Details