Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 8381491-8382011 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACOX3
Entrez Gene ID
8310
Gene Name
acyl-CoA oxidase 3, pristanoyl
Gene Aliases
-
UniGene
Hs.479122
Gene Chromosome Location
Chr.4: 8366263-8440725 on Build GRCh38
Transcripts
NM_001101667.1 NM_003501.2 XM_005248011.4 XM_005248013.3 XM_011513565.2 XM_011513566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4003173 c.1565T>G p.V522G Chr.4: 8381580-8381580 on Build GRCh38
1058212 c.1540G>A p.A514T Chr.4: 8381605-8381605 on Build GRCh38
7086251 c.1596A>G p.K532K Chr.4: 8381549-8381549 on Build GRCh38
3606565 c.1582G>A p.E528K Chr.4: 8381563-8381563 on Build GRCh38
3918259 c.1578C>T p.L526L Chr.4: 8381567-8381567 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs993432393 Chr.4: 8381727-8381727 on Build GRCh38
rs944681357 Chr.4: 8381852-8381852 on Build GRCh38
rs1005700727 Chr.4: 8381704-8381704 on Build GRCh38
rs148853379 Chr.4: 8381565-8381565 on Build GRCh38
rs139566884 Chr.4: 8381934-8381934 on Build GRCh38
rs957217561 Chr.4: 8381779-8381779 on Build GRCh38
rs1040656956 Chr.4: 8381855-8381855 on Build GRCh38
rs773903597 Chr.4: 8381648-8381648 on Build GRCh38
rs575246234 Chr.4: 8381946-8381946 on Build GRCh38
rs775136434 Chr.4: 8381629-8381629 on Build GRCh38
rs756019084 Chr.4: 8381602-8381602 on Build GRCh38
rs368350504 Chr.4: 8381653-8381653 on Build GRCh38
rs772509801 Chr.4: 8381570-8381570 on Build GRCh38
rs1016086957 Chr.4: 8381720-8381720 on Build GRCh38
rs146945288 Chr.4: 8381566-8381566 on Build GRCh38
rs557389295 Chr.4: 8381828-8381828 on Build GRCh38
rs763930986 Chr.4: 8381764-8381764 on Build GRCh38
rs553761531 Chr.4: 8381929-8381929 on Build GRCh38
rs113403746 Chr.4: 8381921-8381921 on Build GRCh38
rs768158127 Chr.4: 8381646-8381646 on Build GRCh38
rs926008639 Chr.4: 8381770-8381770 on Build GRCh38
rs1004619986 Chr.4: 8381895-8381895 on Build GRCh38
rs750396981 Chr.4: 8381588-8381588 on Build GRCh38
rs771555097 Chr.4: 8381626-8381626 on Build GRCh38
rs762728379 Chr.4: 8381576-8381576 on Build GRCh38
rs370052523 Chr.4: 8381625-8381625 on Build GRCh38
rs534898037 Chr.4: 8381896-8381896 on Build GRCh38
rs569758144 Chr.4: 8381840-8381840 on Build GRCh38
rs761225357 Chr.4: 8381574-8381574 on Build GRCh38
rs780165236 Chr.4: 8381605-8381605 on Build GRCh38
rs751396026 Chr.4: 8381806-8381806 on Build GRCh38
rs988635735 Chr.4: 8381787-8381787 on Build GRCh38
rs532868233 Chr.4: 8381644-8381644 on Build GRCh38
rs772897612 Chr.4: 8381616-8381616 on Build GRCh38
rs748355766 Chr.4: 8381613-8381613 on Build GRCh38
rs980163466 Chr.4: 8381766-8381766 on Build GRCh38
rs376449505 Chr.4: 8381612-8381612 on Build GRCh38
rs373553449 Chr.4: 8381606-8381606 on Build GRCh38
rs750309034 Chr.4: 8381524-8381524 on Build GRCh38
rs372321417 Chr.4: 8381963-8381963 on Build GRCh38
rs995929890 Chr.4: 8381663-8381663 on Build GRCh38
rs752586271 Chr.4: 8381549-8381549 on Build GRCh38
rs773880193 Chr.4: 8381573-8381573 on Build GRCh38
rs780315226 Chr.4: 8381891-8381891 on Build GRCh38
rs755016911 Chr.4: 8381611-8381611 on Build GRCh38
rs752208140 Chr.4: 8381872-8381872 on Build GRCh38
rs767257323 Chr.4: 8381587-8381587 on Build GRCh38
rs888630248 Chr.4: 8381701-8381701 on Build GRCh38
rs969990642 Chr.4: 8381742-8381742 on Build GRCh38
rs993295064 Chr.4: 8381893-8381893 on Build GRCh38
rs1014911931 Chr.4: 8381730-8381730 on Build GRCh38
rs763653116 Chr.4: 8381577-8381577 on Build GRCh38
rs17806883 Chr.4: 8381887-8381887 on Build GRCh38
rs900166386 Chr.4: 8381636-8381636 on Build GRCh38
rs1053304556 Chr.4: 8381824-8381829 on Build GRCh38
rs780073703 Chr.4: 8381553-8381553 on Build GRCh38
rs988933000 Chr.4: 8381945-8381945 on Build GRCh38
rs751185160 Chr.4: 8381723-8381723 on Build GRCh38
rs373788067 Chr.4: 8381857-8381857 on Build GRCh38
rs950313283 Chr.4: 8381955-8381955 on Build GRCh38
rs749573613 Chr.4: 8381555-8381555 on Build GRCh38
rs200650508 Chr.4: 8381580-8381580 on Build GRCh38
rs756214944 Chr.4: 8381532-8381532 on Build GRCh38
rs777837184 Chr.4: 8381632-8381632 on Build GRCh38
rs574347357 Chr.4: 8381920-8381920 on Build GRCh38
rs926138189 Chr.4: 8381800-8381800 on Build GRCh38
rs111879117 Chr.4: 8381950-8381950 on Build GRCh38
rs374314740 Chr.4: 8381739-8381739 on Build GRCh38
rs765155172 Chr.4: 8381582-8381582 on Build GRCh38
rs145548429 Chr.4: 8381559-8381559 on Build GRCh38
rs116834870 Chr.4: 8381938-8381938 on Build GRCh38
rs887549892 Chr.4: 8381892-8381892 on Build GRCh38
rs553032282 Chr.4: 8381899-8381899 on Build GRCh38
rs558278672 Chr.4: 8381722-8381722 on Build GRCh38
rs755974858 Chr.4: 8381624-8381626 on Build GRCh38
rs867879967 Chr.4: 8381915-8381915 on Build GRCh38
rs748465089 Chr.4: 8381568-8381568 on Build GRCh38
rs778253388 Chr.4: 8381618-8381618 on Build GRCh38
rs558779534 Chr.4: 8381886-8381886 on Build GRCh38
rs964472345 Chr.4: 8381771-8381771 on Build GRCh38
rs905940987 Chr.4: 8381665-8381665 on Build GRCh38
rs1032963184 Chr.4: 8381959-8381959 on Build GRCh38
rs964700560 Chr.4: 8381935-8381935 on Build GRCh38
rs754534168 Chr.4: 8381799-8381799 on Build GRCh38
rs989301530 Chr.4: 8381978-8381978 on Build GRCh38
rs149706675 Chr.4: 8381966-8381966 on Build GRCh38
rs184901635 Chr.4: 8381699-8381699 on Build GRCh38

View Full Product Details