Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 144591283-144591751 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TPK1
Entrez Gene ID
27010
Gene Name
thiamin pyrophosphokinase 1
Gene Aliases
HTPK1, PP20, THMD5
UniGene
Hs.660232
Gene Chromosome Location
Chr.7: 144451941-144836424 on Build GRCh38
Transcripts
NM_001042482.1 NM_022445.3 XM_005249970.1 XM_011516031.1 XM_011516032.2 XM_011516033.2 XM_011516034.2 XM_011516035.2 XM_011516037.2 XM_011516039.2 XM_011516040.2 XM_011516041.2 XM_011516043.1 XM_011516044.1 XM_011516047.2 XM_011516048.1 XM_011516050.1 XM_017011969.1 XM_017011970.1 XM_017011971.1 XM_017011972.1 XM_017011973.1 XM_017011974.1 XM_017011975.1 XM_017011976.1 XM_017011977.1 XM_017011978.1 XM_017011979.1 XM_017011980.1 XM_017011981.1 XM_017011982.1 XM_017011983.1 XM_017011984.1 XM_017011985.1 XM_017011986.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4587222 c.83C>T p.A28V Chr.7: 144591490-144591490 on Build GRCh38
4490642 c.366C>T p.I122I Chr.7: 144591558-144591558 on Build GRCh38
3259244 c.498A>T p.Q166H Chr.7: 144591426-144591426 on Build GRCh38
4490643 c.15C>T p.I5I Chr.7: 144591558-144591558 on Build GRCh38
4153941 c.69T>A p.N23K Chr.7: 144591504-144591504 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs746578228 Chr.7: 144591479-144591479 on Build GRCh38
rs955870525 Chr.7: 144591363-144591363 on Build GRCh38
rs369707518 Chr.7: 144591519-144591519 on Build GRCh38
rs760881424 Chr.7: 144591542-144591542 on Build GRCh38
rs755359509 Chr.7: 144591442-144591442 on Build GRCh38
rs199657217 Chr.7: 144591409-144591409 on Build GRCh38
rs775364062 Chr.7: 144591513-144591513 on Build GRCh38
rs863224236 Chr.7: 144591558-144591558 on Build GRCh38
rs145770279 Chr.7: 144591675-144591675 on Build GRCh38
rs762729038 Chr.7: 144591520-144591520 on Build GRCh38
rs184792676 Chr.7: 144591370-144591370 on Build GRCh38
rs201237707 Chr.7: 144591426-144591426 on Build GRCh38
rs753088448 Chr.7: 144591444-144591444 on Build GRCh38
rs373811956 Chr.7: 144591320-144591320 on Build GRCh38
rs551242702 Chr.7: 144591658-144591658 on Build GRCh38
rs766748807 Chr.7: 144591402-144591402 on Build GRCh38
rs1026552624 Chr.7: 144591318-144591318 on Build GRCh38
rs561666770 Chr.7: 144591607-144591607 on Build GRCh38
rs149791801 Chr.7: 144591611-144591611 on Build GRCh38
rs147602742 Chr.7: 144591532-144591532 on Build GRCh38
rs529567663 Chr.7: 144591627-144591627 on Build GRCh38
rs866844744 Chr.7: 144591455-144591455 on Build GRCh38
rs373553264 Chr.7: 144591584-144591584 on Build GRCh38
rs376230688 Chr.7: 144591389-144591389 on Build GRCh38
rs759374079 Chr.7: 144591400-144591400 on Build GRCh38
rs374946048 Chr.7: 144591433-144591433 on Build GRCh38
rs763870331 Chr.7: 144591549-144591549 on Build GRCh38
rs776395743 Chr.7: 144591399-144591399 on Build GRCh38
rs751036370 Chr.7: 144591592-144591592 on Build GRCh38
rs759501604 Chr.7: 144591664-144591664 on Build GRCh38
rs1037780458 Chr.7: 144591436-144591436 on Build GRCh38
rs1009112016 Chr.7: 144591605-144591605 on Build GRCh38
rs568693004 Chr.7: 144591573-144591573 on Build GRCh38
rs79334964 Chr.7: 144591327-144591327 on Build GRCh38
rs376340734 Chr.7: 144591619-144591620 on Build GRCh38
rs777144353 Chr.7: 144591475-144591475 on Build GRCh38
rs1026449918 Chr.7: 144591649-144591649 on Build GRCh38
rs189256243 Chr.7: 144591695-144591695 on Build GRCh38
rs528445678 Chr.7: 144591414-144591414 on Build GRCh38
rs770545870 Chr.7: 144591489-144591489 on Build GRCh38
rs374926521 Chr.7: 144591593-144591593 on Build GRCh38
rs547285921 Chr.7: 144591559-144591559 on Build GRCh38
rs978500927 Chr.7: 144591343-144591343 on Build GRCh38
rs748908705 Chr.7: 144591379-144591379 on Build GRCh38
rs745461624 Chr.7: 144591603-144591603 on Build GRCh38
rs1008995175 Chr.7: 144591326-144591326 on Build GRCh38
rs899357375 Chr.7: 144591438-144591438 on Build GRCh38
rs993782095 Chr.7: 144591303-144591303 on Build GRCh38
rs191958558 Chr.7: 144591306-144591306 on Build GRCh38
rs747501466 Chr.7: 144591613-144591614 on Build GRCh38
rs964821209 Chr.7: 144591333-144591333 on Build GRCh38
rs779361801 Chr.7: 144591443-144591443 on Build GRCh38
rs375169579 Chr.7: 144591419-144591419 on Build GRCh38
rs754068899 Chr.7: 144591553-144591553 on Build GRCh38
rs780701936 Chr.7: 144591601-144591601 on Build GRCh38
rs576168961 Chr.7: 144591490-144591490 on Build GRCh38
rs768703805 Chr.7: 144591525-144591525 on Build GRCh38
rs1032842286 Chr.7: 144591346-144591346 on Build GRCh38
rs993693814 Chr.7: 144591639-144591639 on Build GRCh38
rs769525399 Chr.7: 144591498-144591498 on Build GRCh38
rs753174543 Chr.7: 144591579-144591579 on Build GRCh38
rs758819848 Chr.7: 144591583-144591583 on Build GRCh38
rs756715782 Chr.7: 144591595-144591595 on Build GRCh38
rs557906339 Chr.7: 144591441-144591441 on Build GRCh38
rs368326695 Chr.7: 144591404-144591404 on Build GRCh38
rs777357013 Chr.7: 144591381-144591381 on Build GRCh38
rs917150305 Chr.7: 144591393-144591393 on Build GRCh38
rs372057511 Chr.7: 144591307-144591307 on Build GRCh38
rs758949475 Chr.7: 144591445-144591445 on Build GRCh38
rs770639423 Chr.7: 144591390-144591390 on Build GRCh38

View Full Product Details