Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 163143397-163143957 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RGS5
Entrez Gene ID
8490
Gene Name
regulator of G-protein signaling 5
Gene Aliases
MST092, MST106, MST129, MSTP032, MSTP092, MSTP106, MSTP129
UniGene
Hs.24950
Gene Chromosome Location
Chr.1: 163142299-163321791 on Build GRCh38
Transcripts
NM_001195303.2 NM_001254748.1 NM_001254749.1 NM_003617.3

SNP Detail

SNP ID Chromosome Location
rs1029556199 Chr.1: 163143688-163143688 on Build GRCh38
rs908254756 Chr.1: 163143865-163143865 on Build GRCh38
rs955193704 Chr.1: 163143693-163143693 on Build GRCh38
rs892310764 Chr.1: 163143911-163143911 on Build GRCh38
rs200530428 Chr.1: 163143762-163143762 on Build GRCh38
rs566167494 Chr.1: 163143890-163143890 on Build GRCh38
rs941117222 Chr.1: 163143871-163143871 on Build GRCh38
rs968489932 Chr.1: 163143419-163143419 on Build GRCh38
rs1056515 Chr.1: 163143470-163143470 on Build GRCh38
rs982969140 Chr.1: 163143864-163143864 on Build GRCh38
rs975521682 Chr.1: 163143461-163143461 on Build GRCh38
rs747498397 Chr.1: 163143821-163143821 on Build GRCh38
rs917533947 Chr.1: 163143843-163143843 on Build GRCh38
rs778134817 Chr.1: 163143771-163143771 on Build GRCh38
rs937596628 Chr.1: 163143482-163143482 on Build GRCh38
rs548657331 Chr.1: 163143716-163143716 on Build GRCh38
rs370990008 Chr.1: 163143751-163143752 on Build GRCh38
rs2661280 Chr.1: 163143585-163143585 on Build GRCh38
rs997162627 Chr.1: 163143680-163143680 on Build GRCh38
rs920271537 Chr.1: 163143720-163143720 on Build GRCh38
rs184131275 Chr.1: 163143912-163143912 on Build GRCh38
rs968059710 Chr.1: 163143928-163143928 on Build GRCh38
rs138471500 Chr.1: 163143593-163143593 on Build GRCh38
rs1056514 Chr.1: 163143776-163143776 on Build GRCh38
rs932362847 Chr.1: 163143927-163143927 on Build GRCh38
rs535298991 Chr.1: 163143647-163143647 on Build GRCh38
rs551708211 Chr.1: 163143486-163143486 on Build GRCh38
rs758562408 Chr.1: 163143754-163143754 on Build GRCh38
rs56212137 Chr.1: 163143855-163143855 on Build GRCh38
rs549825937 Chr.1: 163143600-163143600 on Build GRCh38
rs534422215 Chr.1: 163143752-163143752 on Build GRCh38
rs574276489 Chr.1: 163143932-163143932 on Build GRCh38
rs950221804 Chr.1: 163143599-163143599 on Build GRCh38
rs191604587 Chr.1: 163143745-163143745 on Build GRCh38
rs886870978 Chr.1: 163143639-163143639 on Build GRCh38
rs1038449084 Chr.1: 163143669-163143669 on Build GRCh38
rs776986534 Chr.1: 163143921-163143921 on Build GRCh38
rs899715964 Chr.1: 163143678-163143678 on Build GRCh38
rs958774277 Chr.1: 163143798-163143798 on Build GRCh38
rs1056083141 Chr.1: 163143516-163143516 on Build GRCh38
rs569489018 Chr.1: 163143632-163143632 on Build GRCh38
rs756990651 Chr.1: 163143456-163143456 on Build GRCh38

View Full Product Details