Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 229512522-229512989 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNER
Entrez Gene ID
92737
Gene Name
delta/notch like EGF repeat containing
Gene Aliases
UNQ26, bet
UniGene
Hs.234074
Gene Chromosome Location
Chr.2: 229357629-229714570 on Build GRCh38
Transcripts
NM_139072.3 XM_005246950.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7238565 c.1117G>T p.G373W Chr.2: 229512813-229512813 on Build GRCh38
394261 c.1061_1062AG>TT p.Q354>? Chr.2: 229512868-229512869 on Build GRCh38
6446599 c.1105G>A p.E369K Chr.2: 229512825-229512825 on Build GRCh38
1017515 c.1097A>T p.D366V Chr.2: 229512833-229512833 on Build GRCh38
3047974 c.1089C>T p.S363S Chr.2: 229512841-229512841 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs564134662 Chr.2: 229512953-229512953 on Build GRCh38
rs868522163 Chr.2: 229512799-229512799 on Build GRCh38
rs747712827 Chr.2: 229512945-229512946 on Build GRCh38
rs1008074479 Chr.2: 229512567-229512567 on Build GRCh38
rs184021791 Chr.2: 229512565-229512565 on Build GRCh38
rs149897676 Chr.2: 229512912-229512912 on Build GRCh38
rs550059066 Chr.2: 229512556-229512556 on Build GRCh38
rs774258388 Chr.2: 229512938-229512938 on Build GRCh38
rs201917911 Chr.2: 229512931-229512931 on Build GRCh38
rs938362517 Chr.2: 229512882-229512882 on Build GRCh38
rs755510223 Chr.2: 229512814-229512814 on Build GRCh38
rs541493138 Chr.2: 229512877-229512877 on Build GRCh38
rs948765993 Chr.2: 229512693-229512693 on Build GRCh38
rs778185598 Chr.2: 229512899-229512899 on Build GRCh38
rs994382228 Chr.2: 229512830-229512830 on Build GRCh38
rs536619598 Chr.2: 229512571-229512571 on Build GRCh38
rs753250384 Chr.2: 229512890-229512890 on Build GRCh38
rs947087705 Chr.2: 229512745-229512745 on Build GRCh38
rs779246523 Chr.2: 229512884-229512884 on Build GRCh38
rs1041832492 Chr.2: 229512616-229512616 on Build GRCh38
rs981489988 Chr.2: 229512700-229512700 on Build GRCh38
rs755132810 Chr.2: 229512880-229512880 on Build GRCh38
rs771603023 Chr.2: 229512743-229512743 on Build GRCh38
rs918105294 Chr.2: 229512783-229512783 on Build GRCh38
rs772516631 Chr.2: 229512820-229512820 on Build GRCh38
rs1019496082 Chr.2: 229512589-229512589 on Build GRCh38
rs1007500039 Chr.2: 229512821-229512821 on Build GRCh38
rs928841886 Chr.2: 229512706-229512706 on Build GRCh38
rs11461195 Chr.2: 229512659-229512660 on Build GRCh38
rs556414531 Chr.2: 229512844-229512844 on Build GRCh38
rs758631766 Chr.2: 229512898-229512898 on Build GRCh38
rs1028093300 Chr.2: 229512550-229512550 on Build GRCh38
rs143345995 Chr.2: 229512852-229512852 on Build GRCh38
rs373389203 Chr.2: 229512904-229512904 on Build GRCh38
rs187948575 Chr.2: 229512689-229512689 on Build GRCh38
rs199643608 Chr.2: 229512826-229512826 on Build GRCh38
rs1037541834 Chr.2: 229512740-229512740 on Build GRCh38
rs745330329 Chr.2: 229512924-229512924 on Build GRCh38
rs200458484 Chr.2: 229512747-229512747 on Build GRCh38
rs748588837 Chr.2: 229512741-229512741 on Build GRCh38
rs953900225 Chr.2: 229512562-229512563 on Build GRCh38
rs752401467 Chr.2: 229512766-229512766 on Build GRCh38
rs369585918 Chr.2: 229512845-229512845 on Build GRCh38
rs751461005 Chr.2: 229512869-229512869 on Build GRCh38
rs972671611 Chr.2: 229512630-229512630 on Build GRCh38
rs758260102 Chr.2: 229512767-229512767 on Build GRCh38
rs768547006 Chr.2: 229512936-229512936 on Build GRCh38
rs776492748 Chr.2: 229512752-229512752 on Build GRCh38
rs146813937 Chr.2: 229512879-229512879 on Build GRCh38
rs938310267 Chr.2: 229512654-229512654 on Build GRCh38
rs919817417 Chr.2: 229512647-229512647 on Build GRCh38
rs952097263 Chr.2: 229512669-229512669 on Build GRCh38
rs749446179 Chr.2: 229512808-229512808 on Build GRCh38
rs369163042 Chr.2: 229512779-229512779 on Build GRCh38
rs749171981 Chr.2: 229512859-229512859 on Build GRCh38
rs897619084 Chr.2: 229512742-229512742 on Build GRCh38
rs780271188 Chr.2: 229512797-229512797 on Build GRCh38
rs945826406 Chr.2: 229512597-229512597 on Build GRCh38
rs6717730 Chr.2: 229512676-229512676 on Build GRCh38
rs779524622 Chr.2: 229512819-229512819 on Build GRCh38
rs759453658 Chr.2: 229512760-229512760 on Build GRCh38
rs775531552 Chr.2: 229512847-229512847 on Build GRCh38
rs769354107 Chr.2: 229512929-229512929 on Build GRCh38
rs1006098172 Chr.2: 229512728-229512728 on Build GRCh38
rs760136535 Chr.2: 229512746-229512746 on Build GRCh38
rs934817561 Chr.2: 229512726-229512726 on Build GRCh38
rs991151351 Chr.2: 229512888-229512888 on Build GRCh38
rs373356577 Chr.2: 229512837-229512837 on Build GRCh38
rs765060539 Chr.2: 229512846-229512846 on Build GRCh38
rs762591673 Chr.2: 229512848-229512848 on Build GRCh38
rs772852791 Chr.2: 229512962-229512963 on Build GRCh38
rs376161329 Chr.2: 229512842-229512842 on Build GRCh38
rs780869420 Chr.2: 229512917-229512917 on Build GRCh38
rs765281747 Chr.2: 229512765-229512765 on Build GRCh38
rs769536132 Chr.2: 229512946-229512946 on Build GRCh38
rs192517100 Chr.2: 229512691-229512691 on Build GRCh38
rs763967495 Chr.2: 229512860-229512860 on Build GRCh38
rs928340081 Chr.2: 229512641-229512641 on Build GRCh38
rs527439071 Chr.2: 229512903-229512903 on Build GRCh38
rs746678179 Chr.2: 229512827-229512827 on Build GRCh38
rs576984523 Chr.2: 229512785-229512785 on Build GRCh38
rs1037992888 Chr.2: 229512958-229512958 on Build GRCh38
rs767598147 Chr.2: 229512777-229512777 on Build GRCh38

View Full Product Details