Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 33432625-33433219 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNGR2
Entrez Gene ID
3460
Gene Name
interferon gamma receptor 2 (interferon gamma transducer 1)
Gene Aliases
AF-1, IFGR2, IFNGT1, IMD28
UniGene
Hs.634632
Gene Chromosome Location
Chr.21: 33402896-33437521 on Build GRCh38
Transcripts
NM_005534.3 XM_005260969.2 XM_011529553.1 XM_011529554.2 NR_040016.1 XM_011529746.1

Gene Symbol
TMEM50B
Entrez Gene ID
757
Gene Name
transmembrane protein 50B
Gene Aliases
C21orf4, HCVP7TP3
UniGene
Hs.433668
Gene Chromosome Location
Chr.21: 33432486-33480009 on Build GRCh38
Transcripts
NM_005534.3 XM_005260969.2 XM_011529553.1 XM_011529554.2 NR_040016.1 XM_011529746.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1413855 c.770C>T p.S257L Chr.21: 33432762-33432762 on Build GRCh38
6683927 c.835T>G p.Y279D Chr.21: 33432827-33432827 on Build GRCh38
419255 c.804C>A p.F268L Chr.21: 33432796-33432796 on Build GRCh38
2843619 c.870G>C p.Q290H Chr.21: 33432862-33432862 on Build GRCh38
1030288 c.874G>A p.E292K Chr.21: 33432866-33432866 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767166047 Chr.21: 33433121-33433122 on Build GRCh38
rs772797835 Chr.21: 33432684-33432684 on Build GRCh38
rs781176667 Chr.21: 33432784-33432784 on Build GRCh38
rs375538161 Chr.21: 33432876-33432876 on Build GRCh38
rs1532 Chr.21: 33432659-33432659 on Build GRCh38
rs1023877064 Chr.21: 33432957-33432957 on Build GRCh38
rs767753849 Chr.21: 33432967-33432967 on Build GRCh38
rs769561382 Chr.21: 33432841-33432841 on Build GRCh38
rs747344228 Chr.21: 33432891-33432892 on Build GRCh38
rs116536727 Chr.21: 33432771-33432771 on Build GRCh38
rs1035633808 Chr.21: 33432988-33432988 on Build GRCh38
rs762681401 Chr.21: 33432697-33432697 on Build GRCh38
rs566154275 Chr.21: 33432787-33432787 on Build GRCh38
rs1004955573 Chr.21: 33432811-33432811 on Build GRCh38
rs1036419209 Chr.21: 33433184-33433184 on Build GRCh38
rs750573735 Chr.21: 33433099-33433099 on Build GRCh38
rs747852852 Chr.21: 33432889-33432892 on Build GRCh38
rs568350481 Chr.21: 33432998-33432998 on Build GRCh38
rs796136357 Chr.21: 33432891-33432892 on Build GRCh38
rs368644044 Chr.21: 33433117-33433117 on Build GRCh38
rs780365785 Chr.21: 33432911-33432911 on Build GRCh38
rs753453319 Chr.21: 33432889-33432891 on Build GRCh38
rs200803530 Chr.21: 33432903-33432904 on Build GRCh38
rs765881972 Chr.21: 33432699-33432699 on Build GRCh38
rs955815573 Chr.21: 33433057-33433057 on Build GRCh38
rs780113680 Chr.21: 33432888-33432890 on Build GRCh38
rs761096880 Chr.21: 33432704-33432704 on Build GRCh38
rs121913216 Chr.21: 33432672-33432672 on Build GRCh38
rs750203174 Chr.21: 33432887-33432888 on Build GRCh38
rs939768829 Chr.21: 33432891-33432893 on Build GRCh38
rs947247157 Chr.21: 33432725-33432725 on Build GRCh38
rs758387117 Chr.21: 33432905-33432905 on Build GRCh38
rs576714760 Chr.21: 33432953-33432953 on Build GRCh38
rs937863192 Chr.21: 33432918-33432919 on Build GRCh38
rs982302588 Chr.21: 33432984-33432984 on Build GRCh38
rs890540411 Chr.21: 33432773-33432773 on Build GRCh38
rs769347613 Chr.21: 33432673-33432673 on Build GRCh38
rs762332617 Chr.21: 33432864-33432864 on Build GRCh38
rs912876600 Chr.21: 33433135-33433135 on Build GRCh38
rs765620198 Chr.21: 33432898-33432898 on Build GRCh38
rs957459108 Chr.21: 33433001-33433001 on Build GRCh38
rs373582182 Chr.21: 33432756-33432756 on Build GRCh38
rs978860725 Chr.21: 33433162-33433162 on Build GRCh38
rs903169657 Chr.21: 33432987-33432987 on Build GRCh38
rs535886101 Chr.21: 33432999-33432999 on Build GRCh38
rs577299511 Chr.21: 33433166-33433166 on Build GRCh38
rs761188502 Chr.21: 33432882-33432882 on Build GRCh38
rs546035322 Chr.21: 33433188-33433188 on Build GRCh38
rs1034039315 Chr.21: 33432707-33432709 on Build GRCh38
rs768605434 Chr.21: 33432670-33432670 on Build GRCh38
rs557489662 Chr.21: 33433026-33433026 on Build GRCh38
rs767366590 Chr.21: 33432886-33432887 on Build GRCh38
rs17883129 Chr.21: 33432890-33432890 on Build GRCh38
rs529890416 Chr.21: 33432650-33432650 on Build GRCh38
rs121913217 Chr.21: 33432772-33432772 on Build GRCh38
rs779060231 Chr.21: 33432757-33432757 on Build GRCh38
rs78311247 Chr.21: 33432903-33432903 on Build GRCh38
rs995022817 Chr.21: 33432917-33432917 on Build GRCh38
rs750400339 Chr.21: 33432762-33432762 on Build GRCh38
rs376105280 Chr.21: 33432706-33432706 on Build GRCh38
rs770663375 Chr.21: 33432875-33432875 on Build GRCh38
rs968384165 Chr.21: 33433152-33433152 on Build GRCh38
rs776462360 Chr.21: 33432698-33432698 on Build GRCh38
rs145216430 Chr.21: 33432716-33432716 on Build GRCh38
rs780332871 Chr.21: 33433187-33433187 on Build GRCh38
rs115458101 Chr.21: 33432763-33432763 on Build GRCh38
rs763026256 Chr.21: 33432885-33432889 on Build GRCh38
rs191792822 Chr.21: 33432749-33432749 on Build GRCh38
rs896429967 Chr.21: 33432932-33432932 on Build GRCh38
rs372109363 Chr.21: 33432886-33432886 on Build GRCh38
rs764187616 Chr.21: 33432909-33432910 on Build GRCh38
rs542070748 Chr.21: 33433177-33433177 on Build GRCh38
rs143025663 Chr.21: 33432861-33432861 on Build GRCh38
rs201322939 Chr.21: 33432888-33432889 on Build GRCh38
rs756332452 Chr.21: 33433181-33433181 on Build GRCh38
rs764540325 Chr.21: 33432891-33432891 on Build GRCh38
rs762385143 Chr.21: 33432895-33432895 on Build GRCh38
rs17885089 Chr.21: 33432677-33432677 on Build GRCh38
rs752966235 Chr.21: 33432707-33432708 on Build GRCh38
rs373845747 Chr.21: 33432892-33432892 on Build GRCh38
rs367731539 Chr.21: 33432908-33432908 on Build GRCh38
rs771477826 Chr.21: 33432891-33432891 on Build GRCh38
rs928385626 Chr.21: 33432660-33432660 on Build GRCh38
rs559219805 Chr.21: 33433087-33433087 on Build GRCh38
rs751639678 Chr.21: 33432790-33432790 on Build GRCh38
rs750536049 Chr.21: 33432900-33432900 on Build GRCh38
rs1033086578 Chr.21: 33432992-33432992 on Build GRCh38
rs187271417 Chr.21: 33432933-33432933 on Build GRCh38
rs186369364 Chr.21: 33432748-33432748 on Build GRCh38
rs751776635 Chr.21: 33432923-33432923 on Build GRCh38
rs1014842439 Chr.21: 33432820-33432820 on Build GRCh38
rs1045514352 Chr.21: 33433190-33433190 on Build GRCh38
rs763266586 Chr.21: 33432900-33432901 on Build GRCh38
rs1028456497 Chr.21: 33433032-33433032 on Build GRCh38
rs143248516 Chr.21: 33432890-33432891 on Build GRCh38
rs1013471054 Chr.21: 33432951-33432951 on Build GRCh38
rs193922682 Chr.21: 33432890-33432890 on Build GRCh38
rs921854002 Chr.21: 33433165-33433165 on Build GRCh38
rs1064579 Chr.21: 33432750-33432750 on Build GRCh38
rs998909098 Chr.21: 33432989-33432989 on Build GRCh38
rs756156431 Chr.21: 33432887-33432891 on Build GRCh38
rs1051596304 Chr.21: 33433173-33433173 on Build GRCh38

View Full Product Details