Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 88010880-88011479 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR65
Entrez Gene ID
8477
Gene Name
G protein-coupled receptor 65
Gene Aliases
TDAG8, hTDAG8
UniGene
Hs.443243
Gene Chromosome Location
Chr.14: 88005124-88014811 on Build GRCh38
Transcripts
NM_003608.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
305621 c.326C>T p.A109V Chr.14: 88011173-88011173 on Build GRCh38
6576306 c.563_572del10 p.C189fs*9 Chr.14: 88011410-88011419 on Build GRCh38
4554837 c.632G>T p.R211L Chr.14: 88011479-88011479 on Build GRCh38
3401494 c.328G>A p.V110I Chr.14: 88011175-88011175 on Build GRCh38
4769379 c.92C>A p.A31D Chr.14: 88010939-88010939 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771285188 Chr.14: 88011265-88011265 on Build GRCh38
rs983718463 Chr.14: 88010962-88010962 on Build GRCh38
rs746121339 Chr.14: 88011385-88011385 on Build GRCh38
rs142533891 Chr.14: 88011022-88011022 on Build GRCh38
rs148741118 Chr.14: 88011099-88011099 on Build GRCh38
rs147419350 Chr.14: 88011191-88011191 on Build GRCh38
rs370595097 Chr.14: 88011199-88011199 on Build GRCh38
rs949144996 Chr.14: 88011245-88011245 on Build GRCh38
rs776921083 Chr.14: 88010957-88010957 on Build GRCh38
rs891012357 Chr.14: 88011259-88011259 on Build GRCh38
rs772388191 Chr.14: 88011182-88011182 on Build GRCh38
rs541031027 Chr.14: 88011410-88011410 on Build GRCh38
rs754855859 Chr.14: 88011058-88011058 on Build GRCh38
rs749751887 Chr.14: 88011172-88011172 on Build GRCh38
rs753641390 Chr.14: 88011034-88011034 on Build GRCh38
rs1055468 Chr.14: 88011453-88011453 on Build GRCh38
rs764112131 Chr.14: 88011201-88011201 on Build GRCh38
rs775939991 Chr.14: 88010994-88010994 on Build GRCh38
rs1050983323 Chr.14: 88011227-88011227 on Build GRCh38
rs770245308 Chr.14: 88010986-88010986 on Build GRCh38
rs1004430544 Chr.14: 88011328-88011328 on Build GRCh38
rs771016297 Chr.14: 88011367-88011367 on Build GRCh38
rs761796311 Chr.14: 88011306-88011306 on Build GRCh38
rs755703665 Chr.14: 88011313-88011313 on Build GRCh38
rs746253097 Chr.14: 88010970-88010970 on Build GRCh38
rs760210365 Chr.14: 88011422-88011422 on Build GRCh38
rs868319581 Chr.14: 88010996-88010996 on Build GRCh38
rs201126670 Chr.14: 88011128-88011128 on Build GRCh38
rs754908890 Chr.14: 88010906-88010906 on Build GRCh38
rs777392229 Chr.14: 88010932-88010932 on Build GRCh38
rs920617864 Chr.14: 88010916-88010916 on Build GRCh38
rs779020059 Chr.14: 88010910-88010910 on Build GRCh38
rs780853135 Chr.14: 88011148-88011148 on Build GRCh38
rs747625763 Chr.14: 88010919-88010919 on Build GRCh38
rs762835121 Chr.14: 88011277-88011277 on Build GRCh38
rs139775422 Chr.14: 88011327-88011327 on Build GRCh38
rs769208743 Chr.14: 88011174-88011174 on Build GRCh38
rs753118671 Chr.14: 88011457-88011457 on Build GRCh38
rs781204922 Chr.14: 88011090-88011090 on Build GRCh38
rs181129859 Chr.14: 88011334-88011334 on Build GRCh38
rs1055465 Chr.14: 88011138-88011138 on Build GRCh38
rs761978597 Chr.14: 88011181-88011181 on Build GRCh38
rs780214420 Chr.14: 88011230-88011230 on Build GRCh38
rs755339728 Chr.14: 88011320-88011320 on Build GRCh38
rs756618557 Chr.14: 88010921-88010921 on Build GRCh38
rs752606649 Chr.14: 88011196-88011196 on Build GRCh38
rs748655974 Chr.14: 88011333-88011333 on Build GRCh38
rs1015581588 Chr.14: 88011031-88011031 on Build GRCh38
rs767453748 Chr.14: 88011411-88011411 on Build GRCh38
rs753507667 Chr.14: 88011444-88011444 on Build GRCh38
rs776879229 Chr.14: 88011371-88011371 on Build GRCh38
rs748264608 Chr.14: 88011123-88011127 on Build GRCh38
rs377442975 Chr.14: 88011185-88011185 on Build GRCh38
rs750989848 Chr.14: 88011210-88011210 on Build GRCh38
rs755830314 Chr.14: 88011219-88011219 on Build GRCh38
rs760532795 Chr.14: 88011190-88011190 on Build GRCh38
rs747102290 Chr.14: 88011356-88011356 on Build GRCh38
rs765848791 Chr.14: 88011427-88011427 on Build GRCh38
rs145362448 Chr.14: 88011400-88011400 on Build GRCh38
rs571054746 Chr.14: 88011258-88011258 on Build GRCh38
rs756852546 Chr.14: 88011133-88011133 on Build GRCh38
rs764064545 Chr.14: 88011004-88011004 on Build GRCh38
rs375417424 Chr.14: 88011212-88011212 on Build GRCh38
rs1027687236 Chr.14: 88010959-88010959 on Build GRCh38
rs867394370 Chr.14: 88011202-88011202 on Build GRCh38
rs925220030 Chr.14: 88011132-88011132 on Build GRCh38
rs745560553 Chr.14: 88011158-88011158 on Build GRCh38
rs775096144 Chr.14: 88011268-88011268 on Build GRCh38
rs267604075 Chr.14: 88011049-88011049 on Build GRCh38
rs200487695 Chr.14: 88011233-88011233 on Build GRCh38
rs139430517 Chr.14: 88010963-88010963 on Build GRCh38
rs755211561 Chr.14: 88010954-88010955 on Build GRCh38
rs548128364 Chr.14: 88011175-88011175 on Build GRCh38
rs1055467 Chr.14: 88011235-88011235 on Build GRCh38
rs757816094 Chr.14: 88010924-88010924 on Build GRCh38
rs566099533 Chr.14: 88011218-88011218 on Build GRCh38
rs772260251 Chr.14: 88011248-88011248 on Build GRCh38
rs773964361 Chr.14: 88010973-88010973 on Build GRCh38
rs916726612 Chr.14: 88011051-88011051 on Build GRCh38
rs6574978 Chr.14: 88011069-88011069 on Build GRCh38
rs767009794 Chr.14: 88011312-88011312 on Build GRCh38
rs139232553 Chr.14: 88011316-88011316 on Build GRCh38
rs761455094 Chr.14: 88011003-88011003 on Build GRCh38
rs1002366983 Chr.14: 88010937-88010937 on Build GRCh38
rs772662213 Chr.14: 88011413-88011413 on Build GRCh38
rs950683606 Chr.14: 88010945-88010945 on Build GRCh38
rs770829884 Chr.14: 88011211-88011211 on Build GRCh38
rs931847819 Chr.14: 88011127-88011127 on Build GRCh38
rs754648962 Chr.14: 88011451-88011451 on Build GRCh38
rs779780445 Chr.14: 88011314-88011314 on Build GRCh38
rs761876928 Chr.14: 88011009-88011009 on Build GRCh38
rs777935834 Chr.14: 88011335-88011335 on Build GRCh38
rs760977268 Chr.14: 88011261-88011261 on Build GRCh38
rs934967805 Chr.14: 88011215-88011215 on Build GRCh38
rs1055466 Chr.14: 88011220-88011220 on Build GRCh38
rs777146067 Chr.14: 88011100-88011100 on Build GRCh38
rs751488297 Chr.14: 88011285-88011285 on Build GRCh38
rs770118299 Chr.14: 88011159-88011159 on Build GRCh38
rs752555068 Chr.14: 88011097-88011097 on Build GRCh38
rs904937461 Chr.14: 88011281-88011281 on Build GRCh38
rs774080995 Chr.14: 88011408-88011408 on Build GRCh38
rs746629867 Chr.14: 88010934-88010934 on Build GRCh38
rs1055264317 Chr.14: 88011036-88011036 on Build GRCh38

View Full Product Details